Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3110037-3110542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA11
Entrez Gene ID
2767
Gene Name
G protein subunit alpha 11
Gene Aliases
FBH, FBH2, FHH2, GNA-11, HHC2, HYPOC2
UniGene
Hs.650575
Gene Chromosome Location
Chr.19: 3094410-3124002 on Build GRCh38
Transcripts
NM_002067.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4827188 c.168C>G p.I56M Chr.19: 3110180-3110180 on Build GRCh38
6960018 c.140C>T p.T47M Chr.19: 3110152-3110152 on Build GRCh38
6672106 c.162G>A p.T54T Chr.19: 3110174-3110174 on Build GRCh38
6232951 c.141G>T p.T47T Chr.19: 3110153-3110153 on Build GRCh38
6672105 c.303C>T p.Y101Y Chr.19: 3110315-3110315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921079769 Chr.19: 3110435-3110435 on Build GRCh38
rs762506413 Chr.19: 3110104-3110104 on Build GRCh38
rs866002209 Chr.19: 3110335-3110335 on Build GRCh38
rs750194467 Chr.19: 3110369-3110369 on Build GRCh38
rs74913870 Chr.19: 3110263-3110263 on Build GRCh38
rs546306678 Chr.19: 3110341-3110341 on Build GRCh38
rs587777021 Chr.19: 3110190-3110190 on Build GRCh38
rs1026750003 Chr.19: 3110381-3110381 on Build GRCh38
rs374969385 Chr.19: 3110121-3110121 on Build GRCh38
rs759067107 Chr.19: 3110162-3110162 on Build GRCh38
rs41276844 Chr.19: 3110346-3110346 on Build GRCh38
rs770729214 Chr.19: 3110246-3110246 on Build GRCh38
rs11085000 Chr.19: 3110351-3110351 on Build GRCh38
rs925026452 Chr.19: 3110477-3110477 on Build GRCh38
rs563944908 Chr.19: 3110140-3110140 on Build GRCh38
rs753693838 Chr.19: 3110286-3110286 on Build GRCh38
rs935057621 Chr.19: 3110421-3110421 on Build GRCh38
rs140301317 Chr.19: 3110229-3110229 on Build GRCh38
rs1051216341 Chr.19: 3110332-3110332 on Build GRCh38
rs147309216 Chr.19: 3110424-3110424 on Build GRCh38
rs760710011 Chr.19: 3110277-3110277 on Build GRCh38
rs143987260 Chr.19: 3110342-3110342 on Build GRCh38
rs7257368 Chr.19: 3110489-3110489 on Build GRCh38
rs975342398 Chr.19: 3110292-3110292 on Build GRCh38
rs765070403 Chr.19: 3110364-3110364 on Build GRCh38
rs373250050 Chr.19: 3110348-3110348 on Build GRCh38
rs760690456 Chr.19: 3110333-3110334 on Build GRCh38
rs933889965 Chr.19: 3110315-3110315 on Build GRCh38
rs1006287849 Chr.19: 3110418-3110418 on Build GRCh38
rs370357749 Chr.19: 3110357-3110357 on Build GRCh38
rs7255425 Chr.19: 3110100-3110100 on Build GRCh38
rs531924554 Chr.19: 3110498-3110498 on Build GRCh38
rs1004230306 Chr.19: 3110476-3110476 on Build GRCh38
rs756991534 Chr.19: 3110299-3110299 on Build GRCh38
rs775099798 Chr.19: 3110352-3110352 on Build GRCh38
rs754043777 Chr.19: 3110381-3110382 on Build GRCh38
rs749256854 Chr.19: 3110303-3110303 on Build GRCh38
rs576887084 Chr.19: 3110207-3110207 on Build GRCh38
rs377224539 Chr.19: 3110163-3110163 on Build GRCh38
rs571571444 Chr.19: 3110127-3110127 on Build GRCh38
rs149108190 Chr.19: 3110174-3110174 on Build GRCh38
rs779008056 Chr.19: 3110129-3110129 on Build GRCh38
rs776843648 Chr.19: 3110150-3110150 on Build GRCh38
rs769129067 Chr.19: 3110143-3110143 on Build GRCh38
rs755663461 Chr.19: 3110204-3110204 on Build GRCh38
rs776369531 Chr.19: 3110362-3110362 on Build GRCh38
rs975225827 Chr.19: 3110431-3110431 on Build GRCh38
rs200107023 Chr.19: 3110113-3110113 on Build GRCh38
rs964177853 Chr.19: 3110132-3110132 on Build GRCh38
rs142364701 Chr.19: 3110512-3110512 on Build GRCh38
rs933651595 Chr.19: 3110108-3110108 on Build GRCh38
rs777673683 Chr.19: 3110195-3110195 on Build GRCh38
rs941191833 Chr.19: 3110287-3110287 on Build GRCh38
rs557415284 Chr.19: 3110404-3110404 on Build GRCh38
rs955491239 Chr.19: 3110436-3110436 on Build GRCh38
rs374089777 Chr.19: 3110268-3110268 on Build GRCh38
rs995605325 Chr.19: 3110347-3110347 on Build GRCh38
rs147528229 Chr.19: 3110205-3110205 on Build GRCh38
rs755993488 Chr.19: 3110179-3110179 on Build GRCh38
rs767445318 Chr.19: 3110216-3110218 on Build GRCh38
rs928807739 Chr.19: 3110074-3110074 on Build GRCh38
rs61731117 Chr.19: 3110153-3110153 on Build GRCh38
rs978950936 Chr.19: 3110413-3110413 on Build GRCh38
rs150293267 Chr.19: 3110114-3110114 on Build GRCh38
rs34074026 Chr.19: 3110374-3110375 on Build GRCh38
rs777099166 Chr.19: 3110247-3110247 on Build GRCh38
rs565336062 Chr.19: 3110471-3110471 on Build GRCh38
rs764110617 Chr.19: 3110361-3110361 on Build GRCh38
rs773216969 Chr.19: 3110330-3110331 on Build GRCh38
rs762714189 Chr.19: 3110152-3110152 on Build GRCh38
rs941298170 Chr.19: 3110069-3110069 on Build GRCh38
rs61634148 Chr.19: 3110439-3110439 on Build GRCh38
rs775915105 Chr.19: 3110139-3110139 on Build GRCh38
rs762226968 Chr.19: 3110234-3110234 on Build GRCh38
rs947503474 Chr.19: 3110405-3110405 on Build GRCh38
rs938891774 Chr.19: 3110086-3110086 on Build GRCh38
rs369699960 Chr.19: 3110216-3110216 on Build GRCh38
rs1056360254 Chr.19: 3110097-3110097 on Build GRCh38
rs780228478 Chr.19: 3110338-3110338 on Build GRCh38
rs761804476 Chr.19: 3110216-3110216 on Build GRCh38
rs193178022 Chr.19: 3110419-3110419 on Build GRCh38
rs752079029 Chr.19: 3110145-3110145 on Build GRCh38
rs61731116 Chr.19: 3110201-3110201 on Build GRCh38
rs1026761665 Chr.19: 3110468-3110468 on Build GRCh38
rs766042469 Chr.19: 3110106-3110106 on Build GRCh38
rs774842486 Chr.19: 3110276-3110276 on Build GRCh38
rs904056028 Chr.19: 3110350-3110350 on Build GRCh38
rs140204057 Chr.19: 3110267-3110267 on Build GRCh38
rs368986120 Chr.19: 3110124-3110124 on Build GRCh38
rs528667271 Chr.19: 3110440-3110440 on Build GRCh38
rs964728772 Chr.19: 3110425-3110425 on Build GRCh38
rs772594536 Chr.19: 3110138-3110138 on Build GRCh38
rs758603549 Chr.19: 3110321-3110321 on Build GRCh38
rs763900975 Chr.19: 3110285-3110285 on Build GRCh38
rs773698583 Chr.19: 3110344-3110344 on Build GRCh38
rs769019097 Chr.19: 3110230-3110230 on Build GRCh38
rs7255519 Chr.19: 3110125-3110125 on Build GRCh38
rs1052126676 Chr.19: 3110422-3110422 on Build GRCh38
rs995780363 Chr.19: 3110462-3110462 on Build GRCh38
rs145033220 Chr.19: 3110252-3110252 on Build GRCh38
rs868681560 Chr.19: 3110209-3110209 on Build GRCh38
rs147570504 Chr.19: 3110156-3110156 on Build GRCh38
rs766579881 Chr.19: 3110334-3110335 on Build GRCh38
rs781234448 Chr.19: 3110119-3110119 on Build GRCh38
rs960279983 Chr.19: 3110497-3110497 on Build GRCh38
rs377441887 Chr.19: 3110506-3110506 on Build GRCh38
rs777092144 Chr.19: 3110232-3110232 on Build GRCh38
rs761054897 Chr.19: 3110099-3110099 on Build GRCh38
rs587777707 Chr.19: 3110191-3110191 on Build GRCh38
rs943975275 Chr.19: 3110433-3110433 on Build GRCh38
rs184614398 Chr.19: 3110466-3110466 on Build GRCh38
rs370558074 Chr.19: 3110343-3110343 on Build GRCh38
rs368095546 Chr.19: 3110101-3110101 on Build GRCh38
rs916041207 Chr.19: 3110220-3110220 on Build GRCh38
rs745912473 Chr.19: 3110133-3110133 on Build GRCh38
rs172613 Chr.19: 3110517-3110517 on Build GRCh38
rs41276846 Chr.19: 3110363-3110363 on Build GRCh38
rs992122997 Chr.19: 3110403-3110403 on Build GRCh38
rs750706655 Chr.19: 3110312-3110312 on Build GRCh38
rs947843389 Chr.19: 3110231-3110231 on Build GRCh38
rs755281796 Chr.19: 3110067-3110067 on Build GRCh38
rs371983140 Chr.19: 3110120-3110120 on Build GRCh38

View Full Product Details