Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3478345-3478846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM24
Entrez Gene ID
284422
Gene Name
small integral membrane protein 24
Gene Aliases
C19orf77, HSPC323
UniGene
Hs.130714
Gene Chromosome Location
Chr.19: 3474407-3480542 on Build GRCh38
Transcripts
NM_001136503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639446 c.175G>A p.A59T Chr.19: 3478822-3478822 on Build GRCh38
3422662 c.204G>A p.T68T Chr.19: 3478454-3478454 on Build GRCh38
6300913 c.204G>T p.T68T Chr.19: 3478454-3478454 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753989516 Chr.19: 3478792-3478792 on Build GRCh38
rs948240768 Chr.19: 3478433-3478433 on Build GRCh38
rs1010893453 Chr.19: 3478624-3478624 on Build GRCh38
rs1008713013 Chr.19: 3478545-3478545 on Build GRCh38
rs957064257 Chr.19: 3478582-3478587 on Build GRCh38
rs1032889937 Chr.19: 3478580-3478580 on Build GRCh38
rs146333097 Chr.19: 3478409-3478409 on Build GRCh38
rs767435632 Chr.19: 3478480-3478480 on Build GRCh38
rs762503020 Chr.19: 3478462-3478462 on Build GRCh38
rs532273268 Chr.19: 3478706-3478706 on Build GRCh38
rs887817004 Chr.19: 3478426-3478426 on Build GRCh38
rs764328345 Chr.19: 3478769-3478769 on Build GRCh38
rs10413312 Chr.19: 3478648-3478648 on Build GRCh38
rs780138726 Chr.19: 3478722-3478722 on Build GRCh38
rs759540118 Chr.19: 3478477-3478477 on Build GRCh38
rs778961545 Chr.19: 3478800-3478800 on Build GRCh38
rs574346801 Chr.19: 3478546-3478546 on Build GRCh38
rs796377093 Chr.19: 3478471-3478471 on Build GRCh38
rs779714096 Chr.19: 3478773-3478773 on Build GRCh38
rs932360486 Chr.19: 3478674-3478674 on Build GRCh38
rs756322863 Chr.19: 3478527-3478527 on Build GRCh38
rs989525465 Chr.19: 3478412-3478412 on Build GRCh38
rs957254872 Chr.19: 3478371-3478371 on Build GRCh38
rs1045122213 Chr.19: 3478808-3478808 on Build GRCh38
rs746435117 Chr.19: 3478801-3478801 on Build GRCh38
rs940713171 Chr.19: 3478741-3478741 on Build GRCh38
rs562640477 Chr.19: 3478733-3478733 on Build GRCh38
rs36076842 Chr.19: 3478630-3478631 on Build GRCh38
rs192284606 Chr.19: 3478394-3478394 on Build GRCh38
rs972313562 Chr.19: 3478690-3478690 on Build GRCh38
rs563246320 Chr.19: 3478719-3478719 on Build GRCh38
rs928137577 Chr.19: 3478664-3478664 on Build GRCh38
rs556308017 Chr.19: 3478450-3478450 on Build GRCh38
rs999185008 Chr.19: 3478586-3478586 on Build GRCh38
rs1050111915 Chr.19: 3478745-3478745 on Build GRCh38
rs928220796 Chr.19: 3478454-3478454 on Build GRCh38
rs747767618 Chr.19: 3478443-3478443 on Build GRCh38
rs200625720 Chr.19: 3478469-3478469 on Build GRCh38
rs542074058 Chr.19: 3478488-3478488 on Build GRCh38
rs1036746322 Chr.19: 3478767-3478767 on Build GRCh38
rs1024118886 Chr.19: 3478511-3478511 on Build GRCh38
rs768486524 Chr.19: 3478796-3478797 on Build GRCh38
rs988850223 Chr.19: 3478604-3478604 on Build GRCh38
rs868090989 Chr.19: 3478583-3478583 on Build GRCh38
rs888955667 Chr.19: 3478368-3478368 on Build GRCh38
rs539631539 Chr.19: 3478421-3478421 on Build GRCh38
rs556025793 Chr.19: 3478510-3478510 on Build GRCh38
rs1050590803 Chr.19: 3478796-3478796 on Build GRCh38
rs183891325 Chr.19: 3478616-3478616 on Build GRCh38
rs767466796 Chr.19: 3478403-3478403 on Build GRCh38
rs765964109 Chr.19: 3478468-3478468 on Build GRCh38
rs531609618 Chr.19: 3478369-3478369 on Build GRCh38
rs534204799 Chr.19: 3478406-3478406 on Build GRCh38
rs958027291 Chr.19: 3478619-3478619 on Build GRCh38
rs969902744 Chr.19: 3478642-3478642 on Build GRCh38
rs145723843 Chr.19: 3478458-3478458 on Build GRCh38
rs757371278 Chr.19: 3478795-3478795 on Build GRCh38
rs978094309 Chr.19: 3478405-3478405 on Build GRCh38
rs932134681 Chr.19: 3478712-3478712 on Build GRCh38
rs746347159 Chr.19: 3478395-3478395 on Build GRCh38
rs548518838 Chr.19: 3478375-3478375 on Build GRCh38
rs531545250 Chr.19: 3478762-3478762 on Build GRCh38
rs1023665180 Chr.19: 3478626-3478626 on Build GRCh38
rs905693789 Chr.19: 3478530-3478530 on Build GRCh38
rs758842538 Chr.19: 3478802-3478802 on Build GRCh38
rs56801248 Chr.19: 3478420-3478420 on Build GRCh38
rs574674330 Chr.19: 3478518-3478518 on Build GRCh38
rs903070375 Chr.19: 3478547-3478547 on Build GRCh38
rs548458768 Chr.19: 3478772-3478772 on Build GRCh38
rs749049385 Chr.19: 3478775-3478801 on Build GRCh38

View Full Product Details