Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3480319-3480815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM24
Entrez Gene ID
284422
Gene Name
small integral membrane protein 24
Gene Aliases
C19orf77, HSPC323
UniGene
Hs.130714
Gene Chromosome Location
Chr.19: 3474407-3480542 on Build GRCh38
Transcripts
NM_001136503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4491409 c.37C>T p.L13L Chr.19: 3480427-3480427 on Build GRCh38
5504435 c.9C>T p.T3T Chr.19: 3480455-3480455 on Build GRCh38
4501867 c.59C>T p.A20V Chr.19: 3480405-3480405 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765317526 Chr.19: 3480415-3480415 on Build GRCh38
rs1031261037 Chr.19: 3480470-3480470 on Build GRCh38
rs934260104 Chr.19: 3480726-3480726 on Build GRCh38
rs950426482 Chr.19: 3480355-3480355 on Build GRCh38
rs750433493 Chr.19: 3480417-3480417 on Build GRCh38
rs1011232109 Chr.19: 3480496-3480496 on Build GRCh38
rs955703997 Chr.19: 3480474-3480474 on Build GRCh38
rs755411138 Chr.19: 3480493-3480493 on Build GRCh38
rs959026511 Chr.19: 3480606-3480606 on Build GRCh38
rs889831820 Chr.19: 3480758-3480758 on Build GRCh38
rs541271251 Chr.19: 3480440-3480440 on Build GRCh38
rs1038467233 Chr.19: 3480767-3480767 on Build GRCh38
rs184425863 Chr.19: 3480345-3480345 on Build GRCh38
rs775169894 Chr.19: 3480365-3480365 on Build GRCh38
rs563419821 Chr.19: 3480574-3480574 on Build GRCh38
rs933999451 Chr.19: 3480495-3480495 on Build GRCh38
rs868608894 Chr.19: 3480775-3480775 on Build GRCh38
rs887887970 Chr.19: 3480383-3480383 on Build GRCh38
rs969857675 Chr.19: 3480531-3480531 on Build GRCh38
rs549226662 Chr.19: 3480696-3480696 on Build GRCh38
rs560123574 Chr.19: 3480381-3480382 on Build GRCh38
rs760828944 Chr.19: 3480513-3480514 on Build GRCh38
rs1051121152 Chr.19: 3480508-3480508 on Build GRCh38
rs942639498 Chr.19: 3480761-3480761 on Build GRCh38
rs997787212 Chr.19: 3480456-3480456 on Build GRCh38
rs914939464 Chr.19: 3480632-3480632 on Build GRCh38
rs1021317089 Chr.19: 3480502-3480502 on Build GRCh38
rs978644557 Chr.19: 3480675-3480675 on Build GRCh38
rs889779365 Chr.19: 3480530-3480530 on Build GRCh38
rs34210841 Chr.19: 3480452-3480453 on Build GRCh38
rs902737637 Chr.19: 3480481-3480482 on Build GRCh38
rs950658576 Chr.19: 3480575-3480575 on Build GRCh38
rs981981153 Chr.19: 3480579-3480579 on Build GRCh38
rs759552464 Chr.19: 3480350-3480350 on Build GRCh38
rs2074961 Chr.19: 3480360-3480360 on Build GRCh38
rs1017433289 Chr.19: 3480548-3480548 on Build GRCh38
rs572154269 Chr.19: 3480412-3480412 on Build GRCh38
rs972672393 Chr.19: 3480571-3480571 on Build GRCh38
rs902067049 Chr.19: 3480422-3480422 on Build GRCh38
rs760568265 Chr.19: 3480381-3480381 on Build GRCh38
rs929277857 Chr.19: 3480790-3480790 on Build GRCh38
rs754130663 Chr.19: 3480405-3480405 on Build GRCh38
rs946833056 Chr.19: 3480455-3480455 on Build GRCh38
rs141469689 Chr.19: 3480580-3480592 on Build GRCh38
rs773105608 Chr.19: 3480475-3480476 on Build GRCh38
rs780558113 Chr.19: 3480450-3480450 on Build GRCh38
rs946920767 Chr.19: 3480674-3480674 on Build GRCh38
rs1039222960 Chr.19: 3480389-3480389 on Build GRCh38
rs564084227 Chr.19: 3480547-3480547 on Build GRCh38
rs748097851 Chr.19: 3480353-3480354 on Build GRCh38
rs960189103 Chr.19: 3480568-3480568 on Build GRCh38
rs1008120078 Chr.19: 3480386-3480386 on Build GRCh38
rs1026248327 Chr.19: 3480572-3480572 on Build GRCh38
rs995049834 Chr.19: 3480564-3480564 on Build GRCh38
rs529360113 Chr.19: 3480665-3480665 on Build GRCh38
rs751986806 Chr.19: 3480481-3480481 on Build GRCh38
rs1049164516 Chr.19: 3480378-3480378 on Build GRCh38
rs919133921 Chr.19: 3480349-3480349 on Build GRCh38
rs924116459 Chr.19: 3480681-3480681 on Build GRCh38
rs549796546 Chr.19: 3480565-3480565 on Build GRCh38
rs981717977 Chr.19: 3480360-3480360 on Build GRCh38
rs1034680298 Chr.19: 3480604-3480604 on Build GRCh38
rs553124781 Chr.19: 3480739-3480739 on Build GRCh38
rs533145681 Chr.19: 3480549-3480549 on Build GRCh38
rs566223852 Chr.19: 3480762-3480762 on Build GRCh38

View Full Product Details