Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3526167-3526659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZR1
Entrez Gene ID
51343
Gene Name
fizzy/cell division cycle 20 related 1
Gene Aliases
CDC20C, CDH1, FZR, FZR2, HCDH, HCDH1
UniGene
Hs.413133
Gene Chromosome Location
Chr.19: 3506115-3536757 on Build GRCh38
Transcripts
NM_001136197.1 NM_001136198.1 NM_016263.3 XM_005259573.4 XM_017026863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3532355 c.252C>A p.N84K Chr.19: 3526176-3526176 on Build GRCh38
7318222 c.372G>C p.K124N Chr.19: 3526371-3526371 on Build GRCh38
994817 c.280_284delCTGCT p.L94fs*3 Chr.19: 3526279-3526283 on Build GRCh38
4823145 c.313G>A p.E105K Chr.19: 3526312-3526312 on Build GRCh38
4823144 c.313G>A p.E105K Chr.19: 3526312-3526312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001543649 Chr.19: 3526599-3526599 on Build GRCh38
rs182059666 Chr.19: 3526278-3526278 on Build GRCh38
rs922495793 Chr.19: 3526446-3526446 on Build GRCh38
rs773652238 Chr.19: 3526346-3526346 on Build GRCh38
rs373146649 Chr.19: 3526343-3526343 on Build GRCh38
rs771849722 Chr.19: 3526221-3526221 on Build GRCh38
rs567376375 Chr.19: 3526361-3526361 on Build GRCh38
rs763499825 Chr.19: 3526350-3526350 on Build GRCh38
rs200856412 Chr.19: 3526341-3526341 on Build GRCh38
rs202225112 Chr.19: 3526411-3526411 on Build GRCh38
rs761842369 Chr.19: 3526344-3526344 on Build GRCh38
rs749255706 Chr.19: 3526207-3526207 on Build GRCh38
rs777457646 Chr.19: 3526261-3526261 on Build GRCh38
rs760294838 Chr.19: 3526397-3526397 on Build GRCh38
rs756770945 Chr.19: 3526281-3526281 on Build GRCh38
rs947218725 Chr.19: 3526605-3526605 on Build GRCh38
rs922938357 Chr.19: 3526536-3526536 on Build GRCh38
rs765708188 Chr.19: 3526234-3526234 on Build GRCh38
rs760425900 Chr.19: 3526362-3526362 on Build GRCh38
rs538601576 Chr.19: 3526420-3526420 on Build GRCh38
rs751978825 Chr.19: 3526598-3526598 on Build GRCh38
rs891620815 Chr.19: 3526355-3526355 on Build GRCh38
rs564919780 Chr.19: 3526522-3526522 on Build GRCh38
rs202047625 Chr.19: 3526499-3526503 on Build GRCh38
rs201859345 Chr.19: 3526219-3526219 on Build GRCh38
rs558538410 Chr.19: 3526438-3526438 on Build GRCh38
rs560780880 Chr.19: 3526637-3526638 on Build GRCh38
rs778781973 Chr.19: 3526290-3526290 on Build GRCh38
rs750701371 Chr.19: 3526199-3526199 on Build GRCh38
rs746851865 Chr.19: 3526410-3526410 on Build GRCh38
rs540841451 Chr.19: 3526481-3526481 on Build GRCh38
rs1040955443 Chr.19: 3526441-3526441 on Build GRCh38
rs374922648 Chr.19: 3526255-3526255 on Build GRCh38
rs199976578 Chr.19: 3526203-3526203 on Build GRCh38
rs763704238 Chr.19: 3526385-3526385 on Build GRCh38
rs367591096 Chr.19: 3526305-3526305 on Build GRCh38
rs770734196 Chr.19: 3526208-3526208 on Build GRCh38
rs762401075 Chr.19: 3526233-3526233 on Build GRCh38
rs188982032 Chr.19: 3526515-3526515 on Build GRCh38
rs537998539 Chr.19: 3526447-3526447 on Build GRCh38
rs144128556 Chr.19: 3526631-3526631 on Build GRCh38
rs888207647 Chr.19: 3526475-3526475 on Build GRCh38
rs1024339954 Chr.19: 3526269-3526269 on Build GRCh38
rs771195275 Chr.19: 3526230-3526230 on Build GRCh38
rs200223586 Chr.19: 3526406-3526406 on Build GRCh38
rs980074962 Chr.19: 3526392-3526392 on Build GRCh38
rs185869982 Chr.19: 3526474-3526474 on Build GRCh38
rs985381245 Chr.19: 3526488-3526488 on Build GRCh38
rs764161476 Chr.19: 3526227-3526227 on Build GRCh38
rs529984678 Chr.19: 3526396-3526396 on Build GRCh38
rs778426775 Chr.19: 3526398-3526398 on Build GRCh38
rs370952568 Chr.19: 3526260-3526260 on Build GRCh38
rs577619430 Chr.19: 3526514-3526514 on Build GRCh38
rs539999401 Chr.19: 3526483-3526483 on Build GRCh38
rs753524046 Chr.19: 3526394-3526394 on Build GRCh38
rs752413539 Chr.19: 3526256-3526256 on Build GRCh38
rs373811234 Chr.19: 3526519-3526519 on Build GRCh38
rs200805373 Chr.19: 3526200-3526200 on Build GRCh38
rs769917168 Chr.19: 3526345-3526345 on Build GRCh38
rs913733697 Chr.19: 3526419-3526419 on Build GRCh38
rs771866008 Chr.19: 3526307-3526307 on Build GRCh38
rs530597525 Chr.19: 3526569-3526569 on Build GRCh38
rs774219644 Chr.19: 3526213-3526213 on Build GRCh38
rs777361963 Chr.19: 3526206-3526206 on Build GRCh38
rs569002775 Chr.19: 3526439-3526439 on Build GRCh38
rs1014807520 Chr.19: 3526509-3526509 on Build GRCh38
rs765662263 Chr.19: 3526198-3526198 on Build GRCh38
rs2286544 Chr.19: 3526500-3526500 on Build GRCh38
rs905717306 Chr.19: 3526628-3526628 on Build GRCh38
rs755746236 Chr.19: 3526204-3526204 on Build GRCh38
rs768815657 Chr.19: 3526329-3526329 on Build GRCh38
rs750913737 Chr.19: 3526236-3526236 on Build GRCh38
rs1042897593 Chr.19: 3526607-3526607 on Build GRCh38
rs371682300 Chr.19: 3526202-3526202 on Build GRCh38
rs538723834 Chr.19: 3526354-3526354 on Build GRCh38
rs748392905 Chr.19: 3526415-3526415 on Build GRCh38
rs779756787 Chr.19: 3526308-3526308 on Build GRCh38
rs760827528 Chr.19: 3526225-3526225 on Build GRCh38
rs933066748 Chr.19: 3526264-3526264 on Build GRCh38
rs2286543 Chr.19: 3526454-3526454 on Build GRCh38
rs776530629 Chr.19: 3526228-3526228 on Build GRCh38
rs766648499 Chr.19: 3526547-3526547 on Build GRCh38
rs758760362 Chr.19: 3526254-3526254 on Build GRCh38
rs775745495 Chr.19: 3526223-3526223 on Build GRCh38
rs753239356 Chr.19: 3526262-3526262 on Build GRCh38
rs957726358 Chr.19: 3526624-3526624 on Build GRCh38
rs988546846 Chr.19: 3526597-3526597 on Build GRCh38
rs747260291 Chr.19: 3526328-3526328 on Build GRCh38

View Full Product Details