Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3613754-3614251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACTIN
Entrez Gene ID
58509
Gene Name
cactin, spliceosome C complex subunit
Gene Aliases
C19orf29, NY-REN-24, fSAPc
UniGene
Hs.128425
Gene Chromosome Location
Chr.19: 3610628-3626815 on Build GRCh38
Transcripts
NM_001080543.1 NM_021231.1 XM_011528160.2 NR_038865.1

Gene Symbol
CACTIN-AS1
Entrez Gene ID
404665
Gene Name
CACTIN antisense RNA 1
Gene Aliases
C19orf29-AS1, C19orf29OS
UniGene
Hs.719726
Gene Chromosome Location
Chr.19: 3607247-3613930 on Build GRCh38
Transcripts
NM_001080543.1 NM_021231.1 XM_011528160.2 NR_038865.1

SNP Detail

SNP ID Chromosome Location
rs530242120 Chr.19: 3614035-3614035 on Build GRCh38
rs1035492062 Chr.19: 3614061-3614061 on Build GRCh38
rs984798637 Chr.19: 3614111-3614111 on Build GRCh38
rs959751523 Chr.19: 3613991-3613991 on Build GRCh38
rs943539348 Chr.19: 3613975-3613975 on Build GRCh38
rs78516459 Chr.19: 3613977-3613977 on Build GRCh38
rs544321919 Chr.19: 3613988-3613988 on Build GRCh38
rs552779545 Chr.19: 3614000-3614000 on Build GRCh38
rs1055073685 Chr.19: 3613788-3613788 on Build GRCh38
rs950770986 Chr.19: 3614100-3614100 on Build GRCh38
rs995061136 Chr.19: 3614152-3614152 on Build GRCh38
rs973041234 Chr.19: 3614097-3614097 on Build GRCh38
rs78259786 Chr.19: 3613959-3613959 on Build GRCh38
rs1035524879 Chr.19: 3613927-3613927 on Build GRCh38
rs574285938 Chr.19: 3614044-3614045 on Build GRCh38
rs919225586 Chr.19: 3613938-3613938 on Build GRCh38
rs1011275164 Chr.19: 3614023-3614023 on Build GRCh38
rs75091789 Chr.19: 3613935-3613935 on Build GRCh38
rs997782280 Chr.19: 3613819-3613819 on Build GRCh38
rs567326681 Chr.19: 3614219-3614219 on Build GRCh38
rs943131636 Chr.19: 3614110-3614110 on Build GRCh38
rs902144584 Chr.19: 3613777-3613777 on Build GRCh38
rs559024571 Chr.19: 3613895-3613895 on Build GRCh38
rs1021381775 Chr.19: 3614024-3614024 on Build GRCh38
rs938046045 Chr.19: 3613786-3613786 on Build GRCh38
rs905539090 Chr.19: 3614226-3614226 on Build GRCh38
rs547123665 Chr.19: 3614065-3614065 on Build GRCh38
rs537177282 Chr.19: 3613831-3613831 on Build GRCh38
rs150313709 Chr.19: 3613980-3613980 on Build GRCh38
rs1007212862 Chr.19: 3613900-3613900 on Build GRCh38
rs183986312 Chr.19: 3614189-3614189 on Build GRCh38
rs979970959 Chr.19: 3613904-3613904 on Build GRCh38
rs978624680 Chr.19: 3614225-3614225 on Build GRCh38
rs557069084 Chr.19: 3613886-3613886 on Build GRCh38
rs114844570 Chr.19: 3613897-3613897 on Build GRCh38
rs1025586540 Chr.19: 3614098-3614098 on Build GRCh38
rs915817540 Chr.19: 3613795-3613799 on Build GRCh38
rs151182741 Chr.19: 3613812-3613812 on Build GRCh38
rs768623508 Chr.19: 3613989-3613989 on Build GRCh38
rs1022219338 Chr.19: 3614218-3614218 on Build GRCh38
rs146118431 Chr.19: 3613856-3613857 on Build GRCh38
rs773298663 Chr.19: 3614198-3614198 on Build GRCh38
rs368743438 Chr.19: 3614224-3614224 on Build GRCh38
rs964733514 Chr.19: 3614132-3614132 on Build GRCh38
rs1001259480 Chr.19: 3614231-3614231 on Build GRCh38
rs1003702880 Chr.19: 3614208-3614208 on Build GRCh38
rs1009802542 Chr.19: 3614020-3614031 on Build GRCh38
rs532880600 Chr.19: 3614125-3614125 on Build GRCh38
rs947628467 Chr.19: 3614217-3614217 on Build GRCh38
rs2074791 Chr.19: 3614138-3614138 on Build GRCh38
rs891437914 Chr.19: 3614022-3614022 on Build GRCh38
rs934529480 Chr.19: 3614051-3614051 on Build GRCh38
rs570120585 Chr.19: 3614215-3614215 on Build GRCh38
rs1001292126 Chr.19: 3614058-3614058 on Build GRCh38
rs560516653 Chr.19: 3614066-3614066 on Build GRCh38
rs762417464 Chr.19: 3614121-3614121 on Build GRCh38
rs903019577 Chr.19: 3613893-3613893 on Build GRCh38
rs902028435 Chr.19: 3613979-3613979 on Build GRCh38
rs537012567 Chr.19: 3614207-3614207 on Build GRCh38
rs891473824 Chr.19: 3613877-3613877 on Build GRCh38
rs909337814 Chr.19: 3613965-3613965 on Build GRCh38
rs1031721342 Chr.19: 3613854-3613854 on Build GRCh38
rs933540809 Chr.19: 3613999-3613999 on Build GRCh38
rs140319021 Chr.19: 3613957-3613957 on Build GRCh38
rs866101847 Chr.19: 3613931-3613931 on Build GRCh38
rs146074848 Chr.19: 3614209-3614224 on Build GRCh38

View Full Product Details