Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12938508-12939016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALR
Entrez Gene ID
811
Gene Name
calreticulin
Gene Aliases
CRT, HEL-S-99n, RO, SSA, cC1qR
UniGene
Hs.515162
Gene Chromosome Location
Chr.19: 12938600-12944490 on Build GRCh38
Transcripts
NM_004343.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7235806 c.19C>T p.L7L Chr.19: 12938698-12938698 on Build GRCh38
5854726 c.91G>A p.D31N Chr.19: 12938770-12938770 on Build GRCh38
6986596 c.37G>C p.G13R Chr.19: 12938716-12938716 on Build GRCh38
7304075 c.34C>T p.L12F Chr.19: 12938713-12938713 on Build GRCh38
4850372 c.63C>G p.V21V Chr.19: 12938742-12938742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984506499 Chr.19: 12938595-12938595 on Build GRCh38
rs563832075 Chr.19: 12938608-12938608 on Build GRCh38
rs559441200 Chr.19: 12938838-12938838 on Build GRCh38
rs190251647 Chr.19: 12938676-12938676 on Build GRCh38
rs28365950 Chr.19: 12938601-12938601 on Build GRCh38
rs930257841 Chr.19: 12938591-12938592 on Build GRCh38
rs761931801 Chr.19: 12938654-12938654 on Build GRCh38
rs371058066 Chr.19: 12938587-12938587 on Build GRCh38
rs878872782 Chr.19: 12938575-12938575 on Build GRCh38
rs541868908 Chr.19: 12938950-12938950 on Build GRCh38
rs991460675 Chr.19: 12938889-12938889 on Build GRCh38
rs147368353 Chr.19: 12938729-12938729 on Build GRCh38
rs775373485 Chr.19: 12938807-12938807 on Build GRCh38
rs777056556 Chr.19: 12938646-12938646 on Build GRCh38
rs1018609569 Chr.19: 12938650-12938650 on Build GRCh38
rs750975499 Chr.19: 12938991-12938991 on Build GRCh38
rs749886719 Chr.19: 12938776-12938776 on Build GRCh38
rs748690113 Chr.19: 12938637-12938637 on Build GRCh38
rs945116541 Chr.19: 12938607-12938607 on Build GRCh38
rs982787079 Chr.19: 12938578-12938578 on Build GRCh38
rs1026372838 Chr.19: 12938628-12938628 on Build GRCh38
rs903070949 Chr.19: 12938629-12938629 on Build GRCh38
rs767448972 Chr.19: 12938664-12938665 on Build GRCh38
rs1005382801 Chr.19: 12938623-12938623 on Build GRCh38
rs941005562 Chr.19: 12938907-12938907 on Build GRCh38
rs780662640 Chr.19: 12938696-12938696 on Build GRCh38
rs750356587 Chr.19: 12938588-12938588 on Build GRCh38
rs774308071 Chr.19: 12938801-12938801 on Build GRCh38
rs371782062 Chr.19: 12938784-12938784 on Build GRCh38
rs977520717 Chr.19: 12938915-12938915 on Build GRCh38
rs760508798 Chr.19: 12938809-12938809 on Build GRCh38
rs763780070 Chr.19: 12938813-12938813 on Build GRCh38
rs769124051 Chr.19: 12938643-12938643 on Build GRCh38
rs541091872 Chr.19: 12938751-12938751 on Build GRCh38
rs770254686 Chr.19: 12938790-12938790 on Build GRCh38
rs765974111 Chr.19: 12938626-12938626 on Build GRCh38
rs924451631 Chr.19: 12938785-12938785 on Build GRCh38
rs771108945 Chr.19: 12938799-12938799 on Build GRCh38
rs966897395 Chr.19: 12938853-12938853 on Build GRCh38
rs772657847 Chr.19: 12938638-12938638 on Build GRCh38
rs889694246 Chr.19: 12938621-12938621 on Build GRCh38
rs993655300 Chr.19: 12938579-12938579 on Build GRCh38
rs539393451 Chr.19: 12938719-12938719 on Build GRCh38
rs764890684 Chr.19: 12938818-12938818 on Build GRCh38
rs978268274 Chr.19: 12938859-12938859 on Build GRCh38
rs866220461 Chr.19: 12938691-12938691 on Build GRCh38
rs759562231 Chr.19: 12938802-12938802 on Build GRCh38
rs767385998 Chr.19: 12938724-12938724 on Build GRCh38
rs1034371074 Chr.19: 12938634-12938634 on Build GRCh38
rs955145749 Chr.19: 12938880-12938880 on Build GRCh38
rs774334892 Chr.19: 12938635-12938635 on Build GRCh38
rs11547567 Chr.19: 12938659-12938659 on Build GRCh38
rs777358098 Chr.19: 12938687-12938687 on Build GRCh38
rs756852999 Chr.19: 12938761-12938761 on Build GRCh38
rs766369235 Chr.19: 12938665-12938665 on Build GRCh38
rs772027359 Chr.19: 12938805-12938805 on Build GRCh38
rs750708082 Chr.19: 12938815-12938816 on Build GRCh38
rs574728641 Chr.19: 12938897-12938897 on Build GRCh38
rs370625713 Chr.19: 12938868-12938868 on Build GRCh38
rs747461787 Chr.19: 12938697-12938697 on Build GRCh38
rs778287096 Chr.19: 12938763-12938763 on Build GRCh38
rs763131258 Chr.19: 12938713-12938713 on Build GRCh38
rs755687463 Chr.19: 12938633-12938633 on Build GRCh38
rs775824052 Chr.19: 12938639-12938639 on Build GRCh38
rs201581541 Chr.19: 12938698-12938698 on Build GRCh38
rs748461962 Chr.19: 12938709-12938709 on Build GRCh38
rs1038162176 Chr.19: 12938624-12938624 on Build GRCh38
rs374628652 Chr.19: 12938632-12938632 on Build GRCh38
rs559305924 Chr.19: 12938548-12938548 on Build GRCh38
rs1025828547 Chr.19: 12938667-12938667 on Build GRCh38
rs749926861 Chr.19: 12938820-12938820 on Build GRCh38
rs781690273 Chr.19: 12938706-12938706 on Build GRCh38
rs762890708 Chr.19: 12938664-12938664 on Build GRCh38
rs751419557 Chr.19: 12938669-12938669 on Build GRCh38
rs531094913 Chr.19: 12938625-12938625 on Build GRCh38
rs774405017 Chr.19: 12938718-12938718 on Build GRCh38
rs1016386672 Chr.19: 12938553-12938553 on Build GRCh38
rs958953477 Chr.19: 12938888-12938888 on Build GRCh38
rs41300080 Chr.19: 12938620-12938620 on Build GRCh38
rs985580958 Chr.19: 12938884-12938884 on Build GRCh38
rs976950498 Chr.19: 12938545-12938545 on Build GRCh38
rs541155081 Chr.19: 12938533-12938533 on Build GRCh38
rs993242043 Chr.19: 12938821-12938821 on Build GRCh38
rs761916531 Chr.19: 12938870-12938870 on Build GRCh38
rs752512916 Chr.19: 12938728-12938728 on Build GRCh38
rs776407451 Chr.19: 12938815-12938815 on Build GRCh38
rs762118641 Chr.19: 12938653-12938653 on Build GRCh38
rs926865745 Chr.19: 12938586-12938586 on Build GRCh38
rs753422007 Chr.19: 12938688-12938688 on Build GRCh38
rs546755913 Chr.19: 12938647-12938647 on Build GRCh38
rs868551669 Chr.19: 12938611-12938611 on Build GRCh38
rs370935055 Chr.19: 12938671-12938671 on Build GRCh38
rs761380506 Chr.19: 12938816-12938816 on Build GRCh38
rs530055503 Chr.19: 12938576-12938576 on Build GRCh38
rs910888797 Chr.19: 12938896-12938896 on Build GRCh38
rs534687510 Chr.19: 12938636-12938636 on Build GRCh38
rs933676459 Chr.19: 12938673-12938673 on Build GRCh38
rs924723480 Chr.19: 12938916-12938916 on Build GRCh38
rs1026739986 Chr.19: 12938573-12938573 on Build GRCh38
rs770160684 Chr.19: 12938710-12938710 on Build GRCh38
rs756694331 Chr.19: 12938695-12938695 on Build GRCh38
rs755777417 Chr.19: 12938678-12938678 on Build GRCh38
rs1022525830 Chr.19: 12938834-12938834 on Build GRCh38
rs918487805 Chr.19: 12938945-12938945 on Build GRCh38
rs929822208 Chr.19: 12938978-12938978 on Build GRCh38
rs993491729 Chr.19: 12938561-12938561 on Build GRCh38
rs755559062 Chr.19: 12938780-12938780 on Build GRCh38
rs1055878255 Chr.19: 12938604-12938604 on Build GRCh38
rs368501056 Chr.19: 12938712-12938712 on Build GRCh38
rs749459501 Chr.19: 12938797-12938797 on Build GRCh38
rs1045566420 Chr.19: 12938980-12938980 on Build GRCh38
rs929780860 Chr.19: 12938716-12938716 on Build GRCh38
rs765485261 Chr.19: 12938655-12938655 on Build GRCh38
rs909868306 Chr.19: 12938600-12938600 on Build GRCh38
rs1029868234 Chr.19: 12938866-12938866 on Build GRCh38
rs572735736 Chr.19: 12938742-12938742 on Build GRCh38
rs922280180 Chr.19: 12938613-12938613 on Build GRCh38
rs747444683 Chr.19: 12938641-12938641 on Build GRCh38
rs1054223858 Chr.19: 12938935-12938935 on Build GRCh38
rs748557624 Chr.19: 12938788-12938788 on Build GRCh38
rs941288230 Chr.19: 12938701-12938701 on Build GRCh38
rs973971403 Chr.19: 12938913-12938913 on Build GRCh38
rs778107764 Chr.19: 12938796-12938796 on Build GRCh38
rs770893383 Chr.19: 12938715-12938715 on Build GRCh38
rs933642918 Chr.19: 12938615-12938615 on Build GRCh38
rs552795097 Chr.19: 12938631-12938631 on Build GRCh38
rs775581620 Chr.19: 12938731-12938731 on Build GRCh38

View Full Product Details