Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43969859-43970349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH2
Entrez Gene ID
1802
Gene Name
DPH2 homolog
Gene Aliases
DPH2L2
UniGene
Hs.632398
Gene Chromosome Location
Chr.1: 43969979-43973371 on Build GRCh38
Transcripts
NM_001039589.1 NM_001319165.1 NM_001319166.1 NM_001319167.1 NM_001319168.1 NM_001319169.1 NM_001319170.1 NM_001319171.1 NM_001384.4 XM_017000502.1 XM_017000503.1 XM_017000504.1 XM_017000505.1 XM_017000506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
357149 c.42G>T p.Q14H Chr.1: 43970217-43970217 on Build GRCh38
5509690 c.70A>G p.T24A Chr.1: 43970245-43970245 on Build GRCh38
7147194 c.44G>A p.R15Q Chr.1: 43970219-43970219 on Build GRCh38
5491100 c.147+4A>T p.? Chr.1: 43970326-43970326 on Build GRCh38
1560603 c.8C>T p.S3L Chr.1: 43970183-43970183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553083384 Chr.1: 43970022-43970022 on Build GRCh38
rs11539398 Chr.1: 43970211-43970211 on Build GRCh38
rs535186172 Chr.1: 43970051-43970051 on Build GRCh38
rs201622210 Chr.1: 43970220-43970220 on Build GRCh38
rs764794496 Chr.1: 43970210-43970210 on Build GRCh38
rs772911342 Chr.1: 43970290-43970290 on Build GRCh38
rs371907827 Chr.1: 43969926-43969926 on Build GRCh38
rs952529160 Chr.1: 43970003-43970003 on Build GRCh38
rs192927657 Chr.1: 43969997-43969997 on Build GRCh38
rs138358323 Chr.1: 43970226-43970226 on Build GRCh38
rs921394758 Chr.1: 43970094-43970094 on Build GRCh38
rs772218135 Chr.1: 43970251-43970251 on Build GRCh38
rs764044865 Chr.1: 43970196-43970196 on Build GRCh38
rs778233301 Chr.1: 43970133-43970133 on Build GRCh38
rs775168381 Chr.1: 43970128-43970133 on Build GRCh38
rs539455304 Chr.1: 43970021-43970021 on Build GRCh38
rs934805192 Chr.1: 43970124-43970124 on Build GRCh38
rs545963890 Chr.1: 43969903-43969903 on Build GRCh38
rs757814133 Chr.1: 43970221-43970221 on Build GRCh38
rs908460094 Chr.1: 43970019-43970019 on Build GRCh38
rs982546363 Chr.1: 43970248-43970248 on Build GRCh38
rs952581528 Chr.1: 43970245-43970245 on Build GRCh38
rs761585568 Chr.1: 43970277-43970277 on Build GRCh38
rs11539397 Chr.1: 43970118-43970118 on Build GRCh38
rs770453378 Chr.1: 43970130-43970130 on Build GRCh38
rs567976339 Chr.1: 43969967-43969967 on Build GRCh38
rs72890645 Chr.1: 43970148-43970148 on Build GRCh38
rs771207387 Chr.1: 43970150-43970150 on Build GRCh38
rs999138871 Chr.1: 43969882-43969882 on Build GRCh38
rs775594787 Chr.1: 43970266-43970266 on Build GRCh38
rs751135996 Chr.1: 43970228-43970228 on Build GRCh38
rs754598526 Chr.1: 43970313-43970313 on Build GRCh38
rs747105105 Chr.1: 43970267-43970267 on Build GRCh38
rs760569805 Chr.1: 43970181-43970181 on Build GRCh38
rs566618785 Chr.1: 43970033-43970033 on Build GRCh38
rs1031912642 Chr.1: 43969886-43969886 on Build GRCh38
rs1013416553 Chr.1: 43969918-43969918 on Build GRCh38
rs904844091 Chr.1: 43970200-43970200 on Build GRCh38
rs904995616 Chr.1: 43970300-43970300 on Build GRCh38
rs530720882 Chr.1: 43969983-43969983 on Build GRCh38
rs749663452 Chr.1: 43970230-43970230 on Build GRCh38
rs1025031384 Chr.1: 43970192-43970192 on Build GRCh38
rs11539399 Chr.1: 43970249-43970249 on Build GRCh38
rs765961038 Chr.1: 43970307-43970307 on Build GRCh38
rs775262625 Chr.1: 43970179-43970179 on Build GRCh38
rs1026662731 Chr.1: 43969973-43969973 on Build GRCh38
rs373058733 Chr.1: 43970284-43970284 on Build GRCh38
rs756626745 Chr.1: 43970229-43970229 on Build GRCh38
rs1057249658 Chr.1: 43970232-43970232 on Build GRCh38
rs548023940 Chr.1: 43969965-43969965 on Build GRCh38
rs975551270 Chr.1: 43970074-43970074 on Build GRCh38
rs995386908 Chr.1: 43969942-43969942 on Build GRCh38
rs72890643 Chr.1: 43970065-43970065 on Build GRCh38
rs751043631 Chr.1: 43970308-43970308 on Build GRCh38
rs747159514 Chr.1: 43970178-43970178 on Build GRCh38
rs760578415 Chr.1: 43970309-43970309 on Build GRCh38
rs570368154 Chr.1: 43970016-43970016 on Build GRCh38
rs892167766 Chr.1: 43969899-43969899 on Build GRCh38
rs979395874 Chr.1: 43970215-43970215 on Build GRCh38
rs41270389 Chr.1: 43970227-43970227 on Build GRCh38
rs534816407 Chr.1: 43970293-43970293 on Build GRCh38
rs976683409 Chr.1: 43970031-43970031 on Build GRCh38
rs746144496 Chr.1: 43970242-43970242 on Build GRCh38
rs148793639 Chr.1: 43970233-43970233 on Build GRCh38
rs760370833 Chr.1: 43970198-43970211 on Build GRCh38
rs41270387 Chr.1: 43969914-43969914 on Build GRCh38
rs761602213 Chr.1: 43970208-43970208 on Build GRCh38
rs768515197 Chr.1: 43970270-43970270 on Build GRCh38
rs905111812 Chr.1: 43969941-43969941 on Build GRCh38
rs759518745 Chr.1: 43970177-43970177 on Build GRCh38
rs753572731 Chr.1: 43970198-43970198 on Build GRCh38
rs774698916 Chr.1: 43970167-43970167 on Build GRCh38
rs1027262256 Chr.1: 43970002-43970002 on Build GRCh38
rs373268510 Chr.1: 43970311-43970311 on Build GRCh38

View Full Product Details