Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43970511-43971018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH2
Entrez Gene ID
1802
Gene Name
DPH2 homolog
Gene Aliases
DPH2L2
UniGene
Hs.632398
Gene Chromosome Location
Chr.1: 43969979-43973371 on Build GRCh38
Transcripts
NM_001039589.1 NM_001319165.1 NM_001319166.1 NM_001319167.1 NM_001319168.1 NM_001319169.1 NM_001319170.1 NM_001319171.1 NM_001384.4 XM_017000502.1 XM_017000503.1 XM_017000504.1 XM_017000505.1 XM_017000506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4008317 c.163C>T p.P55S Chr.1: 43970611-43970611 on Build GRCh38
464668 c.257G>A p.G86D Chr.1: 43970705-43970705 on Build GRCh38
1204482 c.310A>G p.I104V Chr.1: 43971015-43971015 on Build GRCh38
3490047 c.232T>C p.F78L Chr.1: 43970680-43970680 on Build GRCh38
5977988 c.289C>A p.Q97K Chr.1: 43970994-43970994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756206247 Chr.1: 43970771-43970771 on Build GRCh38
rs377624927 Chr.1: 43970623-43970623 on Build GRCh38
rs548765835 Chr.1: 43970776-43970776 on Build GRCh38
rs775084102 Chr.1: 43970744-43970744 on Build GRCh38
rs776517490 Chr.1: 43970547-43970547 on Build GRCh38
rs1004094009 Chr.1: 43970606-43970606 on Build GRCh38
rs760222935 Chr.1: 43970818-43970818 on Build GRCh38
rs988880274 Chr.1: 43970663-43970663 on Build GRCh38
rs745672737 Chr.1: 43970804-43970804 on Build GRCh38
rs751940280 Chr.1: 43970758-43970758 on Build GRCh38
rs993064138 Chr.1: 43970727-43970727 on Build GRCh38
rs572975806 Chr.1: 43970886-43970886 on Build GRCh38
rs775952481 Chr.1: 43970971-43970971 on Build GRCh38
rs759870886 Chr.1: 43970946-43970946 on Build GRCh38
rs980260780 Chr.1: 43970945-43970945 on Build GRCh38
rs781198882 Chr.1: 43970685-43970685 on Build GRCh38
rs757204487 Chr.1: 43970869-43970869 on Build GRCh38
rs370141264 Chr.1: 43970620-43970620 on Build GRCh38
rs1024328667 Chr.1: 43970733-43970733 on Build GRCh38
rs780243702 Chr.1: 43970654-43970654 on Build GRCh38
rs767907879 Chr.1: 43970966-43970966 on Build GRCh38
rs950242509 Chr.1: 43970687-43970687 on Build GRCh38
rs199939707 Chr.1: 43970569-43970569 on Build GRCh38
rs765679623 Chr.1: 43970637-43970637 on Build GRCh38
rs781393933 Chr.1: 43970764-43970764 on Build GRCh38
rs375949212 Chr.1: 43970740-43970740 on Build GRCh38
rs374261150 Chr.1: 43970931-43970931 on Build GRCh38
rs931204152 Chr.1: 43970802-43970802 on Build GRCh38
rs775942877 Chr.1: 43970921-43970921 on Build GRCh38
rs750733521 Chr.1: 43970644-43970644 on Build GRCh38
rs746683174 Chr.1: 43970893-43970893 on Build GRCh38
rs747657600 Chr.1: 43970925-43970925 on Build GRCh38
rs766046002 Chr.1: 43970577-43970577 on Build GRCh38
rs767178285 Chr.1: 43970598-43970598 on Build GRCh38
rs956078693 Chr.1: 43970640-43970640 on Build GRCh38
rs765438963 Chr.1: 43970704-43970704 on Build GRCh38
rs766687160 Chr.1: 43970756-43970756 on Build GRCh38
rs41270391 Chr.1: 43970877-43970877 on Build GRCh38
rs371553445 Chr.1: 43970748-43970748 on Build GRCh38
rs533914131 Chr.1: 43970551-43970577 on Build GRCh38
rs774113170 Chr.1: 43970723-43970723 on Build GRCh38
rs770679075 Chr.1: 43970710-43970710 on Build GRCh38
rs180764410 Chr.1: 43970795-43970795 on Build GRCh38
rs768103521 Chr.1: 43970826-43970826 on Build GRCh38
rs753670741 Chr.1: 43970610-43970610 on Build GRCh38
rs751851993 Chr.1: 43970838-43970838 on Build GRCh38
rs759091784 Chr.1: 43970587-43970587 on Build GRCh38
rs1028015745 Chr.1: 43970651-43970651 on Build GRCh38
rs771025509 Chr.1: 43970792-43970792 on Build GRCh38
rs577504701 Chr.1: 43970856-43970856 on Build GRCh38
rs1041116607 Chr.1: 43970629-43970629 on Build GRCh38
rs931128576 Chr.1: 43970575-43970575 on Build GRCh38
rs775075788 Chr.1: 43970812-43970812 on Build GRCh38
rs757479686 Chr.1: 43970982-43970982 on Build GRCh38
rs1000624774 Chr.1: 43970836-43970836 on Build GRCh38
rs41307832 Chr.1: 43970837-43970837 on Build GRCh38
rs971975088 Chr.1: 43970936-43970936 on Build GRCh38
rs539411300 Chr.1: 43970862-43970862 on Build GRCh38
rs763357329 Chr.1: 43970755-43970755 on Build GRCh38
rs1028240471 Chr.1: 43970561-43970561 on Build GRCh38
rs142635132 Chr.1: 43970747-43970747 on Build GRCh38
rs926468165 Chr.1: 43970574-43970574 on Build GRCh38
rs183925193 Chr.1: 43970700-43970700 on Build GRCh38
rs1000769331 Chr.1: 43970671-43970671 on Build GRCh38
rs765255953 Chr.1: 43970983-43970983 on Build GRCh38
rs778999585 Chr.1: 43970870-43970870 on Build GRCh38
rs771783956 Chr.1: 43970810-43970810 on Build GRCh38
rs996572108 Chr.1: 43970643-43970643 on Build GRCh38
rs377715065 Chr.1: 43970978-43970978 on Build GRCh38
rs550586020 Chr.1: 43970670-43970670 on Build GRCh38
rs372746396 Chr.1: 43970559-43970559 on Build GRCh38
rs759956616 Chr.1: 43970839-43970839 on Build GRCh38
rs759225604 Chr.1: 43970737-43970737 on Build GRCh38
rs761763039 Chr.1: 43970642-43970643 on Build GRCh38
rs371361825 Chr.1: 43970979-43970979 on Build GRCh38
rs745654289 Chr.1: 43970875-43970875 on Build GRCh38
rs758756539 Chr.1: 43970646-43970646 on Build GRCh38
rs752817178 Chr.1: 43970767-43970767 on Build GRCh38
rs199710836 Chr.1: 43970642-43970642 on Build GRCh38
rs773740510 Chr.1: 43970830-43970830 on Build GRCh38
rs372033617 Chr.1: 43970829-43970829 on Build GRCh38
rs374257990 Chr.1: 43970759-43970759 on Build GRCh38
rs917219457 Chr.1: 43970535-43970535 on Build GRCh38
rs749094938 Chr.1: 43970707-43970707 on Build GRCh38
rs756185728 Chr.1: 43970859-43970859 on Build GRCh38
rs750438307 Chr.1: 43970989-43970989 on Build GRCh38
rs917336368 Chr.1: 43970894-43970894 on Build GRCh38
rs768389156 Chr.1: 43970920-43970920 on Build GRCh38
rs750330294 Chr.1: 43970746-43970746 on Build GRCh38
rs28656782 Chr.1: 43970628-43970628 on Build GRCh38
rs774896681 Chr.1: 43970940-43970940 on Build GRCh38
rs564434138 Chr.1: 43970683-43970683 on Build GRCh38
rs555689023 Chr.1: 43970740-43970740 on Build GRCh38
rs749221708 Chr.1: 43970790-43970790 on Build GRCh38
rs763317049 Chr.1: 43970833-43970833 on Build GRCh38
rs1028669285 Chr.1: 43970825-43970825 on Build GRCh38
rs932310991 Chr.1: 43970958-43970958 on Build GRCh38
rs773001958 Chr.1: 43970570-43970570 on Build GRCh38
rs370695865 Chr.1: 43970572-43970572 on Build GRCh38
rs147087261 Chr.1: 43970640-43970641 on Build GRCh38
rs760762295 Chr.1: 43970973-43970973 on Build GRCh38
rs779737706 Chr.1: 43970880-43970880 on Build GRCh38
rs186465478 Chr.1: 43970844-43970844 on Build GRCh38
rs757737025 Chr.1: 43970632-43970632 on Build GRCh38
rs151064317 Chr.1: 43970703-43970703 on Build GRCh38
rs774201032 Chr.1: 43970580-43970580 on Build GRCh38
rs939947757 Chr.1: 43970858-43970858 on Build GRCh38

View Full Product Details