Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44674635-44675143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf228
Entrez Gene ID
339541
Gene Name
chromosome 1 open reading frame 228
Gene Aliases
NCRNA00082, p40
UniGene
Hs.173679
Gene Chromosome Location
Chr.1: 44674696-44725591 on Build GRCh38
Transcripts
NM_001145636.1 XM_006710603.2 XM_006710604.2 XM_011541340.1 XM_011541341.1 XM_011541343.1 XM_011541345.1 XM_011541349.1 XM_011541354.1 XM_011541355.1 XM_017001140.1 XM_017001141.1 XM_017001142.1 XM_017001143.1 XM_017001144.1 XM_017001145.1 XM_017001146.1 XM_017001147.1 XM_017001148.1

SNP Detail

SNP ID Chromosome Location
rs115426107 Chr.1: 44675004-44675004 on Build GRCh38
rs1028038646 Chr.1: 44674860-44674860 on Build GRCh38
rs891906162 Chr.1: 44674903-44674903 on Build GRCh38
rs533683415 Chr.1: 44674951-44674952 on Build GRCh38
rs1016689610 Chr.1: 44675033-44675033 on Build GRCh38
rs905236665 Chr.1: 44674745-44674745 on Build GRCh38
rs775879069 Chr.1: 44674664-44674664 on Build GRCh38
rs992793036 Chr.1: 44675000-44675000 on Build GRCh38
rs1022657248 Chr.1: 44674848-44674848 on Build GRCh38
rs1045212060 Chr.1: 44674721-44674721 on Build GRCh38
rs1048524329 Chr.1: 44674762-44674762 on Build GRCh38
rs571428195 Chr.1: 44674740-44674740 on Build GRCh38
rs564354279 Chr.1: 44675047-44675047 on Build GRCh38
rs934572094 Chr.1: 44674679-44674679 on Build GRCh38
rs1031527435 Chr.1: 44674934-44674934 on Build GRCh38
rs576397010 Chr.1: 44674979-44674979 on Build GRCh38
rs530618584 Chr.1: 44674975-44674978 on Build GRCh38
rs554010999 Chr.1: 44674799-44674799 on Build GRCh38
rs983841332 Chr.1: 44674696-44674696 on Build GRCh38
rs917952882 Chr.1: 44674705-44674705 on Build GRCh38
rs41307886 Chr.1: 44674693-44674693 on Build GRCh38
rs145423590 Chr.1: 44674948-44674951 on Build GRCh38
rs780240689 Chr.1: 44674964-44674965 on Build GRCh38
rs996430159 Chr.1: 44674855-44674855 on Build GRCh38
rs35280501 Chr.1: 44674895-44674895 on Build GRCh38
rs1054457413 Chr.1: 44674886-44674886 on Build GRCh38
rs1020279372 Chr.1: 44674839-44674839 on Build GRCh38
rs573587959 Chr.1: 44674854-44674854 on Build GRCh38
rs972483837 Chr.1: 44675117-44675117 on Build GRCh38
rs1005244699 Chr.1: 44675031-44675031 on Build GRCh38
rs35799772 Chr.1: 44674987-44674988 on Build GRCh38
rs113637216 Chr.1: 44674947-44674948 on Build GRCh38
rs540600102 Chr.1: 44675115-44675115 on Build GRCh38
rs968410743 Chr.1: 44674896-44674896 on Build GRCh38
rs577790483 Chr.1: 44675070-44675070 on Build GRCh38
rs906376736 Chr.1: 44674930-44674930 on Build GRCh38
rs1042237221 Chr.1: 44674927-44674927 on Build GRCh38
rs1033611939 Chr.1: 44674976-44674976 on Build GRCh38
rs372056203 Chr.1: 44674841-44674841 on Build GRCh38
rs369363774 Chr.1: 44674935-44674935 on Build GRCh38
rs949356217 Chr.1: 44674715-44674715 on Build GRCh38
rs1033731784 Chr.1: 44674667-44674667 on Build GRCh38
rs536234792 Chr.1: 44674858-44674858 on Build GRCh38
rs961149375 Chr.1: 44675054-44675054 on Build GRCh38
rs972934444 Chr.1: 44674807-44674807 on Build GRCh38
rs774821413 Chr.1: 44674797-44674797 on Build GRCh38
rs934203030 Chr.1: 44674834-44674834 on Build GRCh38
rs139934200 Chr.1: 44674904-44674904 on Build GRCh38
rs971093693 Chr.1: 44674660-44674660 on Build GRCh38
rs202025534 Chr.1: 44674960-44674961 on Build GRCh38
rs1003832933 Chr.1: 44674976-44674979 on Build GRCh38
rs769071628 Chr.1: 44674776-44674776 on Build GRCh38
rs533859754 Chr.1: 44674759-44674759 on Build GRCh38
rs1012655632 Chr.1: 44674828-44674828 on Build GRCh38

View Full Product Details