Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44689786-44690295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf228
Entrez Gene ID
339541
Gene Name
chromosome 1 open reading frame 228
Gene Aliases
NCRNA00082, p40
UniGene
Hs.173679
Gene Chromosome Location
Chr.1: 44674696-44725591 on Build GRCh38
Transcripts
NM_001145636.1 XM_006710603.2 XM_006710604.2 XM_011541340.1 XM_011541341.1 XM_011541343.1 XM_011541345.1 XM_011541349.1 XM_011541354.1 XM_011541355.1 XM_017001140.1 XM_017001141.1 XM_017001142.1 XM_017001143.1 XM_017001144.1 XM_017001145.1 XM_017001146.1 XM_017001147.1 XM_017001148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381090 c.155C>T p.S52F Chr.1: 44689852-44689852 on Build GRCh38
4597223 c.186G>C p.L62F Chr.1: 44689883-44689883 on Build GRCh38
4754177 c.181C>T p.R61C Chr.1: 44689878-44689878 on Build GRCh38
6443166 c.113C>T p.T38I Chr.1: 44689810-44689810 on Build GRCh38
4639388 c.180A>T p.V60V Chr.1: 44689877-44689877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914568974 Chr.1: 44690185-44690185 on Build GRCh38
rs758152008 Chr.1: 44689922-44689922 on Build GRCh38
rs949953682 Chr.1: 44689958-44689958 on Build GRCh38
rs187961882 Chr.1: 44689948-44689948 on Build GRCh38
rs981284994 Chr.1: 44690267-44690267 on Build GRCh38
rs753854228 Chr.1: 44689903-44689903 on Build GRCh38
rs942952749 Chr.1: 44689919-44689919 on Build GRCh38
rs962384506 Chr.1: 44690040-44690040 on Build GRCh38
rs544882031 Chr.1: 44690084-44690084 on Build GRCh38
rs562834827 Chr.1: 44690043-44690043 on Build GRCh38
rs1023241170 Chr.1: 44690029-44690029 on Build GRCh38
rs764074201 Chr.1: 44689891-44689891 on Build GRCh38
rs1029371567 Chr.1: 44690143-44690143 on Build GRCh38
rs1055098979 Chr.1: 44689989-44689989 on Build GRCh38
rs572770304 Chr.1: 44689947-44689947 on Build GRCh38
rs971321650 Chr.1: 44690219-44690219 on Build GRCh38
rs866711030 Chr.1: 44689852-44689852 on Build GRCh38
rs935800651 Chr.1: 44689945-44689945 on Build GRCh38
rs1025384455 Chr.1: 44690110-44690110 on Build GRCh38
rs947434918 Chr.1: 44689937-44689937 on Build GRCh38
rs766802737 Chr.1: 44689837-44689837 on Build GRCh38
rs1041743939 Chr.1: 44690256-44690256 on Build GRCh38
rs964303174 Chr.1: 44690174-44690174 on Build GRCh38
rs761644249 Chr.1: 44690218-44690219 on Build GRCh38
rs565054488 Chr.1: 44690140-44690140 on Build GRCh38
rs574062940 Chr.1: 44689987-44689987 on Build GRCh38
rs745504241 Chr.1: 44689907-44689907 on Build GRCh38
rs986951498 Chr.1: 44690159-44690159 on Build GRCh38
rs558626016 Chr.1: 44690166-44690166 on Build GRCh38
rs142681287 Chr.1: 44689879-44689879 on Build GRCh38
rs746476249 Chr.1: 44689946-44689946 on Build GRCh38
rs779697673 Chr.1: 44689942-44689942 on Build GRCh38
rs995206416 Chr.1: 44690045-44690045 on Build GRCh38
rs913051389 Chr.1: 44690209-44690209 on Build GRCh38
rs951610546 Chr.1: 44689877-44689877 on Build GRCh38
rs896376352 Chr.1: 44690010-44690010 on Build GRCh38
rs998428723 Chr.1: 44690142-44690142 on Build GRCh38
rs576236740 Chr.1: 44690079-44690079 on Build GRCh38
rs1056710742 Chr.1: 44689999-44689999 on Build GRCh38
rs547265585 Chr.1: 44690207-44690207 on Build GRCh38
rs1046410660 Chr.1: 44689962-44689962 on Build GRCh38
rs767357067 Chr.1: 44690017-44690017 on Build GRCh38
rs774412145 Chr.1: 44689864-44689864 on Build GRCh38
rs527600819 Chr.1: 44690148-44690148 on Build GRCh38
rs543179819 Chr.1: 44690027-44690027 on Build GRCh38
rs544999083 Chr.1: 44690067-44690067 on Build GRCh38
rs951205754 Chr.1: 44690133-44690133 on Build GRCh38
rs957246719 Chr.1: 44690181-44690181 on Build GRCh38
rs906536559 Chr.1: 44689973-44689973 on Build GRCh38
rs373236704 Chr.1: 44689921-44689922 on Build GRCh38
rs756274979 Chr.1: 44689883-44689883 on Build GRCh38
rs1006109030 Chr.1: 44690023-44690023 on Build GRCh38
rs912734973 Chr.1: 44690173-44690173 on Build GRCh38
rs757980900 Chr.1: 44689940-44689940 on Build GRCh38
rs547273108 Chr.1: 44690016-44690016 on Build GRCh38
rs571886999 Chr.1: 44690060-44690060 on Build GRCh38
rs1033148420 Chr.1: 44690167-44690167 on Build GRCh38
rs760169265 Chr.1: 44690182-44690182 on Build GRCh38
rs375684864 Chr.1: 44690028-44690028 on Build GRCh38
rs534461813 Chr.1: 44689877-44689877 on Build GRCh38
rs377494103 Chr.1: 44689819-44689819 on Build GRCh38
rs953657169 Chr.1: 44690146-44690146 on Build GRCh38
rs761747899 Chr.1: 44689904-44689904 on Build GRCh38
rs913118357 Chr.1: 44689955-44689955 on Build GRCh38

View Full Product Details