Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44850415-44850921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B3
Entrez Gene ID
8891
Gene Name
eukaryotic translation initiation factor 2B subunit gamma
Gene Aliases
EIF-2B, EIF2Bgamma
UniGene
Hs.533549
Gene Chromosome Location
Chr.1: 44850522-44986722 on Build GRCh38
Transcripts
NM_001261418.1 NM_020365.4 XM_017002747.1

SNP Detail

SNP ID Chromosome Location
rs776955130 Chr.1: 44850728-44850728 on Build GRCh38
rs201582864 Chr.1: 44850656-44850656 on Build GRCh38
rs141384432 Chr.1: 44850520-44850520 on Build GRCh38
rs966968205 Chr.1: 44850540-44850540 on Build GRCh38
rs893190309 Chr.1: 44850898-44850898 on Build GRCh38
rs942276440 Chr.1: 44850685-44850685 on Build GRCh38
rs930207623 Chr.1: 44850784-44850784 on Build GRCh38
rs559357410 Chr.1: 44850698-44850698 on Build GRCh38
rs973652153 Chr.1: 44850688-44850688 on Build GRCh38
rs916312002 Chr.1: 44850716-44850717 on Build GRCh38
rs779387196 Chr.1: 44850505-44850505 on Build GRCh38
rs1011183927 Chr.1: 44850899-44850899 on Build GRCh38
rs773280710 Chr.1: 44850537-44850537 on Build GRCh38
rs1051490898 Chr.1: 44850754-44850754 on Build GRCh38
rs61325585 Chr.1: 44850876-44850876 on Build GRCh38
rs1038732567 Chr.1: 44850789-44850789 on Build GRCh38
rs1017494635 Chr.1: 44850450-44850450 on Build GRCh38
rs963218115 Chr.1: 44850720-44850720 on Build GRCh38
rs890205369 Chr.1: 44850779-44850779 on Build GRCh38
rs545680194 Chr.1: 44850690-44850690 on Build GRCh38
rs1010809766 Chr.1: 44850896-44850896 on Build GRCh38
rs991449922 Chr.1: 44850669-44850669 on Build GRCh38
rs555023984 Chr.1: 44850519-44850519 on Build GRCh38
rs771038751 Chr.1: 44850684-44850684 on Build GRCh38
rs1051127603 Chr.1: 44850787-44850787 on Build GRCh38
rs760908698 Chr.1: 44850678-44850678 on Build GRCh38
rs543724503 Chr.1: 44850549-44850549 on Build GRCh38
rs1022646336 Chr.1: 44850555-44850555 on Build GRCh38
rs974256615 Chr.1: 44850736-44850736 on Build GRCh38
rs541401892 Chr.1: 44850516-44850516 on Build GRCh38
rs532328702 Chr.1: 44850631-44850631 on Build GRCh38
rs116672991 Chr.1: 44850550-44850550 on Build GRCh38
rs1034641091 Chr.1: 44850662-44850662 on Build GRCh38
rs963128600 Chr.1: 44850570-44850570 on Build GRCh38
rs893504321 Chr.1: 44850838-44850838 on Build GRCh38
rs999187954 Chr.1: 44850617-44850617 on Build GRCh38
rs942677796 Chr.1: 44850878-44850878 on Build GRCh38
rs966581314 Chr.1: 44850585-44850585 on Build GRCh38
rs528195798 Chr.1: 44850750-44850750 on Build GRCh38
rs942993930 Chr.1: 44850781-44850781 on Build GRCh38

View Full Product Details