Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44941282-44941782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B3
Entrez Gene ID
8891
Gene Name
eukaryotic translation initiation factor 2B subunit gamma
Gene Aliases
EIF-2B, EIF2Bgamma
UniGene
Hs.533549
Gene Chromosome Location
Chr.1: 44850522-44986722 on Build GRCh38
Transcripts
NM_001166588.2 NM_001261418.1 NM_020365.4 XM_017002745.1 XM_017002746.1 XM_017002747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582927 c.353T>C p.V118A Chr.1: 44941607-44941607 on Build GRCh38
4008458 c.347A>C p.E116A Chr.1: 44941613-44941613 on Build GRCh38
1627089 c.383T>G p.L128R Chr.1: 44941577-44941577 on Build GRCh38
1343023 c.437A>C p.K146T Chr.1: 44941523-44941523 on Build GRCh38
1492077 c.441G>A p.G147G Chr.1: 44941519-44941519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760489343 Chr.1: 44941566-44941566 on Build GRCh38
rs373262348 Chr.1: 44941622-44941622 on Build GRCh38
rs371230685 Chr.1: 44941600-44941600 on Build GRCh38
rs759538945 Chr.1: 44941554-44941554 on Build GRCh38
rs764856042 Chr.1: 44941585-44941585 on Build GRCh38
rs905204066 Chr.1: 44941512-44941512 on Build GRCh38
rs529461543 Chr.1: 44941528-44941528 on Build GRCh38
rs965074205 Chr.1: 44941322-44941322 on Build GRCh38
rs1004425628 Chr.1: 44941541-44941541 on Build GRCh38
rs966135471 Chr.1: 44941606-44941606 on Build GRCh38
rs771864524 Chr.1: 44941556-44941556 on Build GRCh38
rs761553484 Chr.1: 44941666-44941666 on Build GRCh38
rs114327055 Chr.1: 44941751-44941751 on Build GRCh38
rs576192741 Chr.1: 44941674-44941674 on Build GRCh38
rs564925413 Chr.1: 44941387-44941387 on Build GRCh38
rs755510412 Chr.1: 44941593-44941593 on Build GRCh38
rs758266461 Chr.1: 44941731-44941731 on Build GRCh38
rs778078435 Chr.1: 44941602-44941602 on Build GRCh38
rs771010080 Chr.1: 44941618-44941618 on Build GRCh38
rs776548044 Chr.1: 44941663-44941663 on Build GRCh38
rs987821163 Chr.1: 44941651-44941651 on Build GRCh38
rs765905949 Chr.1: 44941705-44941705 on Build GRCh38
rs886046361 Chr.1: 44941572-44941572 on Build GRCh38
rs749505713 Chr.1: 44941552-44941552 on Build GRCh38
rs544660255 Chr.1: 44941386-44941386 on Build GRCh38
rs978668650 Chr.1: 44941406-44941406 on Build GRCh38
rs113994023 Chr.1: 44941553-44941553 on Build GRCh38
rs925753741 Chr.1: 44941742-44941742 on Build GRCh38
rs978984968 Chr.1: 44941321-44941321 on Build GRCh38
rs934377820 Chr.1: 44941403-44941403 on Build GRCh38
rs752385453 Chr.1: 44941478-44941478 on Build GRCh38
rs373870315 Chr.1: 44941545-44941545 on Build GRCh38
rs144947838 Chr.1: 44941583-44941583 on Build GRCh38
rs201305666 Chr.1: 44941493-44941493 on Build GRCh38
rs148977100 Chr.1: 44941509-44941509 on Build GRCh38
rs756888116 Chr.1: 44941495-44941495 on Build GRCh38
rs753414286 Chr.1: 44941494-44941494 on Build GRCh38
rs762424980 Chr.1: 44941692-44941692 on Build GRCh38
rs760687647 Chr.1: 44941638-44941638 on Build GRCh38
rs768615168 Chr.1: 44941643-44941643 on Build GRCh38
rs764724963 Chr.1: 44941668-44941668 on Build GRCh38
rs745499706 Chr.1: 44941627-44941627 on Build GRCh38
rs759359515 Chr.1: 44941466-44941466 on Build GRCh38
rs763969996 Chr.1: 44941570-44941570 on Build GRCh38
rs377508889 Chr.1: 44941717-44941717 on Build GRCh38
rs781640168 Chr.1: 44941530-44941530 on Build GRCh38
rs749857178 Chr.1: 44941592-44941592 on Build GRCh38
rs749560824 Chr.1: 44941612-44941612 on Build GRCh38
rs529374377 Chr.1: 44941521-44941521 on Build GRCh38
rs576125590 Chr.1: 44941311-44941311 on Build GRCh38
rs373316628 Chr.1: 44941679-44941679 on Build GRCh38
rs77957684 Chr.1: 44941429-44941429 on Build GRCh38
rs779081102 Chr.1: 44941620-44941620 on Build GRCh38
rs770013712 Chr.1: 44941532-44941532 on Build GRCh38
rs968578263 Chr.1: 44941404-44941404 on Build GRCh38
rs755863082 Chr.1: 44941484-44941484 on Build GRCh38
rs917995621 Chr.1: 44941727-44941727 on Build GRCh38
rs368710944 Chr.1: 44941467-44941467 on Build GRCh38
rs775628640 Chr.1: 44941635-44941635 on Build GRCh38
rs925838927 Chr.1: 44941374-44941374 on Build GRCh38
rs753240802 Chr.1: 44941708-44941708 on Build GRCh38
rs934502303 Chr.1: 44941436-44941436 on Build GRCh38
rs748442078 Chr.1: 44941531-44941531 on Build GRCh38
rs753614565 Chr.1: 44941571-44941571 on Build GRCh38
rs748937918 Chr.1: 44941510-44941510 on Build GRCh38
rs993091279 Chr.1: 44941722-44941722 on Build GRCh38
rs770776173 Chr.1: 44941509-44941510 on Build GRCh38
rs975508648 Chr.1: 44941364-44941364 on Build GRCh38
rs924563383 Chr.1: 44941683-44941683 on Build GRCh38
rs773479090 Chr.1: 44941548-44941548 on Build GRCh38
rs775430377 Chr.1: 44941557-44941557 on Build GRCh38
rs921178263 Chr.1: 44941381-44941381 on Build GRCh38
rs965531974 Chr.1: 44941578-44941578 on Build GRCh38
rs151056457 Chr.1: 44941518-44941518 on Build GRCh38
rs745869984 Chr.1: 44941626-44941626 on Build GRCh38
rs756589415 Chr.1: 44941716-44941716 on Build GRCh38

View Full Product Details