Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14529456-14529954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB1
Entrez Gene ID
3337
Gene Name
DnaJ heat shock protein family (Hsp40) member B1
Gene Aliases
HSPF1, Hdj1, Hsp40, RSPH16B, Sis1
UniGene
Hs.515210
Gene Chromosome Location
Chr.19: 14514764-14529821 on Build GRCh38
Transcripts
XM_006722734.3 NR_030369.1 NM_001321170.1 NM_138501.5 NR_038103.1 NR_038104.1 XM_006722945.1 XM_011528442.2

Gene Symbol
MIR639
Entrez Gene ID
693224
Gene Name
microRNA 639
Gene Aliases
MIRN639, hsa-mir-639
UniGene
Gene Chromosome Location
Chr.19: 14529543-14529640 on Build GRCh38
Transcripts
XM_006722734.3 NR_030369.1 NM_001321170.1 NM_138501.5 NR_038103.1 NR_038104.1 XM_006722945.1 XM_011528442.2

Gene Symbol
TECR
Entrez Gene ID
9524
Gene Name
trans-2,3-enoyl-CoA reductase
Gene Aliases
GPSN2, MRT14, SC2, TER
UniGene
Hs.515642
Gene Chromosome Location
Chr.19: 14527760-14565980 on Build GRCh38
Transcripts
XM_006722734.3 NR_030369.1 NM_001321170.1 NM_138501.5 NR_038103.1 NR_038104.1 XM_006722945.1 XM_011528442.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7184154 c.13G>A p.E5K Chr.19: 14529709-14529709 on Build GRCh38
1304095 c.15G>A p.E5E Chr.19: 14529711-14529711 on Build GRCh38
6799244 c.15+2T>C p.? Chr.19: 14529713-14529713 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764238250 Chr.19: 14529663-14529663 on Build GRCh38
rs749206443 Chr.19: 14529564-14529564 on Build GRCh38
rs532362706 Chr.19: 14529795-14529795 on Build GRCh38
rs772597119 Chr.19: 14529644-14529644 on Build GRCh38
rs769798947 Chr.19: 14529856-14529856 on Build GRCh38
rs1011784923 Chr.19: 14529690-14529690 on Build GRCh38
rs762076874 Chr.19: 14529516-14529516 on Build GRCh38
rs768982369 Chr.19: 14529682-14529682 on Build GRCh38
rs746471139 Chr.19: 14529703-14529703 on Build GRCh38
rs375144461 Chr.19: 14529733-14529733 on Build GRCh38
rs769340365 Chr.19: 14529537-14529537 on Build GRCh38
rs901199855 Chr.19: 14529478-14529478 on Build GRCh38
rs1051026262 Chr.19: 14529641-14529641 on Build GRCh38
rs776469956 Chr.19: 14529721-14529721 on Build GRCh38
rs376552790 Chr.19: 14529522-14529522 on Build GRCh38
rs1022190739 Chr.19: 14529715-14529715 on Build GRCh38
rs751580195 Chr.19: 14529664-14529664 on Build GRCh38
rs1044431686 Chr.19: 14529665-14529665 on Build GRCh38
rs756014191 Chr.19: 14529599-14529599 on Build GRCh38
rs776105806 Chr.19: 14529635-14529635 on Build GRCh38
rs543638043 Chr.19: 14529556-14529556 on Build GRCh38
rs45556632 Chr.19: 14529591-14529591 on Build GRCh38
rs777167994 Chr.19: 14529582-14529582 on Build GRCh38
rs926172474 Chr.19: 14529864-14529864 on Build GRCh38
rs761583185 Chr.19: 14529709-14529709 on Build GRCh38
rs758263727 Chr.19: 14529660-14529660 on Build GRCh38
rs530079728 Chr.19: 14529601-14529601 on Build GRCh38
rs563599895 Chr.19: 14529617-14529617 on Build GRCh38
rs776974226 Chr.19: 14529616-14529616 on Build GRCh38
rs199703577 Chr.19: 14529621-14529621 on Build GRCh38
rs757369235 Chr.19: 14529754-14529754 on Build GRCh38
rs530850662 Chr.19: 14529820-14529820 on Build GRCh38
rs542276577 Chr.19: 14529815-14529815 on Build GRCh38
rs35149836 Chr.19: 14529627-14529627 on Build GRCh38
rs201564572 Chr.19: 14529620-14529620 on Build GRCh38
rs760411442 Chr.19: 14529646-14529646 on Build GRCh38
rs752733365 Chr.19: 14529658-14529658 on Build GRCh38
rs915028069 Chr.19: 14529625-14529625 on Build GRCh38
rs1021847353 Chr.19: 14529925-14529925 on Build GRCh38
rs996841773 Chr.19: 14529479-14529479 on Build GRCh38
rs757939788 Chr.19: 14529550-14529550 on Build GRCh38
rs201963947 Chr.19: 14529745-14529745 on Build GRCh38
rs140756444 Chr.19: 14529804-14529804 on Build GRCh38
rs559706248 Chr.19: 14529590-14529590 on Build GRCh38
rs545976635 Chr.19: 14529861-14529861 on Build GRCh38
rs773468256 Chr.19: 14529573-14529573 on Build GRCh38
rs956484051 Chr.19: 14529683-14529683 on Build GRCh38
rs374570462 Chr.19: 14529624-14529624 on Build GRCh38
rs775079725 Chr.19: 14529739-14529739 on Build GRCh38
rs781353464 Chr.19: 14529668-14529668 on Build GRCh38
rs767150176 Chr.19: 14529592-14529592 on Build GRCh38
rs939915013 Chr.19: 14529785-14529785 on Build GRCh38
rs148446064 Chr.19: 14529518-14529518 on Build GRCh38
rs756241661 Chr.19: 14529741-14529743 on Build GRCh38
rs369455862 Chr.19: 14529480-14529480 on Build GRCh38
rs1046194835 Chr.19: 14529801-14529801 on Build GRCh38
rs746060673 Chr.19: 14529671-14529671 on Build GRCh38
rs754981703 Chr.19: 14529629-14529629 on Build GRCh38
rs908495056 Chr.19: 14529781-14529781 on Build GRCh38
rs377724067 Chr.19: 14529631-14529631 on Build GRCh38
rs768392033 Chr.19: 14529622-14529622 on Build GRCh38
rs745590317 Chr.19: 14529509-14529509 on Build GRCh38
rs1039376395 Chr.19: 14529553-14529553 on Build GRCh38
rs114121047 Chr.19: 14529566-14529566 on Build GRCh38
rs766695196 Chr.19: 14529581-14529581 on Build GRCh38
rs142882191 Chr.19: 14529637-14529637 on Build GRCh38
rs888137067 Chr.19: 14529811-14529811 on Build GRCh38
rs113751040 Chr.19: 14529576-14529576 on Build GRCh38
rs565220991 Chr.19: 14529563-14529563 on Build GRCh38
rs765142855 Chr.19: 14529726-14529726 on Build GRCh38
rs554883325 Chr.19: 14529527-14529527 on Build GRCh38
rs970822597 Chr.19: 14529567-14529567 on Build GRCh38
rs200383949 Chr.19: 14529649-14529649 on Build GRCh38
rs1038610009 Chr.19: 14529653-14529653 on Build GRCh38
rs546240195 Chr.19: 14529632-14529632 on Build GRCh38
rs377544418 Chr.19: 14529606-14529606 on Build GRCh38
rs745620953 Chr.19: 14529560-14529560 on Build GRCh38
rs781549405 Chr.19: 14529633-14529633 on Build GRCh38
rs980459706 Chr.19: 14529570-14529574 on Build GRCh38
rs1015279318 Chr.19: 14529650-14529650 on Build GRCh38
rs780269482 Chr.19: 14529678-14529678 on Build GRCh38
rs935972370 Chr.19: 14529686-14529686 on Build GRCh38
rs1012227702 Chr.19: 14529901-14529901 on Build GRCh38
rs770226681 Chr.19: 14529619-14529619 on Build GRCh38
rs541505241 Chr.19: 14529803-14529803 on Build GRCh38
rs377238588 Chr.19: 14529648-14529648 on Build GRCh38
rs766073566 Chr.19: 14529647-14529647 on Build GRCh38
rs771674833 Chr.19: 14529638-14529638 on Build GRCh38
rs756999775 Chr.19: 14529512-14529512 on Build GRCh38
rs373360580 Chr.19: 14529536-14529536 on Build GRCh38
rs1033545317 Chr.19: 14529808-14529808 on Build GRCh38
rs376016848 Chr.19: 14529654-14529654 on Build GRCh38
rs369850035 Chr.19: 14529626-14529626 on Build GRCh38
rs762961634 Chr.19: 14529744-14529744 on Build GRCh38
rs956376335 Chr.19: 14529500-14529500 on Build GRCh38
rs772272016 Chr.19: 14529572-14529572 on Build GRCh38
rs898567454 Chr.19: 14529593-14529593 on Build GRCh38
rs1031933221 Chr.19: 14529677-14529677 on Build GRCh38
rs977329964 Chr.19: 14529822-14529822 on Build GRCh38
rs374421287 Chr.19: 14529667-14529667 on Build GRCh38
rs886959752 Chr.19: 14529584-14529584 on Build GRCh38
rs563538514 Chr.19: 14529816-14529816 on Build GRCh38
rs774759186 Chr.19: 14529510-14529510 on Build GRCh38
rs186954160 Chr.19: 14529812-14529812 on Build GRCh38
rs780192280 Chr.19: 14529764-14529766 on Build GRCh38
rs559207042 Chr.19: 14529914-14529914 on Build GRCh38
rs372602559 Chr.19: 14529587-14529587 on Build GRCh38
rs925913088 Chr.19: 14529684-14529684 on Build GRCh38
rs1029809048 Chr.19: 14529833-14529833 on Build GRCh38
rs988803198 Chr.19: 14529903-14529903 on Build GRCh38
rs576227374 Chr.19: 14529546-14529546 on Build GRCh38
rs754965095 Chr.19: 14529603-14529603 on Build GRCh38
rs900689268 Chr.19: 14529926-14529926 on Build GRCh38
rs973963567 Chr.19: 14529790-14529790 on Build GRCh38
rs780851912 Chr.19: 14529559-14529559 on Build GRCh38
rs748194576 Chr.19: 14529826-14529826 on Build GRCh38
rs767563738 Chr.19: 14529758-14529758 on Build GRCh38
rs747173264 Chr.19: 14529611-14529611 on Build GRCh38
rs182986875 Chr.19: 14529672-14529672 on Build GRCh38
rs771342700 Chr.19: 14529614-14529614 on Build GRCh38
rs778865767 Chr.19: 14529605-14529605 on Build GRCh38
rs147268675 Chr.19: 14529777-14529777 on Build GRCh38
rs772211952 Chr.19: 14529607-14529607 on Build GRCh38
rs748387906 Chr.19: 14529571-14529571 on Build GRCh38
rs1044463836 Chr.19: 14529805-14529805 on Build GRCh38
rs745798123 Chr.19: 14529595-14529595 on Build GRCh38
rs751227381 Chr.19: 14529558-14529558 on Build GRCh38
rs761057001 Chr.19: 14529577-14529577 on Build GRCh38
rs771584929 Chr.19: 14529797-14529797 on Build GRCh38
rs770235598 Chr.19: 14529634-14529634 on Build GRCh38
rs780843064 Chr.19: 14529489-14529489 on Build GRCh38
rs781159055 Chr.19: 14529512-14529513 on Build GRCh38
rs554821641 Chr.19: 14529695-14529695 on Build GRCh38
rs774563123 Chr.19: 14529569-14529569 on Build GRCh38
rs955026211 Chr.19: 14529763-14529763 on Build GRCh38
rs144615833 Chr.19: 14529876-14529876 on Build GRCh38
rs907232562 Chr.19: 14529666-14529666 on Build GRCh38
rs967817233 Chr.19: 14529720-14529720 on Build GRCh38
rs372535209 Chr.19: 14529681-14529681 on Build GRCh38
rs561305115 Chr.19: 14529623-14529623 on Build GRCh38
rs751668259 Chr.19: 14529753-14529753 on Build GRCh38
rs778093423 Chr.19: 14529610-14529610 on Build GRCh38

View Full Product Details