Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17300800-17301304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD8
Entrez Gene ID
79575
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
UniGene
Hs.515664
Gene Chromosome Location
Chr.19: 17292131-17303473 on Build GRCh38
Transcripts
NM_024527.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6590918 c.416G>A p.G139D Chr.19: 17301201-17301201 on Build GRCh38
6590920 c.442C>T p.R148W Chr.19: 17301175-17301175 on Build GRCh38
1680786 c.463A>G p.K155E Chr.19: 17301154-17301154 on Build GRCh38
2156757 c.598C>T p.R200C Chr.19: 17301019-17301019 on Build GRCh38
4443934 c.743T>C p.L248P Chr.19: 17300874-17300874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757916868 Chr.19: 17301166-17301166 on Build GRCh38
rs969538687 Chr.19: 17301065-17301065 on Build GRCh38
rs149325940 Chr.19: 17301275-17301275 on Build GRCh38
rs747389254 Chr.19: 17301182-17301183 on Build GRCh38
rs772271267 Chr.19: 17300840-17300840 on Build GRCh38
rs753121204 Chr.19: 17301130-17301131 on Build GRCh38
rs529850743 Chr.19: 17300853-17300853 on Build GRCh38
rs536054282 Chr.19: 17301092-17301092 on Build GRCh38
rs764146908 Chr.19: 17301129-17301129 on Build GRCh38
rs756925739 Chr.19: 17301174-17301174 on Build GRCh38
rs367899182 Chr.19: 17301059-17301060 on Build GRCh38
rs566396701 Chr.19: 17301061-17301061 on Build GRCh38
rs749306333 Chr.19: 17300959-17300959 on Build GRCh38
rs771791316 Chr.19: 17300984-17300984 on Build GRCh38
rs759388755 Chr.19: 17301218-17301218 on Build GRCh38
rs772396065 Chr.19: 17301214-17301214 on Build GRCh38
rs946527902 Chr.19: 17301202-17301202 on Build GRCh38
rs754863962 Chr.19: 17301159-17301159 on Build GRCh38
rs758982389 Chr.19: 17301156-17301159 on Build GRCh38
rs756405465 Chr.19: 17301263-17301263 on Build GRCh38
rs771338692 Chr.19: 17301022-17301022 on Build GRCh38
rs1014000322 Chr.19: 17301221-17301221 on Build GRCh38
rs202211622 Chr.19: 17300831-17300831 on Build GRCh38
rs759594879 Chr.19: 17300921-17300921 on Build GRCh38
rs750143429 Chr.19: 17301250-17301250 on Build GRCh38
rs373502327 Chr.19: 17301173-17301173 on Build GRCh38
rs755933630 Chr.19: 17301091-17301091 on Build GRCh38
rs768218913 Chr.19: 17300973-17300973 on Build GRCh38
rs748220540 Chr.19: 17301284-17301284 on Build GRCh38
rs749756854 Chr.19: 17301271-17301271 on Build GRCh38
rs183475016 Chr.19: 17301248-17301248 on Build GRCh38
rs765241379 Chr.19: 17301160-17301160 on Build GRCh38
rs765242155 Chr.19: 17301042-17301042 on Build GRCh38
rs776992540 Chr.19: 17301108-17301108 on Build GRCh38
rs776913911 Chr.19: 17300866-17300866 on Build GRCh38
rs1036904356 Chr.19: 17301247-17301247 on Build GRCh38
rs1001048620 Chr.19: 17301212-17301212 on Build GRCh38
rs776062813 Chr.19: 17301197-17301197 on Build GRCh38
rs530712660 Chr.19: 17300845-17300845 on Build GRCh38
rs762808953 Chr.19: 17301231-17301231 on Build GRCh38
rs370047329 Chr.19: 17301164-17301164 on Build GRCh38
rs777148469 Chr.19: 17301167-17301167 on Build GRCh38
rs780127653 Chr.19: 17300940-17300940 on Build GRCh38
rs781494346 Chr.19: 17301178-17301178 on Build GRCh38
rs768184815 Chr.19: 17301272-17301272 on Build GRCh38
rs747666959 Chr.19: 17301095-17301095 on Build GRCh38
rs750203612 Chr.19: 17300935-17300935 on Build GRCh38
rs760040334 Chr.19: 17301041-17301041 on Build GRCh38
rs1002898696 Chr.19: 17301276-17301276 on Build GRCh38
rs979897117 Chr.19: 17301071-17301071 on Build GRCh38
rs761958582 Chr.19: 17301143-17301143 on Build GRCh38
rs774566297 Chr.19: 17301131-17301131 on Build GRCh38
rs777957836 Chr.19: 17301162-17301163 on Build GRCh38
rs907807943 Chr.19: 17301124-17301124 on Build GRCh38
rs778529369 Chr.19: 17301094-17301094 on Build GRCh38
rs766875943 Chr.19: 17300872-17300872 on Build GRCh38
rs771215862 Chr.19: 17301104-17301104 on Build GRCh38
rs748998381 Chr.19: 17301192-17301197 on Build GRCh38
rs765364538 Chr.19: 17300930-17300930 on Build GRCh38
rs770314859 Chr.19: 17301116-17301116 on Build GRCh38
rs1050853182 Chr.19: 17300964-17300964 on Build GRCh38
rs144317638 Chr.19: 17300827-17300827 on Build GRCh38
rs957002325 Chr.19: 17301216-17301216 on Build GRCh38
rs775912056 Chr.19: 17301123-17301123 on Build GRCh38
rs369129606 Chr.19: 17301064-17301064 on Build GRCh38
rs913675091 Chr.19: 17300835-17300835 on Build GRCh38
rs150514651 Chr.19: 17300871-17300871 on Build GRCh38
rs775567753 Chr.19: 17301056-17301056 on Build GRCh38
rs762089945 Chr.19: 17301211-17301211 on Build GRCh38
rs757891883 Chr.19: 17301097-17301097 on Build GRCh38
rs748768481 Chr.19: 17300828-17300828 on Build GRCh38
rs570097692 Chr.19: 17300936-17300936 on Build GRCh38
rs779107395 Chr.19: 17300999-17301000 on Build GRCh38
rs765003550 Chr.19: 17301222-17301222 on Build GRCh38
rs780297888 Chr.19: 17301265-17301265 on Build GRCh38
rs376723965 Chr.19: 17300931-17300931 on Build GRCh38
rs377333754 Chr.19: 17301229-17301229 on Build GRCh38
rs539011418 Chr.19: 17300978-17300978 on Build GRCh38
rs989023450 Chr.19: 17301168-17301168 on Build GRCh38
rs555670452 Chr.19: 17301103-17301103 on Build GRCh38
rs774320476 Chr.19: 17301207-17301207 on Build GRCh38
rs761564968 Chr.19: 17300834-17300834 on Build GRCh38
rs371098180 Chr.19: 17301019-17301019 on Build GRCh38
rs756799431 Chr.19: 17301246-17301246 on Build GRCh38
rs1040080172 Chr.19: 17301167-17301168 on Build GRCh38
rs200255813 Chr.19: 17301201-17301201 on Build GRCh38
rs981703441 Chr.19: 17301156-17301156 on Build GRCh38
rs779677388 Chr.19: 17301191-17301192 on Build GRCh38
rs751059861 Chr.19: 17301243-17301243 on Build GRCh38
rs749764203 Chr.19: 17301200-17301200 on Build GRCh38
rs751789650 Chr.19: 17301086-17301086 on Build GRCh38
rs970263281 Chr.19: 17301128-17301128 on Build GRCh38
rs373958722 Chr.19: 17301240-17301240 on Build GRCh38
rs550188531 Chr.19: 17300929-17300929 on Build GRCh38
rs761049394 Chr.19: 17300858-17300858 on Build GRCh38
rs969801351 Chr.19: 17301227-17301227 on Build GRCh38
rs769371745 Chr.19: 17301183-17301188 on Build GRCh38
rs777114980 Chr.19: 17301038-17301038 on Build GRCh38
rs754390119 Chr.19: 17300911-17300911 on Build GRCh38
rs778584552 Chr.19: 17300975-17300975 on Build GRCh38
rs369279878 Chr.19: 17301147-17301147 on Build GRCh38
rs868204906 Chr.19: 17301052-17301052 on Build GRCh38
rs138354638 Chr.19: 17301062-17301062 on Build GRCh38
rs575924100 Chr.19: 17301113-17301113 on Build GRCh38
rs746198270 Chr.19: 17301196-17301196 on Build GRCh38
rs780799086 Chr.19: 17300934-17300934 on Build GRCh38
rs773620898 Chr.19: 17301217-17301217 on Build GRCh38

View Full Product Details