Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17301280-17301771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD8
Entrez Gene ID
79575
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
UniGene
Hs.515664
Gene Chromosome Location
Chr.19: 17292131-17303473 on Build GRCh38
Transcripts
NM_024527.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438897 c.208G>A p.G70R Chr.19: 17301409-17301409 on Build GRCh38
1391437 c.187G>A p.A63T Chr.19: 17301430-17301430 on Build GRCh38
4398129 c.317G>A p.G106E Chr.19: 17301300-17301300 on Build GRCh38
1579915 c.217T>A p.S73T Chr.19: 17301400-17301400 on Build GRCh38
4406590 c.14T>G p.V5G Chr.19: 17301603-17301603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912465414 Chr.19: 17301366-17301366 on Build GRCh38
rs372720665 Chr.19: 17301314-17301314 on Build GRCh38
rs762989608 Chr.19: 17301664-17301664 on Build GRCh38
rs779278939 Chr.19: 17301361-17301361 on Build GRCh38
rs759236130 Chr.19: 17301505-17301505 on Build GRCh38
rs1022195852 Chr.19: 17301507-17301507 on Build GRCh38
rs888456437 Chr.19: 17301676-17301676 on Build GRCh38
rs780297252 Chr.19: 17301567-17301567 on Build GRCh38
rs552233794 Chr.19: 17301595-17301595 on Build GRCh38
rs374747961 Chr.19: 17301644-17301644 on Build GRCh38
rs761619410 Chr.19: 17301562-17301563 on Build GRCh38
rs754123574 Chr.19: 17301351-17301351 on Build GRCh38
rs147808769 Chr.19: 17301556-17301556 on Build GRCh38
rs370070131 Chr.19: 17301423-17301423 on Build GRCh38
rs755289782 Chr.19: 17301422-17301422 on Build GRCh38
rs949602157 Chr.19: 17301594-17301594 on Build GRCh38
rs765984168 Chr.19: 17301496-17301496 on Build GRCh38
rs766966995 Chr.19: 17301406-17301406 on Build GRCh38
rs780429313 Chr.19: 17301342-17301342 on Build GRCh38
rs775329949 Chr.19: 17301608-17301608 on Build GRCh38
rs546146109 Chr.19: 17301560-17301560 on Build GRCh38
rs61735361 Chr.19: 17301425-17301425 on Build GRCh38
rs11086067 Chr.19: 17301590-17301590 on Build GRCh38
rs764675043 Chr.19: 17301540-17301540 on Build GRCh38
rs768033957 Chr.19: 17301429-17301452 on Build GRCh38
rs751547039 Chr.19: 17301319-17301319 on Build GRCh38
rs750486923 Chr.19: 17301652-17301652 on Build GRCh38
rs932304480 Chr.19: 17301357-17301357 on Build GRCh38
rs773955225 Chr.19: 17301452-17301453 on Build GRCh38
rs200372628 Chr.19: 17301302-17301302 on Build GRCh38
rs976352133 Chr.19: 17301733-17301733 on Build GRCh38
rs769538954 Chr.19: 17301606-17301606 on Build GRCh38
rs920908499 Chr.19: 17301335-17301335 on Build GRCh38
rs147441347 Chr.19: 17301398-17301398 on Build GRCh38
rs867777795 Chr.19: 17301485-17301485 on Build GRCh38
rs748001423 Chr.19: 17301344-17301344 on Build GRCh38
rs371431966 Chr.19: 17301642-17301642 on Build GRCh38
rs3745186 Chr.19: 17301409-17301409 on Build GRCh38
rs746966639 Chr.19: 17301585-17301585 on Build GRCh38
rs867132263 Chr.19: 17301313-17301313 on Build GRCh38
rs763907147 Chr.19: 17301318-17301318 on Build GRCh38
rs764619853 Chr.19: 17301640-17301640 on Build GRCh38
rs1010885468 Chr.19: 17301689-17301689 on Build GRCh38
rs755787357 Chr.19: 17301327-17301327 on Build GRCh38
rs747176177 Chr.19: 17301376-17301376 on Build GRCh38
rs757407162 Chr.19: 17301431-17301431 on Build GRCh38
rs11086066 Chr.19: 17301557-17301557 on Build GRCh38
rs968935995 Chr.19: 17301526-17301526 on Build GRCh38
rs766340046 Chr.19: 17301655-17301655 on Build GRCh38
rs776077620 Chr.19: 17301499-17301499 on Build GRCh38
rs919856528 Chr.19: 17301607-17301607 on Build GRCh38
rs767457257 Chr.19: 17301647-17301647 on Build GRCh38
rs777686823 Chr.19: 17301572-17301572 on Build GRCh38
rs746107446 Chr.19: 17301434-17301434 on Build GRCh38
rs774155542 Chr.19: 17301447-17301447 on Build GRCh38
rs367827324 Chr.19: 17301561-17301561 on Build GRCh38
rs151037617 Chr.19: 17301543-17301543 on Build GRCh38
rs968170107 Chr.19: 17301323-17301323 on Build GRCh38
rs748727592 Chr.19: 17301368-17301368 on Build GRCh38
rs767018432 Chr.19: 17301324-17301324 on Build GRCh38
rs755423619 Chr.19: 17301360-17301360 on Build GRCh38
rs538415951 Chr.19: 17301709-17301709 on Build GRCh38
rs771087992 Chr.19: 17301378-17301378 on Build GRCh38
rs759068394 Chr.19: 17301598-17301598 on Build GRCh38
rs146901681 Chr.19: 17301568-17301568 on Build GRCh38
rs764220135 Chr.19: 17301680-17301680 on Build GRCh38
rs141225960 Chr.19: 17301698-17301698 on Build GRCh38
rs747827990 Chr.19: 17301438-17301438 on Build GRCh38
rs540209346 Chr.19: 17301464-17301464 on Build GRCh38
rs145834574 Chr.19: 17301617-17301617 on Build GRCh38
rs760548258 Chr.19: 17301620-17301620 on Build GRCh38
rs375984470 Chr.19: 17301371-17301371 on Build GRCh38
rs749014928 Chr.19: 17301397-17301397 on Build GRCh38
rs773080943 Chr.19: 17301604-17301604 on Build GRCh38
rs912265508 Chr.19: 17301566-17301566 on Build GRCh38
rs778010815 Chr.19: 17301375-17301375 on Build GRCh38
rs771970137 Chr.19: 17301463-17301463 on Build GRCh38
rs761309621 Chr.19: 17301321-17301321 on Build GRCh38
rs773559603 Chr.19: 17301301-17301301 on Build GRCh38
rs771916946 Chr.19: 17301569-17301569 on Build GRCh38
rs561881827 Chr.19: 17301596-17301596 on Build GRCh38
rs753588501 Chr.19: 17301410-17301410 on Build GRCh38
rs781257066 Chr.19: 17301433-17301433 on Build GRCh38
rs148538259 Chr.19: 17301455-17301455 on Build GRCh38
rs904794051 Chr.19: 17301717-17301717 on Build GRCh38
rs1037237789 Chr.19: 17301387-17301387 on Build GRCh38
rs760274265 Chr.19: 17301408-17301408 on Build GRCh38
rs891822402 Chr.19: 17301686-17301686 on Build GRCh38
rs142856821 Chr.19: 17301538-17301538 on Build GRCh38
rs765645691 Chr.19: 17301523-17301523 on Build GRCh38
rs756237719 Chr.19: 17301674-17301674 on Build GRCh38
rs768277553 Chr.19: 17301443-17301443 on Build GRCh38
rs760095047 Chr.19: 17301490-17301490 on Build GRCh38
rs892039197 Chr.19: 17301484-17301484 on Build GRCh38
rs374386420 Chr.19: 17301430-17301430 on Build GRCh38
rs772071597 Chr.19: 17301571-17301571 on Build GRCh38
rs923158171 Chr.19: 17301467-17301467 on Build GRCh38
rs753140950 Chr.19: 17301527-17301527 on Build GRCh38
rs11671396 Chr.19: 17301334-17301334 on Build GRCh38
rs776950825 Chr.19: 17301396-17301396 on Build GRCh38

View Full Product Details