Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17338862-17339364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP3
Entrez Gene ID
84705
Gene Name
GTP binding protein 3 (mitochondrial)
Gene Aliases
COXPD23, GTPBG3, MSS1, MTGP1, THDF1
UniGene
Hs.334885
Gene Chromosome Location
Chr.19: 17334982-17342731 on Build GRCh38
Transcripts
NM_001128855.2 NM_001195422.1 NM_032620.3 NM_133644.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4689237 c.680A>T p.Y227F Chr.19: 17338976-17338976 on Build GRCh38
5464425 c.624C>T p.F208F Chr.19: 17338986-17338986 on Build GRCh38
6488180 c.712C>G p.Q238E Chr.19: 17339074-17339074 on Build GRCh38
5365130 c.823G>A p.G275R Chr.19: 17339185-17339185 on Build GRCh38
711263 c.789G>T p.V263V Chr.19: 17339151-17339151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760178739 Chr.19: 17339262-17339262 on Build GRCh38
rs747287650 Chr.19: 17339200-17339200 on Build GRCh38
rs762412342 Chr.19: 17339084-17339084 on Build GRCh38
rs779032068 Chr.19: 17339244-17339244 on Build GRCh38
rs768898152 Chr.19: 17339047-17339047 on Build GRCh38
rs371551886 Chr.19: 17339167-17339167 on Build GRCh38
rs780471961 Chr.19: 17339120-17339120 on Build GRCh38
rs376516285 Chr.19: 17339149-17339149 on Build GRCh38
rs764738333 Chr.19: 17338980-17338980 on Build GRCh38
rs758434694 Chr.19: 17339232-17339232 on Build GRCh38
rs752215746 Chr.19: 17338981-17338981 on Build GRCh38
rs377346963 Chr.19: 17339196-17339196 on Build GRCh38
rs763466746 Chr.19: 17339279-17339279 on Build GRCh38
rs751945111 Chr.19: 17339013-17339013 on Build GRCh38
rs774594475 Chr.19: 17339049-17339049 on Build GRCh38
rs867914093 Chr.19: 17339042-17339042 on Build GRCh38
rs770202893 Chr.19: 17339037-17339037 on Build GRCh38
rs369523370 Chr.19: 17339044-17339044 on Build GRCh38
rs551975550 Chr.19: 17339080-17339080 on Build GRCh38
rs778870846 Chr.19: 17339294-17339294 on Build GRCh38
rs766796409 Chr.19: 17339174-17339174 on Build GRCh38
rs774659468 Chr.19: 17338934-17338934 on Build GRCh38
rs761454762 Chr.19: 17339214-17339214 on Build GRCh38
rs758588126 Chr.19: 17339304-17339304 on Build GRCh38
rs202097575 Chr.19: 17338941-17338941 on Build GRCh38
rs760024091 Chr.19: 17339231-17339231 on Build GRCh38
rs768823118 Chr.19: 17338962-17338962 on Build GRCh38
rs764017367 Chr.19: 17339280-17339280 on Build GRCh38
rs759490834 Chr.19: 17338979-17338979 on Build GRCh38
rs867333219 Chr.19: 17339050-17339050 on Build GRCh38
rs759934387 Chr.19: 17339269-17339269 on Build GRCh38
rs963101147 Chr.19: 17339114-17339114 on Build GRCh38
rs752508649 Chr.19: 17339285-17339285 on Build GRCh38
rs771657097 Chr.19: 17339252-17339252 on Build GRCh38
rs370284941 Chr.19: 17339055-17339055 on Build GRCh38
rs778983997 Chr.19: 17339131-17339131 on Build GRCh38
rs556247962 Chr.19: 17339275-17339275 on Build GRCh38
rs767331683 Chr.19: 17339171-17339171 on Build GRCh38
rs776397618 Chr.19: 17339145-17339145 on Build GRCh38
rs773889548 Chr.19: 17338896-17338896 on Build GRCh38
rs760396660 Chr.19: 17339226-17339226 on Build GRCh38
rs770664002 Chr.19: 17339056-17339056 on Build GRCh38
rs1027360050 Chr.19: 17338963-17338963 on Build GRCh38
rs777862926 Chr.19: 17339305-17339305 on Build GRCh38
rs745700518 Chr.19: 17339147-17339147 on Build GRCh38
rs953458324 Chr.19: 17339148-17339148 on Build GRCh38
rs772790542 Chr.19: 17339261-17339261 on Build GRCh38
rs763165541 Chr.19: 17338953-17338953 on Build GRCh38
rs923865761 Chr.19: 17339222-17339222 on Build GRCh38
rs758057930 Chr.19: 17338990-17338990 on Build GRCh38
rs376033224 Chr.19: 17339192-17339192 on Build GRCh38
rs866503337 Chr.19: 17339259-17339259 on Build GRCh38
rs757509612 Chr.19: 17339237-17339237 on Build GRCh38
rs759514120 Chr.19: 17339078-17339078 on Build GRCh38
rs11880362 Chr.19: 17339229-17339229 on Build GRCh38
rs777142267 Chr.19: 17339315-17339315 on Build GRCh38
rs767930667 Chr.19: 17339103-17339103 on Build GRCh38
rs199820900 Chr.19: 17339298-17339298 on Build GRCh38
rs751732138 Chr.19: 17339289-17339289 on Build GRCh38
rs753749740 Chr.19: 17339237-17339237 on Build GRCh38
rs554038809 Chr.19: 17339202-17339202 on Build GRCh38
rs777342196 Chr.19: 17339008-17339008 on Build GRCh38
rs1025794329 Chr.19: 17338987-17338987 on Build GRCh38
rs537512155 Chr.19: 17339079-17339079 on Build GRCh38
rs777934121 Chr.19: 17339234-17339234 on Build GRCh38
rs764967220 Chr.19: 17339277-17339287 on Build GRCh38
rs781775396 Chr.19: 17338947-17338947 on Build GRCh38
rs866069044 Chr.19: 17338971-17338971 on Build GRCh38
rs765975578 Chr.19: 17339047-17339050 on Build GRCh38
rs761548656 Chr.19: 17339169-17339169 on Build GRCh38
rs934249813 Chr.19: 17339333-17339333 on Build GRCh38
rs779713319 Chr.19: 17339240-17339240 on Build GRCh38
rs754248289 Chr.19: 17339182-17339182 on Build GRCh38
rs990911297 Chr.19: 17338882-17338882 on Build GRCh38
rs754945883 Chr.19: 17339273-17339283 on Build GRCh38
rs370391378 Chr.19: 17339212-17339212 on Build GRCh38
rs747437738 Chr.19: 17338943-17338943 on Build GRCh38
rs961834530 Chr.19: 17339038-17339038 on Build GRCh38
rs760508696 Chr.19: 17338966-17338966 on Build GRCh38
rs746411419 Chr.19: 17338948-17338948 on Build GRCh38
rs772697417 Chr.19: 17339223-17339223 on Build GRCh38
rs750881646 Chr.19: 17339116-17339116 on Build GRCh38
rs369146685 Chr.19: 17338933-17338933 on Build GRCh38
rs781522251 Chr.19: 17339291-17339291 on Build GRCh38
rs775044613 Chr.19: 17339081-17339081 on Build GRCh38
rs771816631 Chr.19: 17339218-17339218 on Build GRCh38
rs374198861 Chr.19: 17339173-17339173 on Build GRCh38
rs757658311 Chr.19: 17339030-17339030 on Build GRCh38
rs776837688 Chr.19: 17339266-17339266 on Build GRCh38
rs200476908 Chr.19: 17339119-17339119 on Build GRCh38
rs778897687 Chr.19: 17339185-17339185 on Build GRCh38
rs533258023 Chr.19: 17338892-17338892 on Build GRCh38
rs768784673 Chr.19: 17339211-17339211 on Build GRCh38
rs150612889 Chr.19: 17338965-17338965 on Build GRCh38
rs771544436 Chr.19: 17338945-17338945 on Build GRCh38
rs751211186 Chr.19: 17339088-17339088 on Build GRCh38
rs140048775 Chr.19: 17339184-17339184 on Build GRCh38
rs752344800 Chr.19: 17338940-17338940 on Build GRCh38
rs780173916 Chr.19: 17339043-17339043 on Build GRCh38
rs949520450 Chr.19: 17339258-17339258 on Build GRCh38
rs776429455 Chr.19: 17338921-17338921 on Build GRCh38
rs775449994 Chr.19: 17338951-17338951 on Build GRCh38
rs751677292 Chr.19: 17339235-17339235 on Build GRCh38
rs763644514 Chr.19: 17339157-17339157 on Build GRCh38
rs987865664 Chr.19: 17339216-17339216 on Build GRCh38
rs201374694 Chr.19: 17338949-17338949 on Build GRCh38
rs45541332 Chr.19: 17338890-17338890 on Build GRCh38
rs754776343 Chr.19: 17339293-17339293 on Build GRCh38
rs775371901 Chr.19: 17339204-17339204 on Build GRCh38
rs3810206 Chr.19: 17339207-17339207 on Build GRCh38
rs893912860 Chr.19: 17338954-17338954 on Build GRCh38
rs763730644 Chr.19: 17339085-17339085 on Build GRCh38
rs759990229 Chr.19: 17339243-17339243 on Build GRCh38
rs923793422 Chr.19: 17338938-17338938 on Build GRCh38
rs368666036 Chr.19: 17338920-17338920 on Build GRCh38
rs772049205 Chr.19: 17339303-17339303 on Build GRCh38
rs748124728 Chr.19: 17339251-17339251 on Build GRCh38
rs373370177 Chr.19: 17339123-17339123 on Build GRCh38
rs544669646 Chr.19: 17339035-17339035 on Build GRCh38
rs775253637 Chr.19: 17339150-17339150 on Build GRCh38
rs534233238 Chr.19: 17339198-17339198 on Build GRCh38
rs753650613 Chr.19: 17339292-17339292 on Build GRCh38
rs201491919 Chr.19: 17339265-17339265 on Build GRCh38
rs568919152 Chr.19: 17339057-17339057 on Build GRCh38
rs754233717 Chr.19: 17339230-17339230 on Build GRCh38
rs772348422 Chr.19: 17339140-17339140 on Build GRCh38
rs376546164 Chr.19: 17338922-17338922 on Build GRCh38
rs754970973 Chr.19: 17339130-17339130 on Build GRCh38
rs748311348 Chr.19: 17339137-17339137 on Build GRCh38
rs557723263 Chr.19: 17338921-17338922 on Build GRCh38
rs1004190940 Chr.19: 17339302-17339302 on Build GRCh38
rs149839355 Chr.19: 17338976-17338976 on Build GRCh38
rs762972282 Chr.19: 17339155-17339155 on Build GRCh38
rs139997629 Chr.19: 17338972-17338972 on Build GRCh38

View Full Product Details