Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17340882-17341371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP3
Entrez Gene ID
84705
Gene Name
GTP binding protein 3 (mitochondrial)
Gene Aliases
COXPD23, GTPBG3, MSS1, MTGP1, THDF1
UniGene
Hs.334885
Gene Chromosome Location
Chr.19: 17334982-17342731 on Build GRCh38
Transcripts
NM_001128855.2 NM_001195422.1 NM_032620.3 NM_133644.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676918 c.1241C>T p.P414L Chr.19: 17341244-17341244 on Build GRCh38
128124 c.1161C>T p.A387A Chr.19: 17341134-17341134 on Build GRCh38
6386919 c.1196A>T p.Q399L Chr.19: 17341169-17341169 on Build GRCh38
6276228 c.1209G>T p.L403L Chr.19: 17341182-17341182 on Build GRCh38
5683143 c.1268G>T p.G423V Chr.19: 17341271-17341271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753884625 Chr.19: 17341330-17341330 on Build GRCh38
rs763886108 Chr.19: 17341225-17341225 on Build GRCh38
rs200027460 Chr.19: 17341299-17341299 on Build GRCh38
rs769133364 Chr.19: 17341099-17341099 on Build GRCh38
rs759530754 Chr.19: 17341200-17341200 on Build GRCh38
rs138116759 Chr.19: 17341074-17341074 on Build GRCh38
rs144236062 Chr.19: 17341245-17341245 on Build GRCh38
rs781586705 Chr.19: 17341129-17341129 on Build GRCh38
rs879173908 Chr.19: 17340944-17340944 on Build GRCh38
rs77401427 Chr.19: 17341126-17341126 on Build GRCh38
rs762588490 Chr.19: 17341271-17341271 on Build GRCh38
rs779147862 Chr.19: 17341255-17341255 on Build GRCh38
rs148291786 Chr.19: 17341321-17341321 on Build GRCh38
rs931123101 Chr.19: 17341304-17341304 on Build GRCh38
rs374901243 Chr.19: 17341005-17341005 on Build GRCh38
rs141041710 Chr.19: 17341083-17341083 on Build GRCh38
rs757903626 Chr.19: 17341038-17341038 on Build GRCh38
rs759009222 Chr.19: 17341095-17341095 on Build GRCh38
rs754647739 Chr.19: 17341014-17341014 on Build GRCh38
rs1018519058 Chr.19: 17340979-17340979 on Build GRCh38
rs3745193 Chr.19: 17341172-17341172 on Build GRCh38
rs758786169 Chr.19: 17341019-17341019 on Build GRCh38
rs76171010 Chr.19: 17340921-17340921 on Build GRCh38
rs372394605 Chr.19: 17341281-17341281 on Build GRCh38
rs769827042 Chr.19: 17341269-17341269 on Build GRCh38
rs149578279 Chr.19: 17341244-17341244 on Build GRCh38
rs753358209 Chr.19: 17341009-17341009 on Build GRCh38
rs375380416 Chr.19: 17341337-17341337 on Build GRCh38
rs751938919 Chr.19: 17341227-17341227 on Build GRCh38
rs979235492 Chr.19: 17340929-17340929 on Build GRCh38
rs200990676 Chr.19: 17341209-17341209 on Build GRCh38
rs1035729758 Chr.19: 17341125-17341125 on Build GRCh38
rs77433229 Chr.19: 17340961-17340961 on Build GRCh38
rs746478858 Chr.19: 17341236-17341236 on Build GRCh38
rs563812528 Chr.19: 17340948-17340948 on Build GRCh38
rs374190574 Chr.19: 17341151-17341151 on Build GRCh38
rs775943192 Chr.19: 17341024-17341025 on Build GRCh38
rs774094357 Chr.19: 17341286-17341286 on Build GRCh38
rs866266746 Chr.19: 17340976-17340976 on Build GRCh38
rs886041488 Chr.19: 17341118-17341118 on Build GRCh38
rs536040222 Chr.19: 17341179-17341179 on Build GRCh38
rs760604467 Chr.19: 17341077-17341077 on Build GRCh38
rs993846552 Chr.19: 17340936-17340936 on Build GRCh38
rs772729996 Chr.19: 17341325-17341326 on Build GRCh38
rs749623578 Chr.19: 17341032-17341032 on Build GRCh38
rs750870357 Chr.19: 17341305-17341305 on Build GRCh38
rs200470221 Chr.19: 17341173-17341173 on Build GRCh38
rs144317680 Chr.19: 17340999-17340999 on Build GRCh38
rs757518953 Chr.19: 17341228-17341228 on Build GRCh38
rs775205156 Chr.19: 17341213-17341213 on Build GRCh38
rs745981620 Chr.19: 17341048-17341048 on Build GRCh38
rs760421275 Chr.19: 17341327-17341328 on Build GRCh38
rs756102331 Chr.19: 17341138-17341138 on Build GRCh38
rs764833866 Chr.19: 17341103-17341103 on Build GRCh38
rs758712952 Chr.19: 17341348-17341348 on Build GRCh38
rs879527410 Chr.19: 17341311-17341311 on Build GRCh38
rs932295710 Chr.19: 17340945-17340945 on Build GRCh38
rs939979516 Chr.19: 17340988-17340988 on Build GRCh38
rs147146117 Chr.19: 17340941-17340941 on Build GRCh38
rs921027194 Chr.19: 17340940-17340940 on Build GRCh38
rs770871640 Chr.19: 17341181-17341181 on Build GRCh38
rs755011281 Chr.19: 17341334-17341334 on Build GRCh38
rs764584318 Chr.19: 17341212-17341212 on Build GRCh38
rs75893526 Chr.19: 17340927-17340927 on Build GRCh38
rs148490454 Chr.19: 17340918-17340918 on Build GRCh38
rs368566426 Chr.19: 17341020-17341020 on Build GRCh38
rs752692402 Chr.19: 17341042-17341042 on Build GRCh38
rs372422729 Chr.19: 17341188-17341188 on Build GRCh38
rs1036879572 Chr.19: 17341257-17341257 on Build GRCh38
rs757813646 Chr.19: 17341128-17341128 on Build GRCh38
rs372801486 Chr.19: 17341106-17341106 on Build GRCh38
rs747452180 Chr.19: 17341026-17341026 on Build GRCh38
rs779997383 Chr.19: 17341240-17341240 on Build GRCh38
rs750315195 Chr.19: 17340998-17340998 on Build GRCh38
rs781044822 Chr.19: 17341261-17341261 on Build GRCh38
rs762680174 Chr.19: 17341224-17341224 on Build GRCh38
rs879042978 Chr.19: 17340942-17340942 on Build GRCh38
rs776184112 Chr.19: 17341091-17341091 on Build GRCh38
rs886037735 Chr.19: 17341078-17341078 on Build GRCh38
rs770313292 Chr.19: 17341331-17341332 on Build GRCh38
rs753074664 Chr.19: 17341016-17341016 on Build GRCh38
rs770023317 Chr.19: 17341052-17341052 on Build GRCh38
rs745788110 Chr.19: 17341262-17341262 on Build GRCh38
rs376954517 Chr.19: 17341259-17341259 on Build GRCh38
rs778986039 Chr.19: 17341336-17341336 on Build GRCh38
rs771800563 Chr.19: 17341067-17341067 on Build GRCh38
rs768309067 Chr.19: 17341285-17341285 on Build GRCh38
rs775893367 Chr.19: 17341239-17341239 on Build GRCh38
rs960584148 Chr.19: 17341092-17341092 on Build GRCh38
rs749538509 Chr.19: 17341060-17341060 on Build GRCh38
rs778479594 Chr.19: 17341015-17341015 on Build GRCh38
rs761075836 Chr.19: 17341316-17341316 on Build GRCh38
rs144775733 Chr.19: 17341153-17341153 on Build GRCh38
rs752233769 Chr.19: 17341105-17341105 on Build GRCh38
rs1021789483 Chr.19: 17341154-17341154 on Build GRCh38
rs781316086 Chr.19: 17341039-17341039 on Build GRCh38

View Full Product Details