Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17533509-17534012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM129C
Entrez Gene ID
199786
Gene Name
family with sequence similarity 129 member C
Gene Aliases
BCNP1
UniGene
Hs.434133
Gene Chromosome Location
Chr.19: 17523301-17556755 on Build GRCh38
Transcripts
NM_001098524.1 NM_001321826.1 NM_001321827.1 NM_001321828.1 NM_173544.4 XM_005259813.3 XM_005259814.3 XM_011527781.2 XM_011527786.2 XM_011527787.2 XM_011527789.1 XM_017026453.1 XM_017026454.1 XM_017026455.1 XM_017026456.1 XM_017026457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992817 c.412G>A p.E138K Chr.19: 17533593-17533593 on Build GRCh38
3286250 c.488G>A p.R163H Chr.19: 17533669-17533669 on Build GRCh38
6898591 c.475C>T p.R159* Chr.19: 17533656-17533656 on Build GRCh38
6465618 c.437_438delCA p.S146fs*25 Chr.19: 17533618-17533619 on Build GRCh38
4075373 c.510T>A p.P170P Chr.19: 17533691-17533691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773780731 Chr.19: 17533681-17533681 on Build GRCh38
rs942370595 Chr.19: 17533866-17533866 on Build GRCh38
rs942397171 Chr.19: 17533651-17533651 on Build GRCh38
rs749451070 Chr.19: 17533545-17533545 on Build GRCh38
rs922550346 Chr.19: 17533868-17533868 on Build GRCh38
rs745777584 Chr.19: 17533587-17533587 on Build GRCh38
rs543604873 Chr.19: 17533683-17533683 on Build GRCh38
rs867638811 Chr.19: 17533923-17533923 on Build GRCh38
rs766906157 Chr.19: 17533560-17533561 on Build GRCh38
rs762468692 Chr.19: 17533601-17533601 on Build GRCh38
rs200102226 Chr.19: 17533538-17533538 on Build GRCh38
rs756685544 Chr.19: 17533633-17533633 on Build GRCh38
rs554974229 Chr.19: 17533959-17533959 on Build GRCh38
rs1025009992 Chr.19: 17533751-17533751 on Build GRCh38
rs556326086 Chr.19: 17533762-17533762 on Build GRCh38
rs1031786217 Chr.19: 17533788-17533788 on Build GRCh38
rs1038868305 Chr.19: 17533915-17533915 on Build GRCh38
rs752844228 Chr.19: 17533738-17533738 on Build GRCh38
rs370655406 Chr.19: 17533971-17533974 on Build GRCh38
rs769793668 Chr.19: 17533598-17533598 on Build GRCh38
rs189951684 Chr.19: 17533740-17533740 on Build GRCh38
rs184472022 Chr.19: 17533668-17533668 on Build GRCh38
rs371501512 Chr.19: 17533779-17533779 on Build GRCh38
rs572540834 Chr.19: 17533880-17533880 on Build GRCh38
rs143255653 Chr.19: 17533655-17533655 on Build GRCh38
rs35901148 Chr.19: 17533555-17533556 on Build GRCh38
rs748350394 Chr.19: 17533656-17533656 on Build GRCh38
rs1035803347 Chr.19: 17533715-17533715 on Build GRCh38
rs150700269 Chr.19: 17533599-17533599 on Build GRCh38
rs530989121 Chr.19: 17533712-17533712 on Build GRCh38
rs140293166 Chr.19: 17533640-17533640 on Build GRCh38
rs941133216 Chr.19: 17533907-17533907 on Build GRCh38
rs1019740874 Chr.19: 17533946-17533946 on Build GRCh38
rs779135011 Chr.19: 17533559-17533559 on Build GRCh38
rs114011143 Chr.19: 17533793-17533793 on Build GRCh38
rs768164581 Chr.19: 17533680-17533680 on Build GRCh38
rs542958092 Chr.19: 17533988-17533988 on Build GRCh38
rs74387759 Chr.19: 17533625-17533625 on Build GRCh38
rs975099680 Chr.19: 17533867-17533867 on Build GRCh38
rs766963992 Chr.19: 17533639-17533639 on Build GRCh38
rs575644922 Chr.19: 17533580-17533580 on Build GRCh38
rs761578885 Chr.19: 17533620-17533620 on Build GRCh38
rs763519140 Chr.19: 17533607-17533607 on Build GRCh38
rs201345901 Chr.19: 17533657-17533657 on Build GRCh38
rs761059706 Chr.19: 17533713-17533713 on Build GRCh38
rs774017362 Chr.19: 17533614-17533614 on Build GRCh38
rs986929887 Chr.19: 17533785-17533785 on Build GRCh38
rs771916611 Chr.19: 17533709-17533709 on Build GRCh38
rs754327565 Chr.19: 17533725-17533725 on Build GRCh38
rs886958015 Chr.19: 17533888-17533888 on Build GRCh38
rs541062560 Chr.19: 17533916-17533916 on Build GRCh38
rs773450161 Chr.19: 17533945-17533946 on Build GRCh38
rs914905381 Chr.19: 17533630-17533630 on Build GRCh38
rs770670280 Chr.19: 17533566-17533566 on Build GRCh38
rs755015013 Chr.19: 17533647-17533647 on Build GRCh38
rs558790572 Chr.19: 17533798-17533798 on Build GRCh38
rs770255100 Chr.19: 17533776-17533776 on Build GRCh38
rs1057455602 Chr.19: 17533829-17533829 on Build GRCh38
rs1008066952 Chr.19: 17533946-17533957 on Build GRCh38
rs573966930 Chr.19: 17533971-17533971 on Build GRCh38
rs933878900 Chr.19: 17533876-17533876 on Build GRCh38
rs762733475 Chr.19: 17533736-17533736 on Build GRCh38
rs749713110 Chr.19: 17533906-17533906 on Build GRCh38
rs924756087 Chr.19: 17533819-17533819 on Build GRCh38
rs973994274 Chr.19: 17533778-17533778 on Build GRCh38
rs370043401 Chr.19: 17533669-17533669 on Build GRCh38
rs147499098 Chr.19: 17533678-17533678 on Build GRCh38
rs765331085 Chr.19: 17533730-17533730 on Build GRCh38
rs761525724 Chr.19: 17533700-17533700 on Build GRCh38
rs775259922 Chr.19: 17533651-17533651 on Build GRCh38
rs914959681 Chr.19: 17533822-17533822 on Build GRCh38
rs551210423 Chr.19: 17533564-17533564 on Build GRCh38
rs767969911 Chr.19: 17533622-17533622 on Build GRCh38
rs1013370589 Chr.19: 17533957-17533957 on Build GRCh38
rs1046961454 Chr.19: 17533878-17533878 on Build GRCh38
rs371364428 Chr.19: 17533946-17533949 on Build GRCh38
rs766653053 Chr.19: 17533714-17533714 on Build GRCh38
rs771774754 Chr.19: 17533949-17533950 on Build GRCh38
rs759980761 Chr.19: 17533728-17533728 on Build GRCh38
rs951258960 Chr.19: 17533946-17533953 on Build GRCh38
rs569934561 Chr.19: 17533787-17533787 on Build GRCh38

View Full Product Details