Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18173679-18174183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI30
Entrez Gene ID
10437
Gene Name
IFI30, lysosomal thiol reductase
Gene Aliases
GILT, IFI-30, IP-30, IP30
UniGene
Hs.14623
Gene Chromosome Location
Chr.19: 18173780-18178124 on Build GRCh38
Transcripts
NM_006332.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611823 c.6C>A p.T2T Chr.19: 18173847-18173847 on Build GRCh38
7219895 c.96A>G p.L32L Chr.19: 18173937-18173937 on Build GRCh38
6999100 c.132+6G>A p.? Chr.19: 18173979-18173979 on Build GRCh38
6458489 c.36_38delGCT p.L17delL Chr.19: 18173877-18173879 on Build GRCh38
4913394 c.6C>T p.T2T Chr.19: 18173847-18173847 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965626050 Chr.19: 18174033-18174033 on Build GRCh38
rs966915885 Chr.19: 18173932-18173932 on Build GRCh38
rs1033555629 Chr.19: 18173726-18173726 on Build GRCh38
rs764788458 Chr.19: 18173892-18173892 on Build GRCh38
rs901813214 Chr.19: 18173720-18173720 on Build GRCh38
rs757222810 Chr.19: 18173961-18173961 on Build GRCh38
rs985734167 Chr.19: 18173790-18173790 on Build GRCh38
rs957909402 Chr.19: 18173789-18173789 on Build GRCh38
rs764733870 Chr.19: 18173808-18173808 on Build GRCh38
rs778500182 Chr.19: 18173933-18173933 on Build GRCh38
rs376122150 Chr.19: 18173906-18173906 on Build GRCh38
rs531634659 Chr.19: 18173826-18173826 on Build GRCh38
rs943964796 Chr.19: 18173917-18173917 on Build GRCh38
rs533709947 Chr.19: 18173907-18173907 on Build GRCh38
rs535891325 Chr.19: 18174019-18174019 on Build GRCh38
rs375812223 Chr.19: 18173774-18173774 on Build GRCh38
rs745317270 Chr.19: 18173984-18173984 on Build GRCh38
rs571435275 Chr.19: 18173905-18173905 on Build GRCh38
rs1014258499 Chr.19: 18174144-18174144 on Build GRCh38
rs1027011651 Chr.19: 18173706-18173706 on Build GRCh38
rs779724198 Chr.19: 18173757-18173757 on Build GRCh38
rs761052681 Chr.19: 18173880-18173882 on Build GRCh38
rs930095307 Chr.19: 18174074-18174074 on Build GRCh38
rs966473110 Chr.19: 18173759-18173759 on Build GRCh38
rs531983625 Chr.19: 18174083-18174083 on Build GRCh38
rs1024831735 Chr.19: 18173926-18173926 on Build GRCh38
rs1029041184 Chr.19: 18173751-18173751 on Build GRCh38
rs746282770 Chr.19: 18174018-18174018 on Build GRCh38
rs1031601996 Chr.19: 18174114-18174114 on Build GRCh38
rs777124188 Chr.19: 18173998-18173998 on Build GRCh38
rs374468027 Chr.19: 18173993-18173993 on Build GRCh38
rs937426086 Chr.19: 18174123-18174123 on Build GRCh38
rs959271434 Chr.19: 18173734-18173734 on Build GRCh38
rs903933851 Chr.19: 18174098-18174098 on Build GRCh38
rs747408465 Chr.19: 18173977-18173977 on Build GRCh38
rs373433112 Chr.19: 18173877-18173877 on Build GRCh38
rs776312259 Chr.19: 18173877-18173882 on Build GRCh38
rs771537773 Chr.19: 18173991-18173991 on Build GRCh38
rs370440721 Chr.19: 18173973-18173973 on Build GRCh38
rs1001072455 Chr.19: 18174100-18174100 on Build GRCh38
rs373154530 Chr.19: 18173834-18173834 on Build GRCh38
rs918769229 Chr.19: 18174046-18174046 on Build GRCh38
rs999291819 Chr.19: 18173940-18173940 on Build GRCh38
rs766452229 Chr.19: 18173871-18173871 on Build GRCh38
rs748974019 Chr.19: 18173805-18173805 on Build GRCh38
rs1050532089 Chr.19: 18174093-18174093 on Build GRCh38
rs770107752 Chr.19: 18174045-18174045 on Build GRCh38
rs892613815 Chr.19: 18174115-18174115 on Build GRCh38
rs749705374 Chr.19: 18174022-18174022 on Build GRCh38
rs978221201 Chr.19: 18173764-18173764 on Build GRCh38
rs992396315 Chr.19: 18173809-18173809 on Build GRCh38
rs888423002 Chr.19: 18173718-18173718 on Build GRCh38
rs1025081902 Chr.19: 18173822-18173822 on Build GRCh38
rs273263 Chr.19: 18173754-18173754 on Build GRCh38
rs1001978587 Chr.19: 18173847-18173847 on Build GRCh38
rs996278392 Chr.19: 18173750-18173750 on Build GRCh38
rs909989185 Chr.19: 18174112-18174112 on Build GRCh38
rs780480112 Chr.19: 18173877-18173879 on Build GRCh38
rs1013490269 Chr.19: 18173915-18173915 on Build GRCh38
rs924984493 Chr.19: 18173776-18173776 on Build GRCh38
rs761933196 Chr.19: 18173832-18173832 on Build GRCh38
rs11554160 Chr.19: 18173937-18173937 on Build GRCh38
rs1041228135 Chr.19: 18173818-18173818 on Build GRCh38
rs1055826762 Chr.19: 18174160-18174160 on Build GRCh38
rs557270747 Chr.19: 18174015-18174015 on Build GRCh38
rs1055793899 Chr.19: 18173862-18173862 on Build GRCh38
rs547116614 Chr.19: 18174014-18174014 on Build GRCh38
rs575909363 Chr.19: 18174120-18174120 on Build GRCh38
rs1040763899 Chr.19: 18173717-18173717 on Build GRCh38
rs2241089 Chr.19: 18173841-18173841 on Build GRCh38
rs985125761 Chr.19: 18173990-18173990 on Build GRCh38
rs369358763 Chr.19: 18173845-18173845 on Build GRCh38
rs751571473 Chr.19: 18173960-18173960 on Build GRCh38
rs1006144344 Chr.19: 18173719-18173719 on Build GRCh38

View Full Product Details