Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18177569-18178085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI30
Entrez Gene ID
10437
Gene Name
IFI30, lysosomal thiol reductase
Gene Aliases
GILT, IFI-30, IP-30, IP30
UniGene
Hs.14623
Gene Chromosome Location
Chr.19: 18173780-18178124 on Build GRCh38
Transcripts
NM_006332.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830472 c.721A>G p.T241A Chr.19: 18177879-18177879 on Build GRCh38
5389110 c.679C>T p.Q227* Chr.19: 18177753-18177753 on Build GRCh38
4750644 c.750G>C p.K250N Chr.19: 18177908-18177908 on Build GRCh38
5389109 c.678C>T p.C226C Chr.19: 18177752-18177752 on Build GRCh38
3530738 c.668C>T p.T223I Chr.19: 18177742-18177742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988546380 Chr.19: 18178002-18178002 on Build GRCh38
rs149650847 Chr.19: 18177598-18177598 on Build GRCh38
rs551748369 Chr.19: 18177917-18177917 on Build GRCh38
rs368871171 Chr.19: 18177715-18177715 on Build GRCh38
rs945955468 Chr.19: 18178016-18178016 on Build GRCh38
rs560326328 Chr.19: 18177729-18177729 on Build GRCh38
rs747099778 Chr.19: 18177875-18177875 on Build GRCh38
rs752472421 Chr.19: 18177687-18177687 on Build GRCh38
rs754588977 Chr.19: 18177867-18177867 on Build GRCh38
rs369094218 Chr.19: 18177672-18177674 on Build GRCh38
rs199599038 Chr.19: 18177666-18177666 on Build GRCh38
rs776970950 Chr.19: 18177825-18177825 on Build GRCh38
rs774342582 Chr.19: 18177926-18177926 on Build GRCh38
rs15953 Chr.19: 18177945-18177945 on Build GRCh38
rs753515072 Chr.19: 18177866-18177866 on Build GRCh38
rs775483671 Chr.19: 18177742-18177742 on Build GRCh38
rs575168573 Chr.19: 18177660-18177660 on Build GRCh38
rs753050018 Chr.19: 18177869-18177869 on Build GRCh38
rs1009129816 Chr.19: 18177638-18177638 on Build GRCh38
rs15057 Chr.19: 18178012-18178012 on Build GRCh38
rs868346112 Chr.19: 18177916-18177916 on Build GRCh38
rs766445246 Chr.19: 18177829-18177829 on Build GRCh38
rs938875484 Chr.19: 18177610-18177610 on Build GRCh38
rs751735830 Chr.19: 18177707-18177708 on Build GRCh38
rs575384663 Chr.19: 18178030-18178030 on Build GRCh38
rs745539034 Chr.19: 18177694-18177694 on Build GRCh38
rs757487351 Chr.19: 18177759-18177760 on Build GRCh38
rs949016756 Chr.19: 18177635-18177635 on Build GRCh38
rs61730355 Chr.19: 18177730-18177730 on Build GRCh38
rs747099566 Chr.19: 18177806-18177806 on Build GRCh38
rs566703314 Chr.19: 18177989-18177989 on Build GRCh38
rs767816568 Chr.19: 18177667-18177667 on Build GRCh38
rs928955252 Chr.19: 18177650-18177650 on Build GRCh38
rs1803813 Chr.19: 18177731-18177731 on Build GRCh38
rs760900335 Chr.19: 18177678-18177678 on Build GRCh38
rs749965375 Chr.19: 18177859-18177859 on Build GRCh38
rs77198486 Chr.19: 18177986-18177986 on Build GRCh38
rs779256288 Chr.19: 18177868-18177868 on Build GRCh38
rs539760169 Chr.19: 18177933-18177933 on Build GRCh38
rs77583590 Chr.19: 18177832-18177832 on Build GRCh38
rs751564815 Chr.19: 18177751-18177751 on Build GRCh38
rs375674283 Chr.19: 18177675-18177675 on Build GRCh38
rs773175557 Chr.19: 18177936-18177936 on Build GRCh38
rs3185630 Chr.19: 18178055-18178055 on Build GRCh38
rs374054204 Chr.19: 18177594-18177594 on Build GRCh38
rs370382349 Chr.19: 18177659-18177659 on Build GRCh38
rs766073076 Chr.19: 18177865-18177865 on Build GRCh38
rs1027805866 Chr.19: 18177943-18177943 on Build GRCh38
rs771126518 Chr.19: 18177813-18177813 on Build GRCh38
rs898837198 Chr.19: 18177642-18177642 on Build GRCh38
rs1038317371 Chr.19: 18177637-18177637 on Build GRCh38
rs781380689 Chr.19: 18177883-18177883 on Build GRCh38
rs555420144 Chr.19: 18177606-18177606 on Build GRCh38
rs372940357 Chr.19: 18177663-18177663 on Build GRCh38
rs755776437 Chr.19: 18177764-18177764 on Build GRCh38
rs989782908 Chr.19: 18177593-18177593 on Build GRCh38
rs1028604703 Chr.19: 18177605-18177605 on Build GRCh38
rs367547762 Chr.19: 18177843-18177843 on Build GRCh38
rs370566972 Chr.19: 18177688-18177688 on Build GRCh38
rs2921 Chr.19: 18178000-18178000 on Build GRCh38
rs749642774 Chr.19: 18177717-18177717 on Build GRCh38
rs767473145 Chr.19: 18177858-18177858 on Build GRCh38
rs773179563 Chr.19: 18177803-18177803 on Build GRCh38
rs992942875 Chr.19: 18177621-18177621 on Build GRCh38
rs762836929 Chr.19: 18177743-18177743 on Build GRCh38
rs766939783 Chr.19: 18177759-18177759 on Build GRCh38
rs571503439 Chr.19: 18177927-18177927 on Build GRCh38
rs764970320 Chr.19: 18177685-18177685 on Build GRCh38
rs375751473 Chr.19: 18177795-18177795 on Build GRCh38
rs761666660 Chr.19: 18177849-18177849 on Build GRCh38
rs748272652 Chr.19: 18177726-18177726 on Build GRCh38
rs372016608 Chr.19: 18177831-18177831 on Build GRCh38
rs759782481 Chr.19: 18177828-18177828 on Build GRCh38
rs374435415 Chr.19: 18177690-18177690 on Build GRCh38
rs751790931 Chr.19: 18177944-18177944 on Build GRCh38
rs191513645 Chr.19: 18177911-18177911 on Build GRCh38
rs768042602 Chr.19: 18177860-18177860 on Build GRCh38
rs369048103 Chr.19: 18177737-18177737 on Build GRCh38
rs760196559 Chr.19: 18177860-18177860 on Build GRCh38
rs746091568 Chr.19: 18177891-18177891 on Build GRCh38
rs916050103 Chr.19: 18177625-18177625 on Build GRCh38
rs1042896295 Chr.19: 18178040-18178040 on Build GRCh38
rs772565792 Chr.19: 18177801-18177801 on Build GRCh38
rs766567273 Chr.19: 18177738-18177738 on Build GRCh38
rs749689536 Chr.19: 18177794-18177794 on Build GRCh38
rs757176576 Chr.19: 18177758-18177758 on Build GRCh38
rs755323609 Chr.19: 18177978-18177978 on Build GRCh38
rs200628484 Chr.19: 18177880-18177880 on Build GRCh38
rs998795867 Chr.19: 18177807-18177807 on Build GRCh38
rs555213418 Chr.19: 18178010-18178010 on Build GRCh38
rs184164860 Chr.19: 18177957-18177957 on Build GRCh38
rs758888528 Chr.19: 18177870-18177870 on Build GRCh38
rs778340765 Chr.19: 18177872-18177872 on Build GRCh38
rs202043324 Chr.19: 18177711-18177711 on Build GRCh38
rs913061259 Chr.19: 18177765-18177765 on Build GRCh38
rs779673125 Chr.19: 18177786-18177786 on Build GRCh38
rs995835125 Chr.19: 18177599-18177599 on Build GRCh38
rs749888818 Chr.19: 18177761-18177761 on Build GRCh38
rs764176353 Chr.19: 18177744-18177744 on Build GRCh38
rs368357390 Chr.19: 18177797-18177797 on Build GRCh38
rs980791122 Chr.19: 18177702-18177702 on Build GRCh38

View Full Product Details