Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18596764-18597289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLF1
Entrez Gene ID
9244
Gene Name
cytokine receptor like factor 1
Gene Aliases
CISS, CISS1, CLF, CLF-1, NR6, zcytor5
UniGene
Hs.114948
Gene Chromosome Location
Chr.19: 18593225-18606850 on Build GRCh38
Transcripts
NM_004750.4 XM_011528422.2 XM_011528423.2 XM_011528424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4914123 c.811A>T p.K271* Chr.19: 18596936-18596936 on Build GRCh38
361954 c.713C>T p.P238L Chr.19: 18597034-18597034 on Build GRCh38
3530830 c.717C>T p.P239P Chr.19: 18597030-18597030 on Build GRCh38
5529221 c.852G>A p.W284* Chr.19: 18596895-18596895 on Build GRCh38
1641100 c.827A>G p.Y276C Chr.19: 18596920-18596920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753349471 Chr.19: 18597062-18597062 on Build GRCh38
rs758261006 Chr.19: 18596913-18596913 on Build GRCh38
rs748525943 Chr.19: 18597009-18597009 on Build GRCh38
rs374977336 Chr.19: 18596876-18596876 on Build GRCh38
rs137853145 Chr.19: 18596918-18596918 on Build GRCh38
rs764501088 Chr.19: 18596871-18596871 on Build GRCh38
rs10401510 Chr.19: 18596842-18596842 on Build GRCh38
rs78504242 Chr.19: 18596826-18596826 on Build GRCh38
rs1053965602 Chr.19: 18597026-18597026 on Build GRCh38
rs542145297 Chr.19: 18597142-18597142 on Build GRCh38
rs770368637 Chr.19: 18596864-18596864 on Build GRCh38
rs1010586021 Chr.19: 18597013-18597013 on Build GRCh38
rs373093397 Chr.19: 18597205-18597208 on Build GRCh38
rs553291137 Chr.19: 18597118-18597118 on Build GRCh38
rs1005429225 Chr.19: 18597104-18597104 on Build GRCh38
rs962139480 Chr.19: 18597125-18597125 on Build GRCh38
rs769138565 Chr.19: 18597138-18597138 on Build GRCh38
rs139919292 Chr.19: 18596981-18596981 on Build GRCh38
rs745392621 Chr.19: 18596923-18596923 on Build GRCh38
rs369678434 Chr.19: 18596796-18596796 on Build GRCh38
rs780531297 Chr.19: 18596919-18596919 on Build GRCh38
rs7250623 Chr.19: 18597163-18597163 on Build GRCh38
rs763321938 Chr.19: 18596867-18596867 on Build GRCh38
rs548602967 Chr.19: 18596841-18596841 on Build GRCh38
rs775250588 Chr.19: 18596845-18596845 on Build GRCh38
rs67567163 Chr.19: 18597202-18597205 on Build GRCh38
rs753110535 Chr.19: 18597150-18597151 on Build GRCh38
rs557461110 Chr.19: 18597046-18597046 on Build GRCh38
rs768979541 Chr.19: 18596939-18596939 on Build GRCh38
rs746314944 Chr.19: 18596858-18596858 on Build GRCh38
rs373476055 Chr.19: 18597059-18597059 on Build GRCh38
rs752757281 Chr.19: 18596857-18596857 on Build GRCh38
rs954756781 Chr.19: 18596806-18596806 on Build GRCh38
rs147108114 Chr.19: 18596927-18596927 on Build GRCh38
rs745810107 Chr.19: 18596817-18596818 on Build GRCh38
rs776788550 Chr.19: 18597101-18597101 on Build GRCh38
rs7247346 Chr.19: 18597068-18597068 on Build GRCh38
rs762946618 Chr.19: 18597028-18597028 on Build GRCh38
rs756906640 Chr.19: 18596984-18596984 on Build GRCh38
rs74447334 Chr.19: 18596881-18596881 on Build GRCh38
rs767283349 Chr.19: 18597042-18597042 on Build GRCh38
rs776875106 Chr.19: 18597024-18597024 on Build GRCh38
rs543919645 Chr.19: 18597240-18597240 on Build GRCh38
rs773474924 Chr.19: 18597014-18597014 on Build GRCh38
rs899691434 Chr.19: 18597039-18597039 on Build GRCh38
rs774678811 Chr.19: 18597060-18597061 on Build GRCh38
rs142531140 Chr.19: 18596986-18596986 on Build GRCh38
rs778532329 Chr.19: 18596823-18596823 on Build GRCh38
rs749861042 Chr.19: 18596932-18596932 on Build GRCh38
rs752647111 Chr.19: 18596970-18596970 on Build GRCh38
rs755209911 Chr.19: 18596840-18596840 on Build GRCh38
rs754806079 Chr.19: 18596890-18596890 on Build GRCh38
rs760844216 Chr.19: 18597021-18597021 on Build GRCh38
rs878973438 Chr.19: 18597175-18597175 on Build GRCh38
rs894095040 Chr.19: 18597027-18597027 on Build GRCh38
rs574866353 Chr.19: 18597202-18597202 on Build GRCh38
rs762070024 Chr.19: 18596880-18596880 on Build GRCh38
rs907992641 Chr.19: 18596820-18596820 on Build GRCh38
rs772375122 Chr.19: 18597011-18597011 on Build GRCh38
rs137853928 Chr.19: 18596902-18596903 on Build GRCh38
rs1021454872 Chr.19: 18596821-18596821 on Build GRCh38
rs955273434 Chr.19: 18597167-18597167 on Build GRCh38
rs772447610 Chr.19: 18597076-18597076 on Build GRCh38
rs760792044 Chr.19: 18596962-18596962 on Build GRCh38
rs910731707 Chr.19: 18597020-18597020 on Build GRCh38
rs939557535 Chr.19: 18597185-18597185 on Build GRCh38
rs539995719 Chr.19: 18597110-18597110 on Build GRCh38
rs769181592 Chr.19: 18597000-18597000 on Build GRCh38
rs749171844 Chr.19: 18596853-18596856 on Build GRCh38
rs745894453 Chr.19: 18596846-18596846 on Build GRCh38
rs750546273 Chr.19: 18596886-18596886 on Build GRCh38
rs747979764 Chr.19: 18596824-18596824 on Build GRCh38
rs781318458 Chr.19: 18596790-18596790 on Build GRCh38
rs758456131 Chr.19: 18596979-18596979 on Build GRCh38
rs961332766 Chr.19: 18596942-18596942 on Build GRCh38
rs765230520 Chr.19: 18596902-18596902 on Build GRCh38
rs981347676 Chr.19: 18597157-18597157 on Build GRCh38
rs754963858 Chr.19: 18596814-18596814 on Build GRCh38
rs201795161 Chr.19: 18597032-18597032 on Build GRCh38
rs747203538 Chr.19: 18597084-18597084 on Build GRCh38
rs778045876 Chr.19: 18597079-18597079 on Build GRCh38
rs766431820 Chr.19: 18596966-18596966 on Build GRCh38
rs1033536068 Chr.19: 18596869-18596869 on Build GRCh38
rs1052065594 Chr.19: 18597215-18597215 on Build GRCh38
rs987874365 Chr.19: 18596810-18596810 on Build GRCh38
rs779471836 Chr.19: 18596992-18596992 on Build GRCh38
rs761982168 Chr.19: 18596944-18596944 on Build GRCh38
rs920665373 Chr.19: 18597141-18597141 on Build GRCh38
rs771091973 Chr.19: 18597097-18597097 on Build GRCh38
rs753694843 Chr.19: 18596812-18596812 on Build GRCh38
rs768727082 Chr.19: 18597033-18597034 on Build GRCh38
rs764135517 Chr.19: 18597029-18597029 on Build GRCh38
rs763935340 Chr.19: 18596980-18596980 on Build GRCh38
rs1043478703 Chr.19: 18597251-18597251 on Build GRCh38
rs777747919 Chr.19: 18596917-18596917 on Build GRCh38
rs192302108 Chr.19: 18597130-18597130 on Build GRCh38
rs563798848 Chr.19: 18597258-18597258 on Build GRCh38
rs869312035 Chr.19: 18596901-18596902 on Build GRCh38
rs776256829 Chr.19: 18596855-18596855 on Build GRCh38
rs913539916 Chr.19: 18597225-18597225 on Build GRCh38
rs747382667 Chr.19: 18597131-18597131 on Build GRCh38
rs761655766 Chr.19: 18597030-18597030 on Build GRCh38
rs777551327 Chr.19: 18596839-18596839 on Build GRCh38
rs767783644 Chr.19: 18596954-18596954 on Build GRCh38
rs774718772 Chr.19: 18596941-18596941 on Build GRCh38
rs892215986 Chr.19: 18597008-18597008 on Build GRCh38
rs370364717 Chr.19: 18596955-18596955 on Build GRCh38
rs1022095314 Chr.19: 18597025-18597025 on Build GRCh38
rs755419832 Chr.19: 18597066-18597066 on Build GRCh38
rs749775432 Chr.19: 18596998-18596998 on Build GRCh38
rs769519067 Chr.19: 18596831-18596836 on Build GRCh38
rs149015379 Chr.19: 18596925-18596925 on Build GRCh38
rs540101061 Chr.19: 18596812-18596816 on Build GRCh38
rs748716431 Chr.19: 18597075-18597075 on Build GRCh38
rs775654600 Chr.19: 18596865-18596865 on Build GRCh38
rs369142430 Chr.19: 18597004-18597004 on Build GRCh38
rs137853927 Chr.19: 18596895-18596895 on Build GRCh38
rs780915776 Chr.19: 18596985-18596985 on Build GRCh38
rs943536005 Chr.19: 18597254-18597254 on Build GRCh38
rs1051858920 Chr.19: 18597019-18597019 on Build GRCh38
rs752630628 Chr.19: 18596906-18596906 on Build GRCh38

View Full Product Details