Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18742803-18743296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC1
Entrez Gene ID
23373
Gene Name
CREB regulated transcription coactivator 1
Gene Aliases
MECT1, TORC-1, TORC1, WAMTP1
UniGene
Hs.371096
Gene Chromosome Location
Chr.19: 18683615-18782333 on Build GRCh38
Transcripts
NM_001098482.1 NM_015321.2 XM_005259833.3 XM_005259834.1 XM_005259835.3 XM_005259836.3 XM_006722710.3 XM_011527842.2 XM_017026536.1 XM_017026537.1 XM_017026538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712062 c.164C>T p.P55L Chr.19: 18742947-18742947 on Build GRCh38
1712061 c.164C>T p.P55L Chr.19: 18742947-18742947 on Build GRCh38
4835321 c.214G>A p.G72R Chr.19: 18742997-18742997 on Build GRCh38
1391716 c.224C>T p.T75I Chr.19: 18743007-18743007 on Build GRCh38
6294744 c.173G>T p.G58V Chr.19: 18742956-18742956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902154717 Chr.19: 18743166-18743166 on Build GRCh38
rs1030378134 Chr.19: 18743269-18743269 on Build GRCh38
rs200456495 Chr.19: 18742909-18742909 on Build GRCh38
rs754013603 Chr.19: 18743062-18743062 on Build GRCh38
rs769035575 Chr.19: 18742908-18742908 on Build GRCh38
rs934993402 Chr.19: 18743181-18743181 on Build GRCh38
rs1046190245 Chr.19: 18743163-18743163 on Build GRCh38
rs772517937 Chr.19: 18743017-18743017 on Build GRCh38
rs1018051676 Chr.19: 18743214-18743214 on Build GRCh38
rs745554513 Chr.19: 18742885-18742885 on Build GRCh38
rs776924441 Chr.19: 18743038-18743038 on Build GRCh38
rs748737417 Chr.19: 18742998-18742998 on Build GRCh38
rs751500321 Chr.19: 18742973-18742973 on Build GRCh38
rs779827394 Chr.19: 18742996-18742996 on Build GRCh38
rs950715313 Chr.19: 18743275-18743275 on Build GRCh38
rs983684003 Chr.19: 18743106-18743106 on Build GRCh38
rs1020411596 Chr.19: 18743161-18743161 on Build GRCh38
rs929999199 Chr.19: 18743241-18743241 on Build GRCh38
rs757058905 Chr.19: 18742876-18742876 on Build GRCh38
rs902207156 Chr.19: 18742857-18742857 on Build GRCh38
rs776569331 Chr.19: 18742947-18742947 on Build GRCh38
rs371437723 Chr.19: 18743223-18743223 on Build GRCh38
rs757003640 Chr.19: 18742974-18742974 on Build GRCh38
rs942333173 Chr.19: 18743130-18743130 on Build GRCh38
rs552817914 Chr.19: 18743208-18743208 on Build GRCh38
rs369866685 Chr.19: 18742884-18742884 on Build GRCh38
rs991395483 Chr.19: 18743148-18743148 on Build GRCh38
rs569938199 Chr.19: 18742905-18742905 on Build GRCh38
rs779223908 Chr.19: 18743008-18743008 on Build GRCh38
rs779024029 Chr.19: 18743209-18743209 on Build GRCh38
rs371207750 Chr.19: 18743048-18743048 on Build GRCh38
rs777033896 Chr.19: 18743150-18743150 on Build GRCh38
rs1039580581 Chr.19: 18743124-18743124 on Build GRCh38
rs747419096 Chr.19: 18743029-18743029 on Build GRCh38
rs754533421 Chr.19: 18743002-18743002 on Build GRCh38
rs374969669 Chr.19: 18742906-18742906 on Build GRCh38
rs763996458 Chr.19: 18742898-18742899 on Build GRCh38
rs774222165 Chr.19: 18743057-18743057 on Build GRCh38
rs760998829 Chr.19: 18742936-18742936 on Build GRCh38
rs541696446 Chr.19: 18742969-18742969 on Build GRCh38
rs751456753 Chr.19: 18743061-18743061 on Build GRCh38
rs997532835 Chr.19: 18743236-18743236 on Build GRCh38
rs373049904 Chr.19: 18742903-18742903 on Build GRCh38
rs750264099 Chr.19: 18742976-18742976 on Build GRCh38
rs865890656 Chr.19: 18743174-18743174 on Build GRCh38
rs949100700 Chr.19: 18743155-18743155 on Build GRCh38
rs1006689741 Chr.19: 18743102-18743102 on Build GRCh38
rs974097818 Chr.19: 18743224-18743224 on Build GRCh38
rs773690296 Chr.19: 18743019-18743019 on Build GRCh38
rs538413358 Chr.19: 18743113-18743113 on Build GRCh38
rs1002590929 Chr.19: 18743197-18743197 on Build GRCh38
rs117607802 Chr.19: 18743201-18743201 on Build GRCh38
rs560753245 Chr.19: 18742975-18742975 on Build GRCh38
rs1011235576 Chr.19: 18743149-18743149 on Build GRCh38
rs1005286070 Chr.19: 18743276-18743276 on Build GRCh38
rs909465255 Chr.19: 18743120-18743120 on Build GRCh38
rs748463661 Chr.19: 18742912-18742912 on Build GRCh38
rs755728185 Chr.19: 18742984-18742984 on Build GRCh38
rs543156292 Chr.19: 18743170-18743170 on Build GRCh38
rs574094188 Chr.19: 18743129-18743129 on Build GRCh38
rs771044924 Chr.19: 18743037-18743037 on Build GRCh38
rs748698668 Chr.19: 18743015-18743015 on Build GRCh38
rs890922452 Chr.19: 18743047-18743047 on Build GRCh38
rs117770608 Chr.19: 18742927-18742927 on Build GRCh38
rs375809198 Chr.19: 18742869-18742869 on Build GRCh38

View Full Product Details