Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18869087-18869575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS1
Entrez Gene ID
10715
Gene Name
ceramide synthase 1
Gene Aliases
EPM8, LAG1, LASS1, UOG1
UniGene
Hs.412355
Gene Chromosome Location
Chr.19: 18868546-18896727 on Build GRCh38
Transcripts
NM_021267.4 NM_001492.5

Gene Symbol
GDF1
Entrez Gene ID
2657
Gene Name
growth differentiation factor 1
Gene Aliases
DORV, DTGA3, RAI
UniGene
Hs.412355
Gene Chromosome Location
Chr.19: 18868546-18896144 on Build GRCh38
Transcripts
NM_021267.4 NM_001492.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4993600 c.453_454insGCG p.A158_P159insA Chr.19: 18869262-18869263 on Build GRCh38
7312006 c.353C>T p.A118V Chr.19: 18869363-18869363 on Build GRCh38
7312005 c.353C>T p.A118V Chr.19: 18869363-18869363 on Build GRCh38
4993599 c.453_454insGCG p.A158_P159insA Chr.19: 18869262-18869263 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778442978 Chr.19: 18869355-18869355 on Build GRCh38
rs1023239483 Chr.19: 18869446-18869446 on Build GRCh38
rs778324698 Chr.19: 18869416-18869416 on Build GRCh38
rs186263243 Chr.19: 18869489-18869489 on Build GRCh38
rs764806804 Chr.19: 18869402-18869402 on Build GRCh38
rs754190817 Chr.19: 18869248-18869249 on Build GRCh38
rs543008232 Chr.19: 18869527-18869527 on Build GRCh38
rs978050328 Chr.19: 18869193-18869193 on Build GRCh38
rs929436392 Chr.19: 18869292-18869292 on Build GRCh38
rs573559104 Chr.19: 18869117-18869117 on Build GRCh38
rs903852214 Chr.19: 18869420-18869420 on Build GRCh38
rs973565739 Chr.19: 18869252-18869252 on Build GRCh38
rs375438344 Chr.19: 18869532-18869532 on Build GRCh38
rs777054909 Chr.19: 18869286-18869286 on Build GRCh38
rs975747708 Chr.19: 18869491-18869491 on Build GRCh38
rs752155951 Chr.19: 18869419-18869419 on Build GRCh38
rs781649981 Chr.19: 18869426-18869426 on Build GRCh38
rs758755366 Chr.19: 18869246-18869254 on Build GRCh38
rs761762230 Chr.19: 18869334-18869334 on Build GRCh38
rs990276981 Chr.19: 18869484-18869484 on Build GRCh38
rs771276786 Chr.19: 18869285-18869285 on Build GRCh38
rs759669190 Chr.19: 18869290-18869290 on Build GRCh38
rs555453880 Chr.19: 18869261-18869261 on Build GRCh38
rs571387097 Chr.19: 18869245-18869246 on Build GRCh38
rs765449212 Chr.19: 18869304-18869304 on Build GRCh38
rs121434424 Chr.19: 18869231-18869231 on Build GRCh38
rs776966639 Chr.19: 18869372-18869372 on Build GRCh38
rs766202026 Chr.19: 18869387-18869387 on Build GRCh38
rs558253917 Chr.19: 18869376-18869376 on Build GRCh38
rs868061973 Chr.19: 18869295-18869295 on Build GRCh38
rs767483299 Chr.19: 18869385-18869385 on Build GRCh38
rs912179229 Chr.19: 18869523-18869523 on Build GRCh38
rs766003135 Chr.19: 18869349-18869349 on Build GRCh38
rs868026629 Chr.19: 18869120-18869120 on Build GRCh38
rs1022132445 Chr.19: 18869141-18869141 on Build GRCh38
rs973806207 Chr.19: 18869451-18869451 on Build GRCh38
rs775830151 Chr.19: 18869246-18869248 on Build GRCh38
rs4808863 Chr.19: 18869363-18869363 on Build GRCh38
rs1035327337 Chr.19: 18869223-18869223 on Build GRCh38
rs772483004 Chr.19: 18869242-18869242 on Build GRCh38
rs778213666 Chr.19: 18869362-18869362 on Build GRCh38
rs773574790 Chr.19: 18869249-18869249 on Build GRCh38
rs4808864 Chr.19: 18869404-18869404 on Build GRCh38
rs966648207 Chr.19: 18869167-18869167 on Build GRCh38
rs747631423 Chr.19: 18869356-18869356 on Build GRCh38
rs753537451 Chr.19: 18869351-18869351 on Build GRCh38
rs1010776708 Chr.19: 18869131-18869131 on Build GRCh38
rs918311000 Chr.19: 18869453-18869453 on Build GRCh38
rs767371085 Chr.19: 18869340-18869340 on Build GRCh38
rs773079354 Chr.19: 18869386-18869386 on Build GRCh38
rs1047993053 Chr.19: 18869302-18869302 on Build GRCh38
rs757906133 Chr.19: 18869412-18869412 on Build GRCh38
rs578219025 Chr.19: 18869398-18869398 on Build GRCh38
rs746048741 Chr.19: 18869375-18869375 on Build GRCh38
rs1032216804 Chr.19: 18869447-18869447 on Build GRCh38
rs769994620 Chr.19: 18869429-18869429 on Build GRCh38
rs754449715 Chr.19: 18869352-18869352 on Build GRCh38
rs181918871 Chr.19: 18869328-18869328 on Build GRCh38
rs957568704 Chr.19: 18869462-18869462 on Build GRCh38
rs912090524 Chr.19: 18869309-18869309 on Build GRCh38
rs762948218 Chr.19: 18869380-18869380 on Build GRCh38
rs771475316 Chr.19: 18869371-18869371 on Build GRCh38
rs1017324676 Chr.19: 18869488-18869488 on Build GRCh38
rs779370447 Chr.19: 18869221-18869221 on Build GRCh38
rs746250122 Chr.19: 18869427-18869427 on Build GRCh38
rs895247220 Chr.19: 18869305-18869305 on Build GRCh38
rs944730356 Chr.19: 18869312-18869312 on Build GRCh38
rs950914591 Chr.19: 18869485-18869485 on Build GRCh38
rs758878687 Chr.19: 18869359-18869359 on Build GRCh38
rs999030522 Chr.19: 18869417-18869417 on Build GRCh38
rs747345121 Chr.19: 18869279-18869279 on Build GRCh38
rs777905336 Chr.19: 18869343-18869365 on Build GRCh38
rs757870806 Chr.19: 18869422-18869422 on Build GRCh38
rs920749209 Chr.19: 18869259-18869260 on Build GRCh38
rs900625437 Chr.19: 18869336-18869336 on Build GRCh38
rs529560442 Chr.19: 18869509-18869509 on Build GRCh38
rs755967902 Chr.19: 18869344-18869344 on Build GRCh38
rs907488748 Chr.19: 18869415-18869415 on Build GRCh38
rs891345288 Chr.19: 18869540-18869540 on Build GRCh38
rs867249211 Chr.19: 18869505-18869505 on Build GRCh38
rs944947673 Chr.19: 18869548-18869548 on Build GRCh38
rs747099862 Chr.19: 18869392-18869392 on Build GRCh38
rs900553253 Chr.19: 18869129-18869129 on Build GRCh38
rs775584529 Chr.19: 18869377-18869377 on Build GRCh38

View Full Product Details