Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39539040-39539585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID2
Entrez Gene ID
163126
Gene Name
EP300 interacting inhibitor of differentiation 2
Gene Aliases
CRI2, EID-2
UniGene
Hs.18949
Gene Chromosome Location
Chr.19: 39538806-39540198 on Build GRCh38
Transcripts
NM_153232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349584 c.509G>C p.R170T Chr.19: 39539571-39539571 on Build GRCh38
6276245 c.551C>A p.A184D Chr.19: 39539529-39539529 on Build GRCh38
6151125 c.581C>T p.A194V Chr.19: 39539499-39539499 on Build GRCh38
5495219 c.533G>T p.R178L Chr.19: 39539547-39539547 on Build GRCh38
7366015 c.523G>A p.E175K Chr.19: 39539557-39539557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053655497 Chr.19: 39539129-39539129 on Build GRCh38
rs188264868 Chr.19: 39539368-39539368 on Build GRCh38
rs1039421415 Chr.19: 39539305-39539305 on Build GRCh38
rs747822250 Chr.19: 39539437-39539437 on Build GRCh38
rs780862481 Chr.19: 39539432-39539432 on Build GRCh38
rs769521301 Chr.19: 39539450-39539450 on Build GRCh38
rs950528172 Chr.19: 39539252-39539252 on Build GRCh38
rs201697730 Chr.19: 39539333-39539333 on Build GRCh38
rs1002434028 Chr.19: 39539065-39539065 on Build GRCh38
rs1047427766 Chr.19: 39539344-39539344 on Build GRCh38
rs754244593 Chr.19: 39539518-39539518 on Build GRCh38
rs779348803 Chr.19: 39539401-39539401 on Build GRCh38
rs760988363 Chr.19: 39539495-39539495 on Build GRCh38
rs911690759 Chr.19: 39539260-39539260 on Build GRCh38
rs1020949156 Chr.19: 39539197-39539197 on Build GRCh38
rs778300745 Chr.19: 39539372-39539372 on Build GRCh38
rs958310117 Chr.19: 39539182-39539182 on Build GRCh38
rs200861673 Chr.19: 39539362-39539362 on Build GRCh38
rs368356010 Chr.19: 39539468-39539468 on Build GRCh38
rs571051669 Chr.19: 39539136-39539136 on Build GRCh38
rs911964988 Chr.19: 39539257-39539257 on Build GRCh38
rs751652455 Chr.19: 39539327-39539327 on Build GRCh38
rs542845606 Chr.19: 39539203-39539203 on Build GRCh38
rs772326499 Chr.19: 39539455-39539456 on Build GRCh38
rs987273623 Chr.19: 39539556-39539556 on Build GRCh38
rs150653741 Chr.19: 39539188-39539188 on Build GRCh38
rs778852321 Chr.19: 39539336-39539336 on Build GRCh38
rs752911187 Chr.19: 39539485-39539485 on Build GRCh38
rs763059264 Chr.19: 39539174-39539174 on Build GRCh38
rs1002250028 Chr.19: 39539185-39539185 on Build GRCh38
rs779142364 Chr.19: 39539388-39539389 on Build GRCh38
rs1433108 Chr.19: 39539561-39539561 on Build GRCh38
rs774203684 Chr.19: 39539473-39539473 on Build GRCh38
rs1045389 Chr.19: 39539365-39539365 on Build GRCh38
rs566630361 Chr.19: 39539479-39539479 on Build GRCh38
rs766381806 Chr.19: 39539319-39539319 on Build GRCh38
rs531668290 Chr.19: 39539281-39539281 on Build GRCh38
rs1034359852 Chr.19: 39539089-39539089 on Build GRCh38
rs974661309 Chr.19: 39539293-39539293 on Build GRCh38
rs141893929 Chr.19: 39539186-39539186 on Build GRCh38
rs757748164 Chr.19: 39539537-39539537 on Build GRCh38
rs755069098 Chr.19: 39539330-39539330 on Build GRCh38
rs970481684 Chr.19: 39539180-39539180 on Build GRCh38
rs943216046 Chr.19: 39539283-39539283 on Build GRCh38
rs772998243 Chr.19: 39539461-39539461 on Build GRCh38
rs757842008 Chr.19: 39539390-39539390 on Build GRCh38
rs374369388 Chr.19: 39539341-39539341 on Build GRCh38
rs764483293 Chr.19: 39539508-39539508 on Build GRCh38
rs762892046 Chr.19: 39539318-39539318 on Build GRCh38
rs8105143 Chr.19: 39539346-39539346 on Build GRCh38
rs759628844 Chr.19: 39539476-39539476 on Build GRCh38
rs973351909 Chr.19: 39539216-39539216 on Build GRCh38
rs768095342 Chr.19: 39539417-39539417 on Build GRCh38
rs1000468934 Chr.19: 39539093-39539093 on Build GRCh38
rs987325851 Chr.19: 39539254-39539254 on Build GRCh38
rs372306245 Chr.19: 39539470-39539470 on Build GRCh38
rs748484891 Chr.19: 39539402-39539404 on Build GRCh38
rs746509987 Chr.19: 39539409-39539409 on Build GRCh38
rs1014418018 Chr.19: 39539137-39539137 on Build GRCh38
rs762804502 Chr.19: 39539465-39539465 on Build GRCh38
rs1001964458 Chr.19: 39539416-39539416 on Build GRCh38
rs898824120 Chr.19: 39539455-39539455 on Build GRCh38
rs920500290 Chr.19: 39539317-39539317 on Build GRCh38
rs781626999 Chr.19: 39539251-39539251 on Build GRCh38
rs561134017 Chr.19: 39539248-39539248 on Build GRCh38
rs201197879 Chr.19: 39539378-39539378 on Build GRCh38

View Full Product Details