Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39540110-39540623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID2
Entrez Gene ID
163126
Gene Name
EP300 interacting inhibitor of differentiation 2
Gene Aliases
CRI2, EID-2
UniGene
Hs.18949
Gene Chromosome Location
Chr.19: 39538806-39540198 on Build GRCh38
Transcripts
NM_153232.3

SNP Detail

SNP ID Chromosome Location
rs73553822 Chr.19: 39540261-39540261 on Build GRCh38
rs1003180965 Chr.19: 39540525-39540525 on Build GRCh38
rs930344821 Chr.19: 39540141-39540141 on Build GRCh38
rs1009194302 Chr.19: 39540158-39540158 on Build GRCh38
rs1003371789 Chr.19: 39540556-39540556 on Build GRCh38
rs958417616 Chr.19: 39540216-39540216 on Build GRCh38
rs1042562447 Chr.19: 39540147-39540147 on Build GRCh38
rs375647460 Chr.19: 39540207-39540207 on Build GRCh38
rs981389627 Chr.19: 39540206-39540206 on Build GRCh38
rs759233998 Chr.19: 39540294-39540294 on Build GRCh38
rs1002846894 Chr.19: 39540224-39540224 on Build GRCh38
rs931701676 Chr.19: 39540427-39540427 on Build GRCh38
rs887888062 Chr.19: 39540176-39540176 on Build GRCh38
rs372700402 Chr.19: 39540136-39540136 on Build GRCh38
rs951520614 Chr.19: 39540302-39540302 on Build GRCh38
rs1005004309 Chr.19: 39540407-39540407 on Build GRCh38
rs996639427 Chr.19: 39540170-39540170 on Build GRCh38
rs746845563 Chr.19: 39540178-39540178 on Build GRCh38
rs887858549 Chr.19: 39540395-39540395 on Build GRCh38
rs1049112555 Chr.19: 39540394-39540394 on Build GRCh38
rs533206970 Chr.19: 39540539-39540539 on Build GRCh38
rs1014804996 Chr.19: 39540194-39540194 on Build GRCh38
rs944921234 Chr.19: 39540318-39540318 on Build GRCh38
rs1020307854 Chr.19: 39540212-39540212 on Build GRCh38
rs74906438 Chr.19: 39540423-39540423 on Build GRCh38
rs900471376 Chr.19: 39540373-39540373 on Build GRCh38
rs1034285285 Chr.19: 39540540-39540540 on Build GRCh38
rs989757373 Chr.19: 39540557-39540557 on Build GRCh38
rs1034781473 Chr.19: 39540235-39540235 on Build GRCh38
rs762323197 Chr.19: 39540457-39540457 on Build GRCh38
rs375795602 Chr.19: 39540187-39540187 on Build GRCh38
rs1054262530 Chr.19: 39540177-39540177 on Build GRCh38
rs907037038 Chr.19: 39540484-39540484 on Build GRCh38
rs1040645870 Chr.19: 39540323-39540323 on Build GRCh38
rs974735901 Chr.19: 39540288-39540288 on Build GRCh38
rs989829627 Chr.19: 39540256-39540256 on Build GRCh38
rs538610162 Chr.19: 39540296-39540296 on Build GRCh38
rs374466491 Chr.19: 39540587-39540588 on Build GRCh38
rs978230260 Chr.19: 39540287-39540287 on Build GRCh38
rs945870104 Chr.19: 39540497-39540497 on Build GRCh38
rs764734511 Chr.19: 39540162-39540162 on Build GRCh38
rs762351467 Chr.19: 39540142-39540142 on Build GRCh38
rs140020262 Chr.19: 39540530-39540530 on Build GRCh38
rs1027751538 Chr.19: 39540174-39540174 on Build GRCh38
rs995557935 Chr.19: 39540582-39540582 on Build GRCh38
rs554459393 Chr.19: 39540568-39540568 on Build GRCh38
rs565758809 Chr.19: 39540449-39540449 on Build GRCh38
rs1054778385 Chr.19: 39540431-39540431 on Build GRCh38
rs564219986 Chr.19: 39540217-39540217 on Build GRCh38
rs775965286 Chr.19: 39540180-39540180 on Build GRCh38
rs914311179 Chr.19: 39540257-39540257 on Build GRCh38
rs931954740 Chr.19: 39540377-39540377 on Build GRCh38
rs373484315 Chr.19: 39540400-39540400 on Build GRCh38
rs531901393 Chr.19: 39540168-39540168 on Build GRCh38
rs902747435 Chr.19: 39540151-39540151 on Build GRCh38
rs958901522 Chr.19: 39540555-39540555 on Build GRCh38
rs971711620 Chr.19: 39540202-39540202 on Build GRCh38
rs1015033823 Chr.19: 39540271-39540271 on Build GRCh38
rs1041546155 Chr.19: 39540506-39540506 on Build GRCh38
rs894458301 Chr.19: 39540571-39540571 on Build GRCh38
rs535272133 Chr.19: 39540399-39540399 on Build GRCh38
rs893079292 Chr.19: 39540462-39540462 on Build GRCh38
rs971483277 Chr.19: 39540218-39540218 on Build GRCh38

View Full Product Details