Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39870138-39870635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGBP
Entrez Gene ID
8857
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
FC(GAMMA)BP
UniGene
Hs.111732
Gene Chromosome Location
Chr.19: 39863323-39934626 on Build GRCh38
Transcripts

SNP Detail

SNP ID Chromosome Location
rs920374479 Chr.19: 39870307-39870307 on Build GRCh38
rs769584991 Chr.19: 39870317-39870317 on Build GRCh38
rs778971026 Chr.19: 39870397-39870397 on Build GRCh38
rs754164424 Chr.19: 39870339-39870339 on Build GRCh38
rs145067279 Chr.19: 39870333-39870333 on Build GRCh38
rs200378087 Chr.19: 39870415-39870415 on Build GRCh38
rs768745210 Chr.19: 39870404-39870404 on Build GRCh38
rs373880753 Chr.19: 39870461-39870461 on Build GRCh38
rs139526217 Chr.19: 39870296-39870296 on Build GRCh38
rs747851180 Chr.19: 39870305-39870305 on Build GRCh38
rs767754332 Chr.19: 39870330-39870330 on Build GRCh38
rs764451016 Chr.19: 39870335-39870335 on Build GRCh38
rs766421252 Chr.19: 39870177-39870177 on Build GRCh38
rs757705958 Chr.19: 39870290-39870290 on Build GRCh38
rs3760929 Chr.19: 39870205-39870205 on Build GRCh38
rs936588133 Chr.19: 39870199-39870199 on Build GRCh38
rs887323887 Chr.19: 39870321-39870321 on Build GRCh38
rs112158994 Chr.19: 39870188-39870188 on Build GRCh38
rs957673469 Chr.19: 39870369-39870369 on Build GRCh38
rs540284772 Chr.19: 39870402-39870402 on Build GRCh38
rs776973740 Chr.19: 39870414-39870414 on Build GRCh38
rs754329544 Chr.19: 39870289-39870289 on Build GRCh38
rs753007914 Chr.19: 39870272-39870272 on Build GRCh38
rs964502952 Chr.19: 39870525-39870525 on Build GRCh38
rs766950657 Chr.19: 39870456-39870456 on Build GRCh38
rs983039941 Chr.19: 39870557-39870557 on Build GRCh38
rs34811614 Chr.19: 39870608-39870609 on Build GRCh38
rs749544284 Chr.19: 39870325-39870329 on Build GRCh38
rs763485062 Chr.19: 39870448-39870448 on Build GRCh38
rs754466025 Chr.19: 39870351-39870351 on Build GRCh38
rs1025147946 Chr.19: 39870182-39870182 on Build GRCh38
rs575240993 Chr.19: 39870320-39870320 on Build GRCh38
rs746602229 Chr.19: 39870226-39870226 on Build GRCh38
rs780385460 Chr.19: 39870274-39870275 on Build GRCh38
rs140648438 Chr.19: 39870391-39870391 on Build GRCh38
rs765686847 Chr.19: 39870425-39870425 on Build GRCh38
rs3760931 Chr.19: 39870472-39870472 on Build GRCh38
rs368974681 Chr.19: 39870235-39870235 on Build GRCh38
rs374766909 Chr.19: 39870341-39870341 on Build GRCh38
rs988025387 Chr.19: 39870332-39870332 on Build GRCh38
rs1026537999 Chr.19: 39870299-39870299 on Build GRCh38
rs774614015 Chr.19: 39870442-39870443 on Build GRCh38
rs552303848 Chr.19: 39870588-39870588 on Build GRCh38
rs751758861 Chr.19: 39870178-39870178 on Build GRCh38
rs756605942 Chr.19: 39870202-39870202 on Build GRCh38
rs188932726 Chr.19: 39870594-39870594 on Build GRCh38
rs370106966 Chr.19: 39870398-39870398 on Build GRCh38
rs148526142 Chr.19: 39870510-39870510 on Build GRCh38
rs906571655 Chr.19: 39870186-39870186 on Build GRCh38
rs774301932 Chr.19: 39870266-39870266 on Build GRCh38
rs750884516 Chr.19: 39870427-39870427 on Build GRCh38
rs374569267 Chr.19: 39870227-39870227 on Build GRCh38
rs200125442 Chr.19: 39870297-39870297 on Build GRCh38
rs749023392 Chr.19: 39870383-39870383 on Build GRCh38
rs750398364 Chr.19: 39870167-39870167 on Build GRCh38
rs781636509 Chr.19: 39870189-39870189 on Build GRCh38
rs776318997 Chr.19: 39870229-39870229 on Build GRCh38
rs958009997 Chr.19: 39870224-39870224 on Build GRCh38
rs767840667 Chr.19: 39870271-39870271 on Build GRCh38
rs768835655 Chr.19: 39870420-39870421 on Build GRCh38
rs974572432 Chr.19: 39870536-39870536 on Build GRCh38
rs745822010 Chr.19: 39870401-39870401 on Build GRCh38
rs184463001 Chr.19: 39870334-39870334 on Build GRCh38
rs747761387 Chr.19: 39870366-39870366 on Build GRCh38
rs779459680 Chr.19: 39870217-39870217 on Build GRCh38
rs867136565 Chr.19: 39870340-39870340 on Build GRCh38
rs568268028 Chr.19: 39870230-39870230 on Build GRCh38
rs920931449 Chr.19: 39870364-39870364 on Build GRCh38
rs759774406 Chr.19: 39870269-39870269 on Build GRCh38
rs773181877 Chr.19: 39870240-39870240 on Build GRCh38
rs762823567 Chr.19: 39870260-39870260 on Build GRCh38
rs892314162 Chr.19: 39870209-39870210 on Build GRCh38
rs768929860 Chr.19: 39870490-39870490 on Build GRCh38
rs367934901 Chr.19: 39870373-39870373 on Build GRCh38
rs779557013 Chr.19: 39870293-39870293 on Build GRCh38
rs544566860 Chr.19: 39870505-39870505 on Build GRCh38
rs766444672 Chr.19: 39870263-39870263 on Build GRCh38
rs780707707 Chr.19: 39870352-39870352 on Build GRCh38
rs774405155 Chr.19: 39870325-39870325 on Build GRCh38
rs73930662 Chr.19: 39870372-39870372 on Build GRCh38
rs535080628 Chr.19: 39870166-39870166 on Build GRCh38
rs775132189 Chr.19: 39870256-39870257 on Build GRCh38
rs562621506 Chr.19: 39870506-39870506 on Build GRCh38
rs771560291 Chr.19: 39870215-39870215 on Build GRCh38
rs201020566 Chr.19: 39870329-39870329 on Build GRCh38
rs763061961 Chr.19: 39870176-39870176 on Build GRCh38
rs966666679 Chr.19: 39870256-39870256 on Build GRCh38
rs964867591 Chr.19: 39870578-39870578 on Build GRCh38
rs749807427 Chr.19: 39870211-39870211 on Build GRCh38
rs199570976 Chr.19: 39870294-39870294 on Build GRCh38
rs949786377 Chr.19: 39870603-39870603 on Build GRCh38
rs879130742 Chr.19: 39870499-39870499 on Build GRCh38
rs371263426 Chr.19: 39870319-39870319 on Build GRCh38
rs1053686 Chr.19: 39870288-39870288 on Build GRCh38
rs755284049 Chr.19: 39870180-39870180 on Build GRCh38
rs753126452 Chr.19: 39870196-39870196 on Build GRCh38
rs920315468 Chr.19: 39870551-39870551 on Build GRCh38
rs769620324 Chr.19: 39870238-39870238 on Build GRCh38
rs750965934 Chr.19: 39870342-39870342 on Build GRCh38
rs951804539 Chr.19: 39870552-39870552 on Build GRCh38

View Full Product Details