Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40223184-40223682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTD2
Entrez Gene ID
79935
Gene Name
cyclin N-terminal domain containing 2
Gene Aliases
CCNP
UniGene
Hs.631603
Gene Chromosome Location
Chr.19: 40222208-40227111 on Build GRCh38
Transcripts
NM_024877.3 XM_006723395.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078153 c.620G>A p.G207D Chr.19: 40223440-40223440 on Build GRCh38
1481072 c.681A>T p.L227F Chr.19: 40223295-40223295 on Build GRCh38
6841028 c.746G>A p.R249H Chr.19: 40223230-40223230 on Build GRCh38
4559100 c.791G>A p.G264E Chr.19: 40223185-40223185 on Build GRCh38
3225250 c.610C>A p.H204N Chr.19: 40223450-40223450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529720256 Chr.19: 40223305-40223305 on Build GRCh38
rs1057484353 Chr.19: 40223572-40223579 on Build GRCh38
rs763112058 Chr.19: 40223236-40223236 on Build GRCh38
rs763971811 Chr.19: 40223612-40223612 on Build GRCh38
rs758612858 Chr.19: 40223634-40223634 on Build GRCh38
rs776604616 Chr.19: 40223563-40223564 on Build GRCh38
rs909747680 Chr.19: 40223390-40223390 on Build GRCh38
rs939034770 Chr.19: 40223202-40223202 on Build GRCh38
rs758130327 Chr.19: 40223359-40223359 on Build GRCh38
rs547688635 Chr.19: 40223300-40223300 on Build GRCh38
rs374223999 Chr.19: 40223629-40223629 on Build GRCh38
rs769874245 Chr.19: 40223397-40223397 on Build GRCh38
rs759266233 Chr.19: 40223527-40223527 on Build GRCh38
rs751217378 Chr.19: 40223523-40223523 on Build GRCh38
rs923166530 Chr.19: 40223257-40223257 on Build GRCh38
rs746111929 Chr.19: 40223490-40223490 on Build GRCh38
rs774696618 Chr.19: 40223410-40223410 on Build GRCh38
rs770977936 Chr.19: 40223431-40223431 on Build GRCh38
rs764739980 Chr.19: 40223429-40223429 on Build GRCh38
rs745829109 Chr.19: 40223583-40223583 on Build GRCh38
rs769623782 Chr.19: 40223625-40223626 on Build GRCh38
rs746456585 Chr.19: 40223395-40223401 on Build GRCh38
rs1013081921 Chr.19: 40223231-40223231 on Build GRCh38
rs766174322 Chr.19: 40223450-40223450 on Build GRCh38
rs766660484 Chr.19: 40223240-40223240 on Build GRCh38
rs534784104 Chr.19: 40223564-40223564 on Build GRCh38
rs1013130085 Chr.19: 40223348-40223348 on Build GRCh38
rs570666778 Chr.19: 40223487-40223487 on Build GRCh38
rs998602489 Chr.19: 40223317-40223317 on Build GRCh38
rs769180432 Chr.19: 40223567-40223567 on Build GRCh38
rs775723942 Chr.19: 40223498-40223498 on Build GRCh38
rs543131279 Chr.19: 40223651-40223651 on Build GRCh38
rs780876578 Chr.19: 40223465-40223465 on Build GRCh38
rs761189619 Chr.19: 40223503-40223503 on Build GRCh38
rs373547599 Chr.19: 40223423-40223423 on Build GRCh38
rs547916264 Chr.19: 40223362-40223362 on Build GRCh38
rs756840721 Chr.19: 40223337-40223337 on Build GRCh38
rs1000264091 Chr.19: 40223256-40223256 on Build GRCh38
rs45582332 Chr.19: 40223346-40223346 on Build GRCh38
rs532696748 Chr.19: 40223247-40223247 on Build GRCh38
rs142135057 Chr.19: 40223624-40223624 on Build GRCh38
rs777224614 Chr.19: 40223568-40223568 on Build GRCh38
rs768083473 Chr.19: 40223319-40223319 on Build GRCh38
rs536695271 Chr.19: 40223376-40223376 on Build GRCh38
rs764573952 Chr.19: 40223506-40223506 on Build GRCh38
rs112906736 Chr.19: 40223270-40223270 on Build GRCh38
rs762437458 Chr.19: 40223590-40223590 on Build GRCh38
rs1050530888 Chr.19: 40223264-40223264 on Build GRCh38
rs767140282 Chr.19: 40223598-40223598 on Build GRCh38
rs773166939 Chr.19: 40223400-40223400 on Build GRCh38
rs773431582 Chr.19: 40223218-40223218 on Build GRCh38
rs746865496 Chr.19: 40223369-40223369 on Build GRCh38
rs569301505 Chr.19: 40223367-40223367 on Build GRCh38
rs780186429 Chr.19: 40223643-40223643 on Build GRCh38
rs377464659 Chr.19: 40223619-40223619 on Build GRCh38
rs762598334 Chr.19: 40223521-40223521 on Build GRCh38
rs752491330 Chr.19: 40223536-40223536 on Build GRCh38
rs961849454 Chr.19: 40223228-40223228 on Build GRCh38
rs991912949 Chr.19: 40223660-40223660 on Build GRCh38
rs949005505 Chr.19: 40223479-40223479 on Build GRCh38
rs758118013 Chr.19: 40223436-40223436 on Build GRCh38
rs781239236 Chr.19: 40223387-40223387 on Build GRCh38
rs565440976 Chr.19: 40223203-40223203 on Build GRCh38
rs754770297 Chr.19: 40223455-40223455 on Build GRCh38
rs933222223 Chr.19: 40223258-40223258 on Build GRCh38
rs960775938 Chr.19: 40223642-40223642 on Build GRCh38
rs910177361 Chr.19: 40223290-40223290 on Build GRCh38
rs766006695 Chr.19: 40223522-40223522 on Build GRCh38
rs749467178 Chr.19: 40223405-40223405 on Build GRCh38
rs748068553 Chr.19: 40223424-40223424 on Build GRCh38
rs779000036 Chr.19: 40223563-40223563 on Build GRCh38
rs761118637 Chr.19: 40223428-40223428 on Build GRCh38
rs978538911 Chr.19: 40223544-40223544 on Build GRCh38
rs753260153 Chr.19: 40223320-40223320 on Build GRCh38
rs910252078 Chr.19: 40223534-40223534 on Build GRCh38
rs777613623 Chr.19: 40223543-40223543 on Build GRCh38
rs1056102340 Chr.19: 40223220-40223220 on Build GRCh38
rs756076592 Chr.19: 40223542-40223542 on Build GRCh38
rs565409513 Chr.19: 40223303-40223303 on Build GRCh38
rs771203685 Chr.19: 40223409-40223409 on Build GRCh38
rs36020141 Chr.19: 40223574-40223575 on Build GRCh38
rs947157171 Chr.19: 40223294-40223294 on Build GRCh38
rs764675981 Chr.19: 40223350-40223350 on Build GRCh38
rs375364453 Chr.19: 40223558-40223558 on Build GRCh38
rs776589524 Chr.19: 40223596-40223596 on Build GRCh38
rs904690579 Chr.19: 40223248-40223248 on Build GRCh38
rs778971247 Chr.19: 40223356-40223356 on Build GRCh38
rs770498200 Chr.19: 40223592-40223592 on Build GRCh38
rs748054328 Chr.19: 40223396-40223396 on Build GRCh38
rs760436682 Chr.19: 40223609-40223609 on Build GRCh38
rs752508196 Chr.19: 40223608-40223608 on Build GRCh38
rs770600368 Chr.19: 40223468-40223468 on Build GRCh38
rs747371646 Chr.19: 40223565-40223565 on Build GRCh38
rs896062123 Chr.19: 40223224-40223224 on Build GRCh38
rs553228568 Chr.19: 40223582-40223582 on Build GRCh38
rs772745338 Chr.19: 40223517-40223517 on Build GRCh38
rs759938170 Chr.19: 40223412-40223412 on Build GRCh38
rs896075305 Chr.19: 40223322-40223322 on Build GRCh38
rs369761473 Chr.19: 40223559-40223559 on Build GRCh38
rs1005495679 Chr.19: 40223621-40223621 on Build GRCh38
rs777584297 Chr.19: 40223389-40223390 on Build GRCh38
rs774010708 Chr.19: 40223593-40223593 on Build GRCh38
rs373486581 Chr.19: 40223618-40223618 on Build GRCh38
rs369747903 Chr.19: 40223321-40223321 on Build GRCh38
rs767283083 Chr.19: 40223531-40223531 on Build GRCh38
rs748047515 Chr.19: 40223468-40223468 on Build GRCh38
rs756015975 Chr.19: 40223475-40223475 on Build GRCh38

View Full Product Details