Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40447698-40448199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLVRB
Entrez Gene ID
645
Gene Name
biliverdin reductase B
Gene Aliases
BVRB, FLR, HEL-S-10, SDR43U1
UniGene
Hs.515785
Gene Chromosome Location
Chr.19: 40447784-40465818 on Build GRCh38
Transcripts
NM_000713.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5981535 c.592A>G p.S198G Chr.19: 40447918-40447918 on Build GRCh38
6611815 c.509G>A p.R170Q Chr.19: 40448001-40448001 on Build GRCh38
189359 c.582C>T p.Y194Y Chr.19: 40447928-40447928 on Build GRCh38
4665796 c.525C>A p.V175V Chr.19: 40447985-40447985 on Build GRCh38
4078224 c.580T>C p.Y194H Chr.19: 40447930-40447930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763368632 Chr.19: 40447739-40447739 on Build GRCh38
rs927878328 Chr.19: 40447755-40447755 on Build GRCh38
rs937889845 Chr.19: 40447768-40447768 on Build GRCh38
rs139022515 Chr.19: 40448063-40448063 on Build GRCh38
rs200609801 Chr.19: 40447895-40447895 on Build GRCh38
rs779931948 Chr.19: 40447950-40447950 on Build GRCh38
rs747103556 Chr.19: 40447951-40447951 on Build GRCh38
rs113249041 Chr.19: 40447806-40447806 on Build GRCh38
rs368720101 Chr.19: 40447984-40447984 on Build GRCh38
rs773785470 Chr.19: 40448001-40448001 on Build GRCh38
rs550405658 Chr.19: 40448084-40448084 on Build GRCh38
rs761616455 Chr.19: 40448024-40448024 on Build GRCh38
rs773558164 Chr.19: 40447909-40447909 on Build GRCh38
rs755075525 Chr.19: 40448043-40448043 on Build GRCh38
rs920410956 Chr.19: 40447819-40447819 on Build GRCh38
rs937319298 Chr.19: 40448142-40448142 on Build GRCh38
rs774625796 Chr.19: 40447917-40447917 on Build GRCh38
rs765253159 Chr.19: 40447933-40447933 on Build GRCh38
rs141407327 Chr.19: 40448012-40448012 on Build GRCh38
rs748561718 Chr.19: 40447986-40447986 on Build GRCh38
rs748506660 Chr.19: 40447902-40447902 on Build GRCh38
rs12459890 Chr.19: 40448155-40448155 on Build GRCh38
rs369232148 Chr.19: 40448027-40448027 on Build GRCh38
rs549377727 Chr.19: 40448002-40448002 on Build GRCh38
rs757244672 Chr.19: 40447849-40447849 on Build GRCh38
rs775216213 Chr.19: 40447924-40447924 on Build GRCh38
rs199713428 Chr.19: 40448039-40448039 on Build GRCh38
rs770238288 Chr.19: 40447988-40447988 on Build GRCh38
rs753626041 Chr.19: 40447838-40447838 on Build GRCh38
rs111661396 Chr.19: 40447807-40447807 on Build GRCh38
rs200541223 Chr.19: 40447925-40447925 on Build GRCh38
rs762866114 Chr.19: 40448030-40448030 on Build GRCh38
rs1012157248 Chr.19: 40447774-40447774 on Build GRCh38
rs372232630 Chr.19: 40447931-40447931 on Build GRCh38
rs376563591 Chr.19: 40447927-40447927 on Build GRCh38
rs780343690 Chr.19: 40447882-40447882 on Build GRCh38
rs746002639 Chr.19: 40447876-40447876 on Build GRCh38
rs760156948 Chr.19: 40448013-40448013 on Build GRCh38
rs756826463 Chr.19: 40448062-40448063 on Build GRCh38
rs183585098 Chr.19: 40448161-40448161 on Build GRCh38
rs1061340 Chr.19: 40447858-40447858 on Build GRCh38
rs752912171 Chr.19: 40448061-40448061 on Build GRCh38
rs981447602 Chr.19: 40447742-40447742 on Build GRCh38
rs780964540 Chr.19: 40448072-40448072 on Build GRCh38
rs930488568 Chr.19: 40448102-40448102 on Build GRCh38
rs985015381 Chr.19: 40448006-40448006 on Build GRCh38
rs778801788 Chr.19: 40447866-40447866 on Build GRCh38
rs375620073 Chr.19: 40447867-40447867 on Build GRCh38
rs930501285 Chr.19: 40447827-40447827 on Build GRCh38
rs777197975 Chr.19: 40447901-40447901 on Build GRCh38
rs764519831 Chr.19: 40447916-40447916 on Build GRCh38
rs374753276 Chr.19: 40448025-40448025 on Build GRCh38
rs768339077 Chr.19: 40448019-40448019 on Build GRCh38
rs527721383 Chr.19: 40447985-40447985 on Build GRCh38
rs376664931 Chr.19: 40447937-40447937 on Build GRCh38
rs766322422 Chr.19: 40448038-40448038 on Build GRCh38
rs867422949 Chr.19: 40447989-40447989 on Build GRCh38
rs944397907 Chr.19: 40448153-40448153 on Build GRCh38
rs778070390 Chr.19: 40448070-40448070 on Build GRCh38
rs947524932 Chr.19: 40448159-40448159 on Build GRCh38
rs1048053975 Chr.19: 40448111-40448111 on Build GRCh38
rs10281 Chr.19: 40447864-40447864 on Build GRCh38
rs920532200 Chr.19: 40448091-40448091 on Build GRCh38
rs560626645 Chr.19: 40447936-40447936 on Build GRCh38
rs781593642 Chr.19: 40448055-40448055 on Build GRCh38
rs928545555 Chr.19: 40448107-40448107 on Build GRCh38
rs111302674 Chr.19: 40448130-40448130 on Build GRCh38
rs45460395 Chr.19: 40447807-40447807 on Build GRCh38
rs770469496 Chr.19: 40447905-40447905 on Build GRCh38
rs900173763 Chr.19: 40447928-40447928 on Build GRCh38
rs1041913808 Chr.19: 40447787-40447787 on Build GRCh38
rs1055838995 Chr.19: 40448150-40448150 on Build GRCh38
rs191818368 Chr.19: 40447833-40447833 on Build GRCh38

View Full Product Details