Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40458065-40458557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLVRB
Entrez Gene ID
645
Gene Name
biliverdin reductase B
Gene Aliases
BVRB, FLR, HEL-S-10, SDR43U1
UniGene
Hs.515785
Gene Chromosome Location
Chr.19: 40447784-40465818 on Build GRCh38
Transcripts
NM_000713.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4878593 c.271G>A p.A91T Chr.19: 40458218-40458218 on Build GRCh38
6423144 c.319G>T p.V107L Chr.19: 40458170-40458170 on Build GRCh38
4665797 c.147C>T p.H49H Chr.19: 40458478-40458478 on Build GRCh38
6420929 c.214A>T p.I72F Chr.19: 40458411-40458411 on Build GRCh38
4650486 c.233G>A p.R78H Chr.19: 40458392-40458392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050862630 Chr.19: 40458170-40458170 on Build GRCh38
rs777409601 Chr.19: 40458133-40458133 on Build GRCh38
rs949506555 Chr.19: 40458485-40458485 on Build GRCh38
rs369378608 Chr.19: 40458468-40458468 on Build GRCh38
rs776057942 Chr.19: 40458477-40458477 on Build GRCh38
rs763328417 Chr.19: 40458208-40458208 on Build GRCh38
rs751269661 Chr.19: 40458277-40458278 on Build GRCh38
rs141043815 Chr.19: 40458272-40458272 on Build GRCh38
rs988242581 Chr.19: 40458483-40458483 on Build GRCh38
rs754185247 Chr.19: 40458238-40458240 on Build GRCh38
rs762159909 Chr.19: 40458095-40458095 on Build GRCh38
rs987482722 Chr.19: 40458143-40458143 on Build GRCh38
rs750357961 Chr.19: 40458290-40458298 on Build GRCh38
rs750041694 Chr.19: 40458236-40458236 on Build GRCh38
rs748735268 Chr.19: 40458139-40458139 on Build GRCh38
rs570592751 Chr.19: 40458313-40458313 on Build GRCh38
rs369621328 Chr.19: 40458292-40458292 on Build GRCh38
rs370052045 Chr.19: 40458284-40458284 on Build GRCh38
rs746900518 Chr.19: 40458182-40458182 on Build GRCh38
rs763616711 Chr.19: 40458249-40458253 on Build GRCh38
rs376487651 Chr.19: 40458283-40458283 on Build GRCh38
rs755863671 Chr.19: 40458518-40458518 on Build GRCh38
rs1018126723 Chr.19: 40458325-40458325 on Build GRCh38
rs142505402 Chr.19: 40458237-40458237 on Build GRCh38
rs1038324392 Chr.19: 40458533-40458533 on Build GRCh38
rs746764496 Chr.19: 40458249-40458249 on Build GRCh38
rs147457356 Chr.19: 40458521-40458521 on Build GRCh38
rs374308013 Chr.19: 40458194-40458194 on Build GRCh38
rs773382989 Chr.19: 40458206-40458206 on Build GRCh38
rs758730489 Chr.19: 40458125-40458125 on Build GRCh38
rs757366650 Chr.19: 40458525-40458525 on Build GRCh38
rs149698066 Chr.19: 40458157-40458157 on Build GRCh38
rs546557763 Chr.19: 40458329-40458329 on Build GRCh38
rs200026690 Chr.19: 40458215-40458215 on Build GRCh38
rs768683696 Chr.19: 40458251-40458251 on Build GRCh38
rs201096602 Chr.19: 40458218-40458218 on Build GRCh38
rs781212524 Chr.19: 40458366-40458366 on Build GRCh38
rs202228301 Chr.19: 40458181-40458181 on Build GRCh38
rs375352127 Chr.19: 40458156-40458156 on Build GRCh38
rs779212747 Chr.19: 40458457-40458457 on Build GRCh38
rs777658262 Chr.19: 40458438-40458438 on Build GRCh38
rs138251638 Chr.19: 40458530-40458530 on Build GRCh38
rs754797657 Chr.19: 40458365-40458366 on Build GRCh38
rs778971943 Chr.19: 40458366-40458366 on Build GRCh38
rs561982669 Chr.19: 40458274-40458274 on Build GRCh38
rs765428187 Chr.19: 40458098-40458098 on Build GRCh38
rs761772907 Chr.19: 40458191-40458191 on Build GRCh38
rs367905273 Chr.19: 40458275-40458275 on Build GRCh38
rs747405072 Chr.19: 40458532-40458532 on Build GRCh38
rs945697656 Chr.19: 40458480-40458480 on Build GRCh38
rs768101034 Chr.19: 40458287-40458287 on Build GRCh38
rs1031072264 Chr.19: 40458312-40458312 on Build GRCh38
rs376568747 Chr.19: 40458377-40458377 on Build GRCh38
rs867193418 Chr.19: 40458362-40458362 on Build GRCh38
rs772030200 Chr.19: 40458485-40458485 on Build GRCh38
rs749951740 Chr.19: 40458335-40458335 on Build GRCh38
rs535726000 Chr.19: 40458342-40458342 on Build GRCh38
rs865900670 Chr.19: 40458185-40458185 on Build GRCh38
rs553282639 Chr.19: 40458153-40458153 on Build GRCh38
rs138694963 Chr.19: 40458432-40458432 on Build GRCh38
rs766901756 Chr.19: 40458128-40458128 on Build GRCh38
rs756702541 Chr.19: 40458278-40458280 on Build GRCh38
rs369364430 Chr.19: 40458492-40458492 on Build GRCh38
rs372718660 Chr.19: 40458288-40458288 on Build GRCh38
rs768471398 Chr.19: 40458186-40458186 on Build GRCh38
rs780389636 Chr.19: 40458531-40458531 on Build GRCh38
rs903862888 Chr.19: 40458298-40458298 on Build GRCh38
rs148832924 Chr.19: 40458224-40458224 on Build GRCh38
rs776827642 Chr.19: 40458187-40458187 on Build GRCh38
rs557627615 Chr.19: 40458367-40458367 on Build GRCh38
rs867835861 Chr.19: 40458408-40458408 on Build GRCh38
rs764681599 Chr.19: 40458402-40458402 on Build GRCh38
rs1061197 Chr.19: 40458405-40458405 on Build GRCh38
rs146531246 Chr.19: 40458380-40458380 on Build GRCh38
rs761424530 Chr.19: 40458290-40458290 on Build GRCh38
rs766082597 Chr.19: 40458409-40458409 on Build GRCh38
rs753721054 Chr.19: 40458522-40458522 on Build GRCh38
rs149363787 Chr.19: 40458497-40458497 on Build GRCh38
rs45487299 Chr.19: 40458433-40458433 on Build GRCh38
rs912858548 Chr.19: 40458296-40458296 on Build GRCh38
rs766670702 Chr.19: 40458211-40458211 on Build GRCh38
rs865805057 Chr.19: 40458392-40458392 on Build GRCh38
rs771947418 Chr.19: 40458159-40458159 on Build GRCh38
rs1008032567 Chr.19: 40458319-40458319 on Build GRCh38
rs377461982 Chr.19: 40458152-40458152 on Build GRCh38
rs773405252 Chr.19: 40458514-40458514 on Build GRCh38
rs770038459 Chr.19: 40458192-40458192 on Build GRCh38
rs371470529 Chr.19: 40458276-40458276 on Build GRCh38
rs1050831693 Chr.19: 40458322-40458322 on Build GRCh38
rs748449923 Chr.19: 40458457-40458457 on Build GRCh38
rs772894298 Chr.19: 40458403-40458403 on Build GRCh38
rs957128371 Chr.19: 40458481-40458481 on Build GRCh38
rs999969106 Chr.19: 40458307-40458307 on Build GRCh38
rs746270927 Chr.19: 40458376-40458376 on Build GRCh38
rs747352017 Chr.19: 40458478-40458478 on Build GRCh38
rs779547545 Chr.19: 40458336-40458336 on Build GRCh38
rs552288177 Chr.19: 40458302-40458302 on Build GRCh38
rs767148727 Chr.19: 40458420-40458420 on Build GRCh38
rs534721669 Chr.19: 40458318-40458318 on Build GRCh38
rs911512920 Chr.19: 40458307-40458312 on Build GRCh38
rs762544157 Chr.19: 40458489-40458489 on Build GRCh38
rs759988479 Chr.19: 40458248-40458251 on Build GRCh38
rs769324337 Chr.19: 40458484-40458484 on Build GRCh38
rs757236173 Chr.19: 40458448-40458448 on Build GRCh38
rs199771756 Chr.19: 40458368-40458368 on Build GRCh38
rs45558132 Chr.19: 40458371-40458371 on Build GRCh38
rs754685644 Chr.19: 40458355-40458355 on Build GRCh38
rs771047106 Chr.19: 40458280-40458280 on Build GRCh38
rs200439555 Chr.19: 40458338-40458338 on Build GRCh38
rs144849024 Chr.19: 40458418-40458418 on Build GRCh38
rs779979935 Chr.19: 40458363-40458364 on Build GRCh38
rs150990121 Chr.19: 40458270-40458270 on Build GRCh38
rs890915133 Chr.19: 40458328-40458328 on Build GRCh38
rs45604234 Chr.19: 40458365-40458365 on Build GRCh38
rs764777291 Chr.19: 40458294-40458294 on Build GRCh38
rs755981881 Chr.19: 40458293-40458294 on Build GRCh38
rs1043786163 Chr.19: 40458196-40458196 on Build GRCh38
rs778733922 Chr.19: 40458225-40458225 on Build GRCh38
rs898374223 Chr.19: 40458219-40458219 on Build GRCh38
rs1675404 Chr.19: 40458417-40458417 on Build GRCh38
rs563411888 Chr.19: 40458238-40458238 on Build GRCh38
rs752054595 Chr.19: 40458130-40458130 on Build GRCh38
rs145500296 Chr.19: 40458214-40458214 on Build GRCh38
rs373217922 Chr.19: 40458131-40458131 on Build GRCh38
rs557914261 Chr.19: 40458393-40458393 on Build GRCh38
rs1045094526 Chr.19: 40458339-40458339 on Build GRCh38
rs11547746 Chr.19: 40458488-40458488 on Build GRCh38
rs750831377 Chr.19: 40458114-40458114 on Build GRCh38
rs774056404 Chr.19: 40458493-40458493 on Build GRCh38
rs150228804 Chr.19: 40458447-40458447 on Build GRCh38
rs746014592 Chr.19: 40458458-40458458 on Build GRCh38
rs752531257 Chr.19: 40458506-40458506 on Build GRCh38
rs201539982 Chr.19: 40458289-40458289 on Build GRCh38
rs750507070 Chr.19: 40458529-40458529 on Build GRCh38

View Full Product Details