Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41016504-41017000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2B6
Entrez Gene ID
1555
Gene Name
cytochrome P450 family 2 subfamily B member 6
Gene Aliases
CPB6, CYP2B, CYP2B7, CYP2B7P, CYPIIB6, EFVM, IIB1, P450
UniGene
Hs.1360
Gene Chromosome Location
Chr.19: 40991200-41018402 on Build GRCh38
Transcripts
NM_000767.4 XM_005258569.4 XM_006723050.3 XM_011526547.2 XM_011526548.2 XM_011526549.2 XM_011526550.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
383237 c.1460G>T p.R487L Chr.19: 41016811-41016811 on Build GRCh38
4826300 c.1371C>T p.F457F Chr.19: 41016722-41016722 on Build GRCh38
6639174 c.1301G>A p.R434Q Chr.19: 41016652-41016652 on Build GRCh38
5855859 c.1295-1G>T p.? Chr.19: 41016645-41016645 on Build GRCh38
4675934 c.1323C>T p.I441I Chr.19: 41016674-41016674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761555802 Chr.19: 41016611-41016611 on Build GRCh38
rs370338813 Chr.19: 41016800-41016800 on Build GRCh38
rs745459109 Chr.19: 41016838-41016838 on Build GRCh38
rs757421947 Chr.19: 41016700-41016700 on Build GRCh38
rs375367839 Chr.19: 41016734-41016734 on Build GRCh38
rs765620489 Chr.19: 41016706-41016706 on Build GRCh38
rs559298344 Chr.19: 41016673-41016673 on Build GRCh38
rs779388845 Chr.19: 41016596-41016596 on Build GRCh38
rs148520554 Chr.19: 41016823-41016823 on Build GRCh38
rs146956370 Chr.19: 41016686-41016686 on Build GRCh38
rs996988645 Chr.19: 41016953-41016953 on Build GRCh38
rs143488417 Chr.19: 41016674-41016674 on Build GRCh38
rs371613060 Chr.19: 41016584-41016584 on Build GRCh38
rs368531402 Chr.19: 41016737-41016737 on Build GRCh38
rs149386398 Chr.19: 41016811-41016811 on Build GRCh38
rs764180145 Chr.19: 41016767-41016767 on Build GRCh38
rs760667555 Chr.19: 41016684-41016684 on Build GRCh38
rs200993638 Chr.19: 41016743-41016743 on Build GRCh38
rs144728904 Chr.19: 41016818-41016818 on Build GRCh38
rs3211373 Chr.19: 41016889-41016889 on Build GRCh38
rs773855178 Chr.19: 41016617-41016618 on Build GRCh38
rs28399501 Chr.19: 41016853-41016853 on Build GRCh38
rs541481501 Chr.19: 41016675-41016675 on Build GRCh38
rs762824521 Chr.19: 41016622-41016622 on Build GRCh38
rs937977291 Chr.19: 41016830-41016830 on Build GRCh38
rs373442191 Chr.19: 41016738-41016738 on Build GRCh38
rs771841772 Chr.19: 41016550-41016550 on Build GRCh38
rs2291287 Chr.19: 41016546-41016546 on Build GRCh38
rs759346857 Chr.19: 41016639-41016639 on Build GRCh38
rs760602974 Chr.19: 41016766-41016766 on Build GRCh38
rs537701423 Chr.19: 41016606-41016606 on Build GRCh38
rs374099483 Chr.19: 41016679-41016679 on Build GRCh38
rs745638870 Chr.19: 41016752-41016752 on Build GRCh38
rs3211372 Chr.19: 41016878-41016878 on Build GRCh38
rs368451099 Chr.19: 41016682-41016682 on Build GRCh38
rs751814945 Chr.19: 41016956-41016956 on Build GRCh38
rs767509932 Chr.19: 41016796-41016797 on Build GRCh38
rs3211371 Chr.19: 41016810-41016810 on Build GRCh38
rs868672028 Chr.19: 41016947-41016947 on Build GRCh38
rs534996226 Chr.19: 41016954-41016954 on Build GRCh38
rs571199677 Chr.19: 41016949-41016949 on Build GRCh38
rs772840375 Chr.19: 41016819-41016819 on Build GRCh38
rs758789145 Chr.19: 41016670-41016670 on Build GRCh38
rs913613457 Chr.19: 41016602-41016602 on Build GRCh38
rs769297320 Chr.19: 41016612-41016612 on Build GRCh38
rs766696039 Chr.19: 41016793-41016793 on Build GRCh38
rs146837827 Chr.19: 41016653-41016653 on Build GRCh38
rs989185037 Chr.19: 41016555-41016555 on Build GRCh38
rs761596792 Chr.19: 41016792-41016792 on Build GRCh38
rs138053056 Chr.19: 41016906-41016906 on Build GRCh38
rs747756796 Chr.19: 41016729-41016729 on Build GRCh38
rs370591767 Chr.19: 41016651-41016651 on Build GRCh38
rs3211374 Chr.19: 41016935-41016935 on Build GRCh38
rs536428142 Chr.19: 41016559-41016559 on Build GRCh38
rs746980213 Chr.19: 41016846-41016846 on Build GRCh38
rs796553275 Chr.19: 41016536-41016536 on Build GRCh38
rs982356966 Chr.19: 41016792-41016792 on Build GRCh38
rs34749331 Chr.19: 41016899-41016899 on Build GRCh38
rs147991149 Chr.19: 41016678-41016678 on Build GRCh38
rs575892862 Chr.19: 41016871-41016871 on Build GRCh38
rs3211369 Chr.19: 41016726-41016726 on Build GRCh38
rs762060128 Chr.19: 41016774-41016774 on Build GRCh38
rs775699069 Chr.19: 41016683-41016683 on Build GRCh38
rs773187037 Chr.19: 41016621-41016621 on Build GRCh38
rs371998889 Chr.19: 41016725-41016725 on Build GRCh38
rs201028567 Chr.19: 41016727-41016727 on Build GRCh38
rs754073421 Chr.19: 41016695-41016695 on Build GRCh38
rs779190572 Chr.19: 41016662-41016662 on Build GRCh38
rs117872433 Chr.19: 41016741-41016741 on Build GRCh38
rs766486432 Chr.19: 41016630-41016630 on Build GRCh38
rs764288403 Chr.19: 41016652-41016652 on Build GRCh38
rs28399502 Chr.19: 41016965-41016965 on Build GRCh38
rs377238819 Chr.19: 41016877-41016877 on Build GRCh38
rs3211375 Chr.19: 41016980-41016980 on Build GRCh38
rs564083989 Chr.19: 41016778-41016778 on Build GRCh38
rs771776838 Chr.19: 41016842-41016842 on Build GRCh38
rs560614884 Chr.19: 41016543-41016543 on Build GRCh38
rs147410578 Chr.19: 41016765-41016765 on Build GRCh38
rs1028026025 Chr.19: 41016799-41016799 on Build GRCh38
rs775953206 Chr.19: 41016607-41016607 on Build GRCh38
rs772114611 Chr.19: 41016755-41016755 on Build GRCh38
rs148856816 Chr.19: 41016664-41016664 on Build GRCh38
rs779849859 Chr.19: 41016845-41016845 on Build GRCh38
rs763219706 Chr.19: 41016779-41016779 on Build GRCh38
rs3211370 Chr.19: 41016746-41016746 on Build GRCh38
rs767669267 Chr.19: 41016644-41016644 on Build GRCh38
rs776738671 Chr.19: 41016859-41016859 on Build GRCh38
rs770065427 Chr.19: 41016873-41016873 on Build GRCh38
rs576578621 Chr.19: 41016594-41016594 on Build GRCh38
rs751560753 Chr.19: 41016631-41016631 on Build GRCh38
rs927874683 Chr.19: 41016848-41016848 on Build GRCh38
rs748895233 Chr.19: 41016676-41016676 on Build GRCh38
rs746306498 Chr.19: 41016598-41016598 on Build GRCh38
rs945106154 Chr.19: 41016820-41016820 on Build GRCh38
rs1004284929 Chr.19: 41016897-41016897 on Build GRCh38
rs757031032 Chr.19: 41016822-41016822 on Build GRCh38
rs199591919 Chr.19: 41016758-41016758 on Build GRCh38

View Full Product Details