Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41276213-41276724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4602651 c.724C>A p.L242M Chr.19: 41276236-41276236 on Build GRCh38
6525027 c.733G>A p.G245R Chr.19: 41276245-41276245 on Build GRCh38
6244034 c.764G>T p.G255V Chr.19: 41276276-41276276 on Build GRCh38
7202180 c.786G>A p.K262K Chr.19: 41276298-41276298 on Build GRCh38
7112937 c.575C>G p.T192R Chr.19: 41276216-41276216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780083454 Chr.19: 41276365-41276365 on Build GRCh38
rs367849996 Chr.19: 41276325-41276325 on Build GRCh38
rs910855945 Chr.19: 41276551-41276551 on Build GRCh38
rs144718578 Chr.19: 41276289-41276289 on Build GRCh38
rs190035301 Chr.19: 41276472-41276472 on Build GRCh38
rs888567568 Chr.19: 41276425-41276425 on Build GRCh38
rs77028936 Chr.19: 41276397-41276397 on Build GRCh38
rs767128887 Chr.19: 41276295-41276295 on Build GRCh38
rs574006891 Chr.19: 41276694-41276694 on Build GRCh38
rs1001803635 Chr.19: 41276631-41276631 on Build GRCh38
rs141554220 Chr.19: 41276438-41276438 on Build GRCh38
rs200941091 Chr.19: 41276338-41276338 on Build GRCh38
rs748399776 Chr.19: 41276381-41276381 on Build GRCh38
rs935207979 Chr.19: 41276590-41276590 on Build GRCh38
rs747445224 Chr.19: 41276270-41276270 on Build GRCh38
rs1004686512 Chr.19: 41276437-41276437 on Build GRCh38
rs747132376 Chr.19: 41276370-41276370 on Build GRCh38
rs959591186 Chr.19: 41276493-41276493 on Build GRCh38
rs769253732 Chr.19: 41276273-41276273 on Build GRCh38
rs149059713 Chr.19: 41276258-41276258 on Build GRCh38
rs994378504 Chr.19: 41276699-41276699 on Build GRCh38
rs753513620 Chr.19: 41276324-41276324 on Build GRCh38
rs986415098 Chr.19: 41276310-41276310 on Build GRCh38
rs145084008 Chr.19: 41276613-41276613 on Build GRCh38
rs116292826 Chr.19: 41276371-41276371 on Build GRCh38
rs752292786 Chr.19: 41276308-41276308 on Build GRCh38
rs374879490 Chr.19: 41276312-41276312 on Build GRCh38
rs762268558 Chr.19: 41276279-41276279 on Build GRCh38
rs955290580 Chr.19: 41276494-41276494 on Build GRCh38
rs750640915 Chr.19: 41276351-41276351 on Build GRCh38
rs932546406 Chr.19: 41276684-41276684 on Build GRCh38
rs990013708 Chr.19: 41276331-41276331 on Build GRCh38
rs1014298530 Chr.19: 41276384-41276384 on Build GRCh38
rs913354438 Chr.19: 41276600-41276600 on Build GRCh38
rs759840852 Chr.19: 41276571-41276571 on Build GRCh38
rs1048674379 Chr.19: 41276417-41276417 on Build GRCh38
rs139663340 Chr.19: 41276579-41276579 on Build GRCh38
rs965049633 Chr.19: 41276594-41276594 on Build GRCh38
rs765713526 Chr.19: 41276548-41276548 on Build GRCh38
rs968846164 Chr.19: 41276346-41276346 on Build GRCh38
rs1011088723 Chr.19: 41276515-41276515 on Build GRCh38
rs150909142 Chr.19: 41276572-41276572 on Build GRCh38
rs770265326 Chr.19: 41276395-41276395 on Build GRCh38
rs770641111 Chr.19: 41276281-41276281 on Build GRCh38
rs962885789 Chr.19: 41276445-41276446 on Build GRCh38
rs762479094 Chr.19: 41276638-41276638 on Build GRCh38
rs759317274 Chr.19: 41276293-41276293 on Build GRCh38
rs950074303 Chr.19: 41276600-41276600 on Build GRCh38
rs905833871 Chr.19: 41276614-41276614 on Build GRCh38
rs921201739 Chr.19: 41276636-41276636 on Build GRCh38
rs977573959 Chr.19: 41276577-41276577 on Build GRCh38
rs994724119 Chr.19: 41276445-41276445 on Build GRCh38
rs181245835 Chr.19: 41276541-41276541 on Build GRCh38
rs867953637 Chr.19: 41276285-41276285 on Build GRCh38
rs781724680 Chr.19: 41276377-41276377 on Build GRCh38
rs772125858 Chr.19: 41276252-41276252 on Build GRCh38
rs1038638623 Chr.19: 41276635-41276635 on Build GRCh38
rs760023830 Chr.19: 41276510-41276510 on Build GRCh38
rs753983947 Chr.19: 41276368-41276368 on Build GRCh38
rs768661300 Chr.19: 41276383-41276384 on Build GRCh38
rs764126037 Chr.19: 41276321-41276321 on Build GRCh38
rs760853894 Chr.19: 41276378-41276378 on Build GRCh38
rs144925593 Chr.19: 41276247-41276247 on Build GRCh38
rs1003462380 Chr.19: 41276483-41276483 on Build GRCh38
rs906116689 Chr.19: 41276410-41276410 on Build GRCh38
rs1033894080 Chr.19: 41276492-41276492 on Build GRCh38
rs776963747 Chr.19: 41276455-41276455 on Build GRCh38
rs762819825 Chr.19: 41276381-41276382 on Build GRCh38
rs932504945 Chr.19: 41276401-41276401 on Build GRCh38
rs986434760 Chr.19: 41276546-41276546 on Build GRCh38
rs758661018 Chr.19: 41276359-41276359 on Build GRCh38
rs977145663 Chr.19: 41276361-41276361 on Build GRCh38
rs551034665 Chr.19: 41276588-41276592 on Build GRCh38
rs773776201 Chr.19: 41276396-41276396 on Build GRCh38

View Full Product Details