Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41294029-41294527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4938877 c.1307C>A p.T436K Chr.19: 41294378-41294378 on Build GRCh38
4938878 c.965C>A p.T322K Chr.19: 41294378-41294378 on Build GRCh38
6319159 c.941G>T p.G314V Chr.19: 41294354-41294354 on Build GRCh38
3990054 c.947C>G p.P316R Chr.19: 41294360-41294360 on Build GRCh38
6681472 c.941G>A p.G314D Chr.19: 41294354-41294354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928691522 Chr.19: 41294131-41294131 on Build GRCh38
rs900491518 Chr.19: 41294286-41294286 on Build GRCh38
rs981566568 Chr.19: 41294116-41294116 on Build GRCh38
rs991448203 Chr.19: 41294142-41294142 on Build GRCh38
rs780146476 Chr.19: 41294397-41294397 on Build GRCh38
rs545171512 Chr.19: 41294262-41294262 on Build GRCh38
rs143230050 Chr.19: 41294103-41294103 on Build GRCh38
rs540785702 Chr.19: 41294483-41294483 on Build GRCh38
rs764631374 Chr.19: 41294352-41294352 on Build GRCh38
rs748283525 Chr.19: 41294439-41294439 on Build GRCh38
rs573805535 Chr.19: 41294187-41294187 on Build GRCh38
rs937393270 Chr.19: 41294132-41294132 on Build GRCh38
rs1008919376 Chr.19: 41294239-41294239 on Build GRCh38
rs375451305 Chr.19: 41294492-41294492 on Build GRCh38
rs753429835 Chr.19: 41294321-41294321 on Build GRCh38
rs758375580 Chr.19: 41294386-41294386 on Build GRCh38
rs988908623 Chr.19: 41294403-41294403 on Build GRCh38
rs140222084 Chr.19: 41294281-41294281 on Build GRCh38
rs754804563 Chr.19: 41294415-41294415 on Build GRCh38
rs749838914 Chr.19: 41294499-41294499 on Build GRCh38
rs774657569 Chr.19: 41294308-41294308 on Build GRCh38
rs557806341 Chr.19: 41294100-41294100 on Build GRCh38
rs763347436 Chr.19: 41294304-41294304 on Build GRCh38
rs1041925715 Chr.19: 41294177-41294177 on Build GRCh38
rs866379796 Chr.19: 41294160-41294160 on Build GRCh38
rs544216064 Chr.19: 41294224-41294224 on Build GRCh38
rs371503869 Chr.19: 41294466-41294466 on Build GRCh38
rs947647833 Chr.19: 41294171-41294171 on Build GRCh38
rs781105837 Chr.19: 41294428-41294428 on Build GRCh38
rs751344376 Chr.19: 41294407-41294407 on Build GRCh38
rs975008262 Chr.19: 41294076-41294076 on Build GRCh38
rs766127859 Chr.19: 41294280-41294280 on Build GRCh38
rs376862674 Chr.19: 41294334-41294334 on Build GRCh38
rs1040155035 Chr.19: 41294191-41294191 on Build GRCh38
rs965354180 Chr.19: 41294057-41294057 on Build GRCh38
rs550673973 Chr.19: 41294067-41294067 on Build GRCh38
rs771359373 Chr.19: 41294473-41294473 on Build GRCh38
rs773081722 Chr.19: 41294186-41294186 on Build GRCh38
rs766893451 Chr.19: 41294305-41294305 on Build GRCh38
rs60968872 Chr.19: 41294225-41294225 on Build GRCh38
rs961927036 Chr.19: 41294340-41294340 on Build GRCh38
rs767930128 Chr.19: 41294312-41294312 on Build GRCh38
rs555251451 Chr.19: 41294178-41294178 on Build GRCh38
rs546013811 Chr.19: 41294090-41294102 on Build GRCh38
rs181945110 Chr.19: 41294095-41294095 on Build GRCh38
rs374735263 Chr.19: 41294467-41294467 on Build GRCh38
rs974032809 Chr.19: 41294165-41294165 on Build GRCh38
rs139573139 Chr.19: 41294102-41294102 on Build GRCh38
rs774827461 Chr.19: 41294476-41294476 on Build GRCh38
rs577545565 Chr.19: 41294230-41294230 on Build GRCh38
rs901579689 Chr.19: 41294214-41294214 on Build GRCh38
rs772395072 Chr.19: 41294484-41294484 on Build GRCh38
rs1020008976 Chr.19: 41294475-41294475 on Build GRCh38
rs560335753 Chr.19: 41294217-41294217 on Build GRCh38
rs535606398 Chr.19: 41294316-41294316 on Build GRCh38
rs560280359 Chr.19: 41294276-41294276 on Build GRCh38
rs887406044 Chr.19: 41294278-41294278 on Build GRCh38
rs981490257 Chr.19: 41294115-41294115 on Build GRCh38
rs937285757 Chr.19: 41294174-41294174 on Build GRCh38
rs957530078 Chr.19: 41294094-41294094 on Build GRCh38
rs373882817 Chr.19: 41294309-41294309 on Build GRCh38
rs780818495 Chr.19: 41294300-41294312 on Build GRCh38
rs569346568 Chr.19: 41294093-41294093 on Build GRCh38
rs949760050 Chr.19: 41294106-41294106 on Build GRCh38
rs926962014 Chr.19: 41294164-41294164 on Build GRCh38
rs775973037 Chr.19: 41294488-41294488 on Build GRCh38
rs748396647 Chr.19: 41294256-41294256 on Build GRCh38
rs866591872 Chr.19: 41294458-41294458 on Build GRCh38
rs749974827 Chr.19: 41294358-41294358 on Build GRCh38

View Full Product Details