Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41810233-41810744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM3
Entrez Gene ID
1084
Gene Name
carcinoembryonic antigen related cell adhesion molecule 3
Gene Aliases
CD66D, CEA, CGM1, W264, W282
UniGene
Hs.11
Gene Chromosome Location
Chr.19: 41796590-41811553 on Build GRCh38
Transcripts
NM_001277163.2 NM_001815.4 NR_102333.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1393997 c.619G>A p.A207T Chr.19: 41810346-41810346 on Build GRCh38
3286407 c.609C>T p.S203S Chr.19: 41810336-41810336 on Build GRCh38
237180 c.621C>A p.A207A Chr.19: 41810348-41810348 on Build GRCh38
4581093 c.599G>A p.R200H Chr.19: 41810326-41810326 on Build GRCh38
6840823 c.596-1G>T p.? Chr.19: 41810322-41810322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782128256 Chr.19: 41810364-41810364 on Build GRCh38
rs138403931 Chr.19: 41810353-41810353 on Build GRCh38
rs781367238 Chr.19: 41810626-41810626 on Build GRCh38
rs957357272 Chr.19: 41810684-41810684 on Build GRCh38
rs782008014 Chr.19: 41810345-41810345 on Build GRCh38
rs531574507 Chr.19: 41810273-41810273 on Build GRCh38
rs936045029 Chr.19: 41810428-41810428 on Build GRCh38
rs368817066 Chr.19: 41810351-41810351 on Build GRCh38
rs782255494 Chr.19: 41810349-41810349 on Build GRCh38
rs565000338 Chr.19: 41810354-41810354 on Build GRCh38
rs1024487795 Chr.19: 41810340-41810340 on Build GRCh38
rs980155410 Chr.19: 41810671-41810671 on Build GRCh38
rs567752972 Chr.19: 41810643-41810643 on Build GRCh38
rs1027946813 Chr.19: 41810283-41810283 on Build GRCh38
rs115812254 Chr.19: 41810535-41810535 on Build GRCh38
rs113198277 Chr.19: 41810485-41810485 on Build GRCh38
rs782616839 Chr.19: 41810306-41810306 on Build GRCh38
rs781864005 Chr.19: 41810285-41810285 on Build GRCh38
rs781865775 Chr.19: 41810720-41810720 on Build GRCh38
rs1024852718 Chr.19: 41810629-41810629 on Build GRCh38
rs890297923 Chr.19: 41810487-41810487 on Build GRCh38
rs900165570 Chr.19: 41810531-41810531 on Build GRCh38
rs372629633 Chr.19: 41810386-41810386 on Build GRCh38
rs116256605 Chr.19: 41810577-41810577 on Build GRCh38
rs782206698 Chr.19: 41810307-41810307 on Build GRCh38
rs543380385 Chr.19: 41810522-41810522 on Build GRCh38
rs111750417 Chr.19: 41810486-41810486 on Build GRCh38
rs200290615 Chr.19: 41810326-41810326 on Build GRCh38
rs782760672 Chr.19: 41810374-41810374 on Build GRCh38
rs542744463 Chr.19: 41810613-41810613 on Build GRCh38
rs112654786 Chr.19: 41810257-41810257 on Build GRCh38
rs112608111 Chr.19: 41810527-41810527 on Build GRCh38
rs782726213 Chr.19: 41810403-41810403 on Build GRCh38
rs781956340 Chr.19: 41810352-41810352 on Build GRCh38
rs782582099 Chr.19: 41810323-41810323 on Build GRCh38
rs964383820 Chr.19: 41810258-41810258 on Build GRCh38
rs377076909 Chr.19: 41810390-41810390 on Build GRCh38
rs373324140 Chr.19: 41810286-41810286 on Build GRCh38
rs782406730 Chr.19: 41810338-41810338 on Build GRCh38
rs369634785 Chr.19: 41810276-41810276 on Build GRCh38
rs1034422377 Chr.19: 41810675-41810675 on Build GRCh38
rs1005149610 Chr.19: 41810620-41810620 on Build GRCh38
rs782750651 Chr.19: 41810274-41810274 on Build GRCh38
rs945329080 Chr.19: 41810476-41810476 on Build GRCh38
rs980157132 Chr.19: 41810376-41810376 on Build GRCh38
rs913249804 Chr.19: 41810703-41810703 on Build GRCh38
rs781862763 Chr.19: 41810272-41810272 on Build GRCh38
rs556219871 Chr.19: 41810694-41810694 on Build GRCh38
rs782010813 Chr.19: 41810363-41810363 on Build GRCh38
rs782739227 Chr.19: 41810288-41810288 on Build GRCh38
rs782319908 Chr.19: 41810308-41810308 on Build GRCh38
rs538084735 Chr.19: 41810644-41810644 on Build GRCh38
rs944405878 Chr.19: 41810513-41810513 on Build GRCh38
rs140895670 Chr.19: 41810471-41810471 on Build GRCh38
rs989340343 Chr.19: 41810445-41810445 on Build GRCh38

View Full Product Details