Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42201908-42202409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEDD2
Entrez Gene ID
162989
Gene Name
death effector domain containing 2
Gene Aliases
FLAME-3
UniGene
Hs.515432
Gene Chromosome Location
Chr.19: 42198593-42220152 on Build GRCh38
Transcripts
NM_001270614.1 NM_001270615.1 NM_133328.3 NR_073046.1 NR_073047.1 NR_073048.1 NR_073049.1 NR_073050.1 NR_073051.1 NR_073052.1 XM_011526569.1 XM_011526571.1 XM_011526572.1 XM_017026402.1 XM_017026403.1

SNP Detail

SNP ID Chromosome Location
rs772510268 Chr.19: 42202233-42202233 on Build GRCh38
rs889344568 Chr.19: 42202022-42202022 on Build GRCh38
rs895535729 Chr.19: 42202372-42202372 on Build GRCh38
rs941004983 Chr.19: 42202319-42202319 on Build GRCh38
rs968001582 Chr.19: 42202206-42202206 on Build GRCh38
rs905540559 Chr.19: 42202145-42202145 on Build GRCh38
rs896874553 Chr.19: 42202324-42202324 on Build GRCh38
rs528590908 Chr.19: 42202147-42202147 on Build GRCh38
rs1037331736 Chr.19: 42202091-42202091 on Build GRCh38
rs895631509 Chr.19: 42202146-42202146 on Build GRCh38
rs943631146 Chr.19: 42202257-42202257 on Build GRCh38
rs759196460 Chr.19: 42202172-42202172 on Build GRCh38
rs898870619 Chr.19: 42202104-42202104 on Build GRCh38
rs935497137 Chr.19: 42202002-42202002 on Build GRCh38
rs749789868 Chr.19: 42202201-42202201 on Build GRCh38
rs993994420 Chr.19: 42202188-42202188 on Build GRCh38
rs993935587 Chr.19: 42202328-42202328 on Build GRCh38
rs1014426948 Chr.19: 42202376-42202376 on Build GRCh38
rs1045713209 Chr.19: 42202301-42202301 on Build GRCh38
rs896703233 Chr.19: 42202080-42202083 on Build GRCh38
rs969965187 Chr.19: 42202164-42202164 on Build GRCh38
rs910714254 Chr.19: 42202244-42202244 on Build GRCh38
rs569011694 Chr.19: 42202109-42202109 on Build GRCh38
rs918099302 Chr.19: 42202245-42202246 on Build GRCh38
rs770563474 Chr.19: 42202351-42202351 on Build GRCh38
rs979440385 Chr.19: 42202219-42202219 on Build GRCh38
rs772943633 Chr.19: 42202258-42202258 on Build GRCh38
rs988168743 Chr.19: 42202220-42202220 on Build GRCh38
rs1006745958 Chr.19: 42202339-42202339 on Build GRCh38
rs943626884 Chr.19: 42202042-42202042 on Build GRCh38
rs914061798 Chr.19: 42201947-42201947 on Build GRCh38
rs146832952 Chr.19: 42202018-42202018 on Build GRCh38
rs769100877 Chr.19: 42202202-42202202 on Build GRCh38
rs924017419 Chr.19: 42202231-42202231 on Build GRCh38
rs550553136 Chr.19: 42202330-42202330 on Build GRCh38
rs756127124 Chr.19: 42202072-42202072 on Build GRCh38
rs948303585 Chr.19: 42202261-42202261 on Build GRCh38
rs928978604 Chr.19: 42202332-42202332 on Build GRCh38
rs760409620 Chr.19: 42202322-42202322 on Build GRCh38
rs758113613 Chr.19: 42202165-42202165 on Build GRCh38
rs888054611 Chr.19: 42202302-42202302 on Build GRCh38
rs933529717 Chr.19: 42202017-42202017 on Build GRCh38
rs896947178 Chr.19: 42202159-42202159 on Build GRCh38
rs779313102 Chr.19: 42202208-42202208 on Build GRCh38
rs186145213 Chr.19: 42202198-42202198 on Build GRCh38
rs987624187 Chr.19: 42202239-42202239 on Build GRCh38
rs954972263 Chr.19: 42202234-42202234 on Build GRCh38
rs577280802 Chr.19: 42201996-42201996 on Build GRCh38
rs955293727 Chr.19: 42202194-42202194 on Build GRCh38
rs781228411 Chr.19: 42201944-42201944 on Build GRCh38
rs764799458 Chr.19: 42202173-42202173 on Build GRCh38
rs530261812 Chr.19: 42201995-42201996 on Build GRCh38
rs1043774747 Chr.19: 42201934-42201934 on Build GRCh38
rs753511208 Chr.19: 42202169-42202171 on Build GRCh38
rs1029912309 Chr.19: 42202189-42202189 on Build GRCh38
rs1044044971 Chr.19: 42202132-42202132 on Build GRCh38
rs1000562467 Chr.19: 42202193-42202193 on Build GRCh38
rs16975684 Chr.19: 42202158-42202158 on Build GRCh38
rs1026097242 Chr.19: 42202117-42202117 on Build GRCh38
rs547041596 Chr.19: 42202150-42202150 on Build GRCh38
rs748422201 Chr.19: 42202210-42202210 on Build GRCh38

View Full Product Details