Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44818431-44818928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAM
Entrez Gene ID
4059
Gene Name
basal cell adhesion molecule (Lutheran blood group)
Gene Aliases
AU, CD239, LU, MSK19
UniGene
Hs.625725
Gene Chromosome Location
Chr.19: 44809059-44821421 on Build GRCh38
Transcripts
NM_001013257.2 NM_005581.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4665301 c.1296G>A p.P432P Chr.19: 44818842-44818842 on Build GRCh38
3535770 c.1155C>T p.S385S Chr.19: 44818598-44818598 on Build GRCh38
3892841 c.1248C>T p.F416F Chr.19: 44818794-44818794 on Build GRCh38
4515611 c.1206_1207CC>TT p.(=) Chr.19: 44818752-44818753 on Build GRCh38
474905 c.1143C>A p.V381V Chr.19: 44818586-44818586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770973186 Chr.19: 44818661-44818661 on Build GRCh38
rs760277909 Chr.19: 44818877-44818877 on Build GRCh38
rs1029962265 Chr.19: 44818466-44818466 on Build GRCh38
rs906138144 Chr.19: 44818808-44818808 on Build GRCh38
rs751089732 Chr.19: 44818700-44818700 on Build GRCh38
rs754260197 Chr.19: 44818775-44818775 on Build GRCh38
rs747031001 Chr.19: 44818795-44818795 on Build GRCh38
rs28399626 Chr.19: 44818584-44818584 on Build GRCh38
rs767791090 Chr.19: 44818610-44818610 on Build GRCh38
rs755900842 Chr.19: 44818518-44818518 on Build GRCh38
rs367831700 Chr.19: 44818688-44818688 on Build GRCh38
rs534423898 Chr.19: 44818667-44818667 on Build GRCh38
rs200051019 Chr.19: 44818900-44818900 on Build GRCh38
rs753687027 Chr.19: 44818620-44818620 on Build GRCh38
rs372231529 Chr.19: 44818673-44818673 on Build GRCh38
rs770828608 Chr.19: 44818868-44818868 on Build GRCh38
rs558228341 Chr.19: 44818580-44818580 on Build GRCh38
rs980789272 Chr.19: 44818799-44818799 on Build GRCh38
rs766172229 Chr.19: 44818697-44818697 on Build GRCh38
rs28399628 Chr.19: 44818725-44818727 on Build GRCh38
rs745593719 Chr.19: 44818874-44818874 on Build GRCh38
rs757889532 Chr.19: 44818632-44818632 on Build GRCh38
rs754916313 Chr.19: 44818695-44818697 on Build GRCh38
rs778691502 Chr.19: 44818869-44818869 on Build GRCh38
rs369357316 Chr.19: 44818854-44818854 on Build GRCh38
rs771472781 Chr.19: 44818733-44818733 on Build GRCh38
rs138787345 Chr.19: 44818534-44818534 on Build GRCh38
rs867902598 Chr.19: 44818603-44818603 on Build GRCh38
rs148674435 Chr.19: 44818813-44818813 on Build GRCh38
rs749557664 Chr.19: 44818722-44818722 on Build GRCh38
rs756633287 Chr.19: 44818564-44818564 on Build GRCh38
rs758616131 Chr.19: 44818786-44818786 on Build GRCh38
rs948029601 Chr.19: 44818616-44818616 on Build GRCh38
rs144034004 Chr.19: 44818802-44818802 on Build GRCh38
rs752402705 Chr.19: 44818767-44818767 on Build GRCh38
rs774984772 Chr.19: 44818823-44818823 on Build GRCh38
rs745963661 Chr.19: 44818736-44818736 on Build GRCh38
rs953011979 Chr.19: 44818491-44818491 on Build GRCh38
rs139711671 Chr.19: 44818454-44818454 on Build GRCh38
rs558036872 Chr.19: 44818779-44818779 on Build GRCh38
rs879043866 Chr.19: 44818562-44818562 on Build GRCh38
rs763974366 Chr.19: 44818520-44818520 on Build GRCh38
rs200459189 Chr.19: 44818715-44818717 on Build GRCh38
rs746927559 Chr.19: 44818473-44818473 on Build GRCh38
rs774503138 Chr.19: 44818604-44818604 on Build GRCh38
rs748544427 Chr.19: 44818654-44818654 on Build GRCh38
rs760062678 Chr.19: 44818826-44818826 on Build GRCh38
rs755804065 Chr.19: 44818772-44818772 on Build GRCh38
rs150054323 Chr.19: 44818875-44818875 on Build GRCh38
rs775357305 Chr.19: 44818876-44818876 on Build GRCh38
rs759453985 Chr.19: 44818816-44818816 on Build GRCh38
rs761656868 Chr.19: 44818626-44818626 on Build GRCh38
rs141978868 Chr.19: 44818565-44818565 on Build GRCh38
rs764928507 Chr.19: 44818896-44818896 on Build GRCh38
rs779562413 Chr.19: 44818513-44818514 on Build GRCh38
rs765048649 Chr.19: 44818677-44818677 on Build GRCh38
rs150798131 Chr.19: 44818852-44818852 on Build GRCh38
rs373866917 Chr.19: 44818726-44818728 on Build GRCh38
rs374739045 Chr.19: 44818675-44818675 on Build GRCh38
rs754642486 Chr.19: 44818646-44818646 on Build GRCh38
rs543049924 Chr.19: 44818628-44818628 on Build GRCh38
rs1057374445 Chr.19: 44818834-44818834 on Build GRCh38
rs757873954 Chr.19: 44818553-44818553 on Build GRCh38
rs183403648 Chr.19: 44818453-44818453 on Build GRCh38
rs368492974 Chr.19: 44818671-44818671 on Build GRCh38
rs141093684 Chr.19: 44818529-44818529 on Build GRCh38
rs763023271 Chr.19: 44818495-44818495 on Build GRCh38
rs775480071 Chr.19: 44818517-44818517 on Build GRCh38
rs142196330 Chr.19: 44818841-44818841 on Build GRCh38
rs772253850 Chr.19: 44818737-44818737 on Build GRCh38
rs184565168 Chr.19: 44818758-44818758 on Build GRCh38
rs767383930 Chr.19: 44818817-44818817 on Build GRCh38
rs756536064 Chr.19: 44818710-44818710 on Build GRCh38
rs766542678 Chr.19: 44818496-44818496 on Build GRCh38
rs775892338 Chr.19: 44818742-44818742 on Build GRCh38
rs200421757 Chr.19: 44818627-44818627 on Build GRCh38
rs750824810 Chr.19: 44818899-44818899 on Build GRCh38
rs748757091 Chr.19: 44818677-44818677 on Build GRCh38
rs200516188 Chr.19: 44818587-44818587 on Build GRCh38
rs748454641 Chr.19: 44818579-44818579 on Build GRCh38
rs758989604 Chr.19: 44818844-44818844 on Build GRCh38
rs560312954 Chr.19: 44818486-44818486 on Build GRCh38
rs748364445 Chr.19: 44818757-44818757 on Build GRCh38
rs201837153 Chr.19: 44818809-44818809 on Build GRCh38
rs776971856 Chr.19: 44818577-44818577 on Build GRCh38
rs542634988 Chr.19: 44818889-44818889 on Build GRCh38
rs371664685 Chr.19: 44818776-44818776 on Build GRCh38
rs78539981 Chr.19: 44818502-44818502 on Build GRCh38
rs28399627 Chr.19: 44818648-44818648 on Build GRCh38
rs757677631 Chr.19: 44818842-44818842 on Build GRCh38
rs778185476 Chr.19: 44818714-44818714 on Build GRCh38
rs935419772 Chr.19: 44818586-44818586 on Build GRCh38
rs188855455 Chr.19: 44818533-44818533 on Build GRCh38
rs750396520 Chr.19: 44818678-44818678 on Build GRCh38
rs766985414 Chr.19: 44818763-44818763 on Build GRCh38
rs767917162 Chr.19: 44818545-44818545 on Build GRCh38
rs547538508 Chr.19: 44818701-44818701 on Build GRCh38
rs755732107 Chr.19: 44818479-44818479 on Build GRCh38
rs556188971 Chr.19: 44818752-44818752 on Build GRCh38
rs560266052 Chr.19: 44818856-44818856 on Build GRCh38
rs1037856539 Chr.19: 44818804-44818804 on Build GRCh38
rs192417687 Chr.19: 44818660-44818660 on Build GRCh38
rs1047778550 Chr.19: 44818608-44818608 on Build GRCh38
rs776409244 Chr.19: 44818892-44818892 on Build GRCh38
rs757273835 Chr.19: 44818530-44818530 on Build GRCh38
rs1031587411 Chr.19: 44818761-44818761 on Build GRCh38
rs759819008 Chr.19: 44818605-44818605 on Build GRCh38
rs781526566 Chr.19: 44818864-44818864 on Build GRCh38
rs896156795 Chr.19: 44818840-44818840 on Build GRCh38
rs370475792 Chr.19: 44818645-44818645 on Build GRCh38
rs536226868 Chr.19: 44818741-44818741 on Build GRCh38
rs752326816 Chr.19: 44818706-44818706 on Build GRCh38
rs28399629 Chr.19: 44818773-44818773 on Build GRCh38
rs201016308 Chr.19: 44818895-44818895 on Build GRCh38
rs1016964617 Chr.19: 44818642-44818642 on Build GRCh38
rs189399192 Chr.19: 44818794-44818794 on Build GRCh38
rs191092575 Chr.19: 44818812-44818812 on Build GRCh38
rs773077483 Chr.19: 44818753-44818753 on Build GRCh38
rs76476923 Chr.19: 44818470-44818470 on Build GRCh38
rs751031540 Chr.19: 44818843-44818843 on Build GRCh38
rs138302587 Chr.19: 44818599-44818599 on Build GRCh38
rs374322411 Chr.19: 44818664-44818664 on Build GRCh38
rs368301946 Chr.19: 44818656-44818656 on Build GRCh38
rs974633070 Chr.19: 44818903-44818903 on Build GRCh38
rs376225549 Chr.19: 44818853-44818853 on Build GRCh38
rs371311462 Chr.19: 44818547-44818547 on Build GRCh38
rs774225110 Chr.19: 44818759-44818759 on Build GRCh38
rs760977151 Chr.19: 44818750-44818750 on Build GRCh38
rs763826249 Chr.19: 44818835-44818835 on Build GRCh38
rs567457664 Chr.19: 44818704-44818704 on Build GRCh38

View Full Product Details