Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44944669-44945178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOC4
Entrez Gene ID
346
Gene Name
apolipoprotein C4
Gene Aliases
APO-CIV, APOC-IV
UniGene
Hs.731599
Gene Chromosome Location
Chr.19: 44942238-44945496 on Build GRCh38
Transcripts
NM_001646.2 NR_037932.1

Gene Symbol
APOC4-APOC2
Entrez Gene ID
100533990
Gene Name
APOC4-APOC2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.731599
Gene Chromosome Location
Chr.19: 44942238-44949565 on Build GRCh38
Transcripts
NM_001646.2 NR_037932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4132109 c.107T>C p.L36P Chr.19: 44944779-44944779 on Build GRCh38
32603 c.224C>A p.P75Q Chr.19: 44945145-44945145 on Build GRCh38
7263122 c.240C>A p.G80G Chr.19: 44945161-44945161 on Build GRCh38
7264499 c.219-4G>A p.? Chr.19: 44945136-44945136 on Build GRCh38
5426477 c.235C>T p.R79W Chr.19: 44945156-44945156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751351526 Chr.19: 44944739-44944739 on Build GRCh38
rs145341903 Chr.19: 44944846-44944846 on Build GRCh38
rs1002018645 Chr.19: 44945002-44945002 on Build GRCh38
rs966549945 Chr.19: 44944970-44944970 on Build GRCh38
rs531833142 Chr.19: 44944784-44944784 on Build GRCh38
rs569192609 Chr.19: 44945072-44945072 on Build GRCh38
rs551847201 Chr.19: 44944785-44944785 on Build GRCh38
rs758859455 Chr.19: 44944874-44944874 on Build GRCh38
rs547911169 Chr.19: 44944997-44944997 on Build GRCh38
rs5165 Chr.19: 44945073-44945073 on Build GRCh38
rs753474703 Chr.19: 44944762-44944762 on Build GRCh38
rs949447567 Chr.19: 44945070-44945070 on Build GRCh38
rs780724569 Chr.19: 44944788-44944788 on Build GRCh38
rs752572376 Chr.19: 44944756-44944756 on Build GRCh38
rs767445579 Chr.19: 44944750-44944750 on Build GRCh38
rs10423683 Chr.19: 44944783-44944783 on Build GRCh38
rs1037871408 Chr.19: 44945127-44945127 on Build GRCh38
rs961092017 Chr.19: 44944731-44944731 on Build GRCh38
rs368881819 Chr.19: 44944906-44944906 on Build GRCh38
rs375438169 Chr.19: 44945122-44945122 on Build GRCh38
rs371145349 Chr.19: 44945112-44945112 on Build GRCh38
rs573619009 Chr.19: 44944700-44944700 on Build GRCh38
rs118114277 Chr.19: 44945019-44945019 on Build GRCh38
rs151184718 Chr.19: 44944875-44944875 on Build GRCh38
rs759339514 Chr.19: 44944838-44944838 on Build GRCh38
rs776748082 Chr.19: 44944809-44944809 on Build GRCh38
rs751963712 Chr.19: 44945139-44945139 on Build GRCh38
rs1020777831 Chr.19: 44944943-44944943 on Build GRCh38
rs914025505 Chr.19: 44945052-44945052 on Build GRCh38
rs767541073 Chr.19: 44944846-44944846 on Build GRCh38
rs370516106 Chr.19: 44945156-44945156 on Build GRCh38
rs112321936 Chr.19: 44945026-44945026 on Build GRCh38
rs369933487 Chr.19: 44944780-44944780 on Build GRCh38
rs763645870 Chr.19: 44944866-44944866 on Build GRCh38
rs932258351 Chr.19: 44945131-44945131 on Build GRCh38
rs753096367 Chr.19: 44945145-44945145 on Build GRCh38
rs943523396 Chr.19: 44944816-44944816 on Build GRCh38
rs986622264 Chr.19: 44944703-44944703 on Build GRCh38
rs375945781 Chr.19: 44945146-44945146 on Build GRCh38
rs150665000 Chr.19: 44944758-44944758 on Build GRCh38
rs186448850 Chr.19: 44945128-44945128 on Build GRCh38
rs202196766 Chr.19: 44945149-44945149 on Build GRCh38
rs966659607 Chr.19: 44945058-44945058 on Build GRCh38
rs527786250 Chr.19: 44944947-44944947 on Build GRCh38
rs1132899 Chr.19: 44944779-44944779 on Build GRCh38
rs753375245 Chr.19: 44944868-44944868 on Build GRCh38
rs748260064 Chr.19: 44944907-44944907 on Build GRCh38
rs756927594 Chr.19: 44944869-44944869 on Build GRCh38
rs768894898 Chr.19: 44944789-44944789 on Build GRCh38
rs12691090 Chr.19: 44944873-44944873 on Build GRCh38
rs755642155 Chr.19: 44944757-44944757 on Build GRCh38
rs200355132 Chr.19: 44944861-44944861 on Build GRCh38
rs5164 Chr.19: 44944813-44944813 on Build GRCh38
rs775244436 Chr.19: 44945099-44945099 on Build GRCh38
rs781728790 Chr.19: 44945143-44945143 on Build GRCh38
rs552498742 Chr.19: 44945029-44945029 on Build GRCh38
rs962961182 Chr.19: 44944992-44944992 on Build GRCh38
rs991222035 Chr.19: 44945048-44945048 on Build GRCh38
rs773505291 Chr.19: 44944920-44944920 on Build GRCh38
rs140812055 Chr.19: 44944849-44944849 on Build GRCh38
rs371012807 Chr.19: 44945140-44945140 on Build GRCh38
rs12691089 Chr.19: 44944827-44944827 on Build GRCh38
rs755526225 Chr.19: 44944889-44944889 on Build GRCh38
rs781191811 Chr.19: 44944903-44944903 on Build GRCh38
rs34523211 Chr.19: 44945012-44945012 on Build GRCh38
rs773240618 Chr.19: 44944828-44944828 on Build GRCh38
rs769704371 Chr.19: 44944909-44944909 on Build GRCh38
rs147674610 Chr.19: 44944847-44944847 on Build GRCh38
rs770894239 Chr.19: 44944973-44944973 on Build GRCh38
rs766906156 Chr.19: 44945133-44945133 on Build GRCh38
rs1007986127 Chr.19: 44944919-44944920 on Build GRCh38
rs113292373 Chr.19: 44945097-44945097 on Build GRCh38
rs111851340 Chr.19: 44945138-44945138 on Build GRCh38
rs764950395 Chr.19: 44944871-44944871 on Build GRCh38
rs1003837333 Chr.19: 44944914-44944914 on Build GRCh38
rs5166 Chr.19: 44945124-44945124 on Build GRCh38
rs761402543 Chr.19: 44945118-44945118 on Build GRCh38
rs750133809 Chr.19: 44945123-44945123 on Build GRCh38
rs867200293 Chr.19: 44945060-44945060 on Build GRCh38
rs777229733 Chr.19: 44944981-44944981 on Build GRCh38
rs747564547 Chr.19: 44944876-44944876 on Build GRCh38
rs760665826 Chr.19: 44945103-44945103 on Build GRCh38
rs766133260 Chr.19: 44944721-44944721 on Build GRCh38
rs749470744 Chr.19: 44945094-44945094 on Build GRCh38
rs373733705 Chr.19: 44945144-44945144 on Build GRCh38
rs777741441 Chr.19: 44945045-44945045 on Build GRCh38
rs759455284 Chr.19: 44944747-44944747 on Build GRCh38
rs764088242 Chr.19: 44945110-44945110 on Build GRCh38

View Full Product Details