Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45059182-45059676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASRP
Entrez Gene ID
11129
Gene Name
CLK4 associating serine/arginine rich protein
Gene Aliases
CLASP, SFRS16, SWAP2
UniGene
Hs.466917
Gene Chromosome Location
Chr.19: 45039040-45070956 on Build GRCh38
Transcripts
NM_001278439.1 NM_007056.2 NR_103529.1 XM_011526396.2 XM_011526397.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5671166 c.616G>T p.V206L Chr.19: 45059270-45059270 on Build GRCh38
474911 c.643C>A p.Q215K Chr.19: 45059297-45059297 on Build GRCh38
334122 c.652G>T p.V218L Chr.19: 45059306-45059306 on Build GRCh38
6629748 c.628G>A p.V210M Chr.19: 45059282-45059282 on Build GRCh38
33554 c.638T>C p.L213S Chr.19: 45059292-45059292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005367931 Chr.19: 45059215-45059215 on Build GRCh38
rs1036828179 Chr.19: 45059513-45059513 on Build GRCh38
rs564174686 Chr.19: 45059514-45059514 on Build GRCh38
rs748440657 Chr.19: 45059220-45059220 on Build GRCh38
rs555648853 Chr.19: 45059432-45059432 on Build GRCh38
rs1024500970 Chr.19: 45059572-45059572 on Build GRCh38
rs559705747 Chr.19: 45059617-45059617 on Build GRCh38
rs371679555 Chr.19: 45059388-45059388 on Build GRCh38
rs571165211 Chr.19: 45059364-45059364 on Build GRCh38
rs754584233 Chr.19: 45059317-45059317 on Build GRCh38
rs762690474 Chr.19: 45059524-45059524 on Build GRCh38
rs764872009 Chr.19: 45059385-45059385 on Build GRCh38
rs374822691 Chr.19: 45059403-45059403 on Build GRCh38
rs1015002415 Chr.19: 45059423-45059423 on Build GRCh38
rs778216168 Chr.19: 45059347-45059347 on Build GRCh38
rs112316091 Chr.19: 45059585-45059585 on Build GRCh38
rs940808028 Chr.19: 45059434-45059434 on Build GRCh38
rs765099804 Chr.19: 45059275-45059275 on Build GRCh38
rs60962959 Chr.19: 45059255-45059255 on Build GRCh38
rs772166581 Chr.19: 45059362-45059362 on Build GRCh38
rs368514703 Chr.19: 45059270-45059270 on Build GRCh38
rs2304195 Chr.19: 45059613-45059613 on Build GRCh38
rs761821294 Chr.19: 45059378-45059378 on Build GRCh38
rs773267958 Chr.19: 45059237-45059237 on Build GRCh38
rs916369722 Chr.19: 45059557-45059557 on Build GRCh38
rs149597400 Chr.19: 45059404-45059404 on Build GRCh38
rs201266374 Chr.19: 45059269-45059269 on Build GRCh38
rs538127445 Chr.19: 45059236-45059236 on Build GRCh38
rs139288411 Chr.19: 45059258-45059258 on Build GRCh38
rs183322884 Chr.19: 45059638-45059638 on Build GRCh38
rs766201052 Chr.19: 45059287-45059287 on Build GRCh38
rs751101053 Chr.19: 45059296-45059296 on Build GRCh38
rs756386543 Chr.19: 45059338-45059338 on Build GRCh38
rs770234179 Chr.19: 45059233-45059233 on Build GRCh38
rs144325640 Chr.19: 45059424-45059424 on Build GRCh38
rs60565639 Chr.19: 45059502-45059502 on Build GRCh38
rs751010059 Chr.19: 45059635-45059635 on Build GRCh38
rs532813530 Chr.19: 45059515-45059515 on Build GRCh38
rs760869853 Chr.19: 45059332-45059332 on Build GRCh38
rs771142252 Chr.19: 45059350-45059350 on Build GRCh38
rs369229486 Chr.19: 45059589-45059589 on Build GRCh38
rs537954447 Chr.19: 45059228-45059228 on Build GRCh38
rs936311454 Chr.19: 45059386-45059386 on Build GRCh38
rs758072117 Chr.19: 45059281-45059281 on Build GRCh38
rs1014953869 Chr.19: 45059229-45059229 on Build GRCh38
rs529587555 Chr.19: 45059622-45059622 on Build GRCh38
rs543982570 Chr.19: 45059454-45059454 on Build GRCh38
rs367946378 Chr.19: 45059647-45059647 on Build GRCh38
rs549395326 Chr.19: 45059433-45059433 on Build GRCh38
rs1034349469 Chr.19: 45059520-45059520 on Build GRCh38
rs766107907 Chr.19: 45059395-45059395 on Build GRCh38
rs770944052 Chr.19: 45059244-45059244 on Build GRCh38
rs370825293 Chr.19: 45059393-45059393 on Build GRCh38
rs977758410 Chr.19: 45059488-45059488 on Build GRCh38
rs371841201 Chr.19: 45059276-45059276 on Build GRCh38
rs940665144 Chr.19: 45059646-45059646 on Build GRCh38
rs199842664 Chr.19: 45059348-45059348 on Build GRCh38
rs1014908660 Chr.19: 45059529-45059529 on Build GRCh38
rs368668967 Chr.19: 45059360-45059360 on Build GRCh38
rs767159683 Chr.19: 45059405-45059405 on Build GRCh38
rs781756057 Chr.19: 45059331-45059331 on Build GRCh38

View Full Product Details