Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45061866-45062364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASRP
Entrez Gene ID
11129
Gene Name
CLK4 associating serine/arginine rich protein
Gene Aliases
CLASP, SFRS16, SWAP2
UniGene
Hs.466917
Gene Chromosome Location
Chr.19: 45039040-45070956 on Build GRCh38
Transcripts
NM_001278439.1 NM_007056.2 NR_103529.1 XM_011526396.2 XM_011526397.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998127 c.875G>A p.R292Q Chr.19: 45062165-45062165 on Build GRCh38
267848 c.905G>A p.R302Q Chr.19: 45062195-45062195 on Build GRCh38
5565497 c.904C>T p.R302W Chr.19: 45062194-45062194 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755161131 Chr.19: 45062229-45062229 on Build GRCh38
rs901142875 Chr.19: 45062056-45062056 on Build GRCh38
rs558505065 Chr.19: 45062119-45062119 on Build GRCh38
rs761369068 Chr.19: 45062187-45062187 on Build GRCh38
rs1019633088 Chr.19: 45062044-45062044 on Build GRCh38
rs933109726 Chr.19: 45062291-45062291 on Build GRCh38
rs779054791 Chr.19: 45062133-45062133 on Build GRCh38
rs964359472 Chr.19: 45062261-45062261 on Build GRCh38
rs905746937 Chr.19: 45062335-45062335 on Build GRCh38
rs138743378 Chr.19: 45062299-45062299 on Build GRCh38
rs751761241 Chr.19: 45062105-45062105 on Build GRCh38
rs186242903 Chr.19: 45062151-45062151 on Build GRCh38
rs923025297 Chr.19: 45062276-45062276 on Build GRCh38
rs776383104 Chr.19: 45062317-45062318 on Build GRCh38
rs755037319 Chr.19: 45062118-45062118 on Build GRCh38
rs745639921 Chr.19: 45062135-45062135 on Build GRCh38
rs749173386 Chr.19: 45062121-45062121 on Build GRCh38
rs1035786411 Chr.19: 45062062-45062062 on Build GRCh38
rs757259410 Chr.19: 45062130-45062130 on Build GRCh38
rs776111579 Chr.19: 45062186-45062186 on Build GRCh38
rs777806013 Chr.19: 45061926-45061926 on Build GRCh38
rs747266807 Chr.19: 45062033-45062033 on Build GRCh38
rs771807239 Chr.19: 45062137-45062137 on Build GRCh38
rs548819793 Chr.19: 45062013-45062017 on Build GRCh38
rs988568364 Chr.19: 45062292-45062292 on Build GRCh38
rs938315913 Chr.19: 45062230-45062230 on Build GRCh38
rs574046149 Chr.19: 45061980-45061980 on Build GRCh38
rs775154995 Chr.19: 45062148-45062148 on Build GRCh38
rs770844714 Chr.19: 45062082-45062082 on Build GRCh38
rs895866042 Chr.19: 45062113-45062113 on Build GRCh38
rs763336939 Chr.19: 45062221-45062221 on Build GRCh38
rs988694428 Chr.19: 45062256-45062256 on Build GRCh38
rs765710332 Chr.19: 45062189-45062189 on Build GRCh38
rs952859431 Chr.19: 45062143-45062143 on Build GRCh38
rs370518308 Chr.19: 45062313-45062313 on Build GRCh38
rs947220298 Chr.19: 45062323-45062323 on Build GRCh38
rs984327098 Chr.19: 45062149-45062149 on Build GRCh38
rs913192664 Chr.19: 45062319-45062319 on Build GRCh38
rs767920491 Chr.19: 45062233-45062233 on Build GRCh38
rs969857750 Chr.19: 45062165-45062165 on Build GRCh38
rs1011939346 Chr.19: 45061901-45061901 on Build GRCh38
rs377728333 Chr.19: 45062223-45062223 on Build GRCh38
rs1042870142 Chr.19: 45061994-45061994 on Build GRCh38
rs538229503 Chr.19: 45061931-45061931 on Build GRCh38
rs768142018 Chr.19: 45062177-45062177 on Build GRCh38
rs991361547 Chr.19: 45062141-45062141 on Build GRCh38
rs752813945 Chr.19: 45062239-45062239 on Build GRCh38
rs928333640 Chr.19: 45062227-45062227 on Build GRCh38
rs994434726 Chr.19: 45062068-45062068 on Build GRCh38
rs773747275 Chr.19: 45062212-45062212 on Build GRCh38
rs974345539 Chr.19: 45062272-45062272 on Build GRCh38
rs138187879 Chr.19: 45062010-45062014 on Build GRCh38
rs889909397 Chr.19: 45061998-45061998 on Build GRCh38
rs1042963397 Chr.19: 45061978-45061978 on Build GRCh38
rs751748382 Chr.19: 45062228-45062228 on Build GRCh38
rs1043382328 Chr.19: 45062331-45062331 on Build GRCh38

View Full Product Details