Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45785457-45785955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMWD
Entrez Gene ID
1762
Gene Name
dystrophia myotonica, WD repeat containing
Gene Aliases
D19S593E, DMR-N9, DMRN9, gene59
UniGene
Hs.515474
Gene Chromosome Location
Chr.19: 45783006-45792802 on Build GRCh38
Transcripts
NM_004943.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3104627 c.1706C>T p.P569L Chr.19: 45785790-45785790 on Build GRCh38
3104624 c.1725C>T p.P575P Chr.19: 45785771-45785771 on Build GRCh38
3104623 c.1757G>A p.R586H Chr.19: 45785739-45785739 on Build GRCh38
6645748 c.1592G>A p.R531Q Chr.19: 45785904-45785904 on Build GRCh38
3104620 c.1803C>T p.I601I Chr.19: 45785693-45785693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776616640 Chr.19: 45785882-45785884 on Build GRCh38
rs760497739 Chr.19: 45785640-45785640 on Build GRCh38
rs753566101 Chr.19: 45785553-45785553 on Build GRCh38
rs778487763 Chr.19: 45785921-45785921 on Build GRCh38
rs897895942 Chr.19: 45785480-45785480 on Build GRCh38
rs375610071 Chr.19: 45785915-45785915 on Build GRCh38
rs757420017 Chr.19: 45785787-45785787 on Build GRCh38
rs773690612 Chr.19: 45785773-45785773 on Build GRCh38
rs960959552 Chr.19: 45785889-45785889 on Build GRCh38
rs529500976 Chr.19: 45785717-45785717 on Build GRCh38
rs1046884577 Chr.19: 45785749-45785749 on Build GRCh38
rs772758156 Chr.19: 45785583-45785584 on Build GRCh38
rs745868047 Chr.19: 45785789-45785789 on Build GRCh38
rs761349705 Chr.19: 45785567-45785567 on Build GRCh38
rs776485718 Chr.19: 45785741-45785741 on Build GRCh38
rs773062519 Chr.19: 45785753-45785753 on Build GRCh38
rs781640052 Chr.19: 45785693-45785693 on Build GRCh38
rs908604943 Chr.19: 45785487-45785487 on Build GRCh38
rs780647222 Chr.19: 45785593-45785593 on Build GRCh38
rs892911515 Chr.19: 45785917-45785917 on Build GRCh38
rs761140423 Chr.19: 45785853-45785853 on Build GRCh38
rs756069432 Chr.19: 45785904-45785904 on Build GRCh38
rs548973387 Chr.19: 45785722-45785722 on Build GRCh38
rs760146638 Chr.19: 45785817-45785831 on Build GRCh38
rs752150745 Chr.19: 45785678-45785678 on Build GRCh38
rs568725613 Chr.19: 45785732-45785732 on Build GRCh38
rs61738390 Chr.19: 45785831-45785831 on Build GRCh38
rs371055861 Chr.19: 45785849-45785849 on Build GRCh38
rs993992192 Chr.19: 45785493-45785493 on Build GRCh38
rs764861343 Chr.19: 45785569-45785569 on Build GRCh38
rs773920306 Chr.19: 45785830-45785830 on Build GRCh38
rs773121811 Chr.19: 45785804-45785804 on Build GRCh38
rs753119516 Chr.19: 45785779-45785779 on Build GRCh38
rs762559342 Chr.19: 45785555-45785555 on Build GRCh38
rs759256917 Chr.19: 45785889-45785890 on Build GRCh38
rs764464056 Chr.19: 45785860-45785860 on Build GRCh38
rs374890935 Chr.19: 45785927-45785927 on Build GRCh38
rs772013097 Chr.19: 45785633-45785633 on Build GRCh38
rs763784225 Chr.19: 45785547-45785547 on Build GRCh38
rs61744161 Chr.19: 45785819-45785819 on Build GRCh38
rs755420525 Chr.19: 45785682-45785682 on Build GRCh38
rs775271191 Chr.19: 45785541-45785541 on Build GRCh38
rs777914299 Chr.19: 45785720-45785720 on Build GRCh38
rs765980263 Chr.19: 45785677-45785677 on Build GRCh38
rs540419380 Chr.19: 45785496-45785496 on Build GRCh38
rs749962215 Chr.19: 45785667-45785667 on Build GRCh38
rs367851825 Chr.19: 45785931-45785931 on Build GRCh38
rs897928471 Chr.19: 45785734-45785734 on Build GRCh38
rs750640345 Chr.19: 45785895-45785895 on Build GRCh38
rs902923922 Chr.19: 45785554-45785554 on Build GRCh38
rs202210183 Chr.19: 45785739-45785739 on Build GRCh38
rs748148849 Chr.19: 45785901-45785901 on Build GRCh38
rs765315845 Chr.19: 45785894-45785894 on Build GRCh38
rs760124669 Chr.19: 45785774-45785774 on Build GRCh38
rs765984271 Chr.19: 45785781-45785781 on Build GRCh38
rs1045597203 Chr.19: 45785552-45785552 on Build GRCh38
rs749299818 Chr.19: 45785624-45785624 on Build GRCh38
rs868223204 Chr.19: 45785898-45785898 on Build GRCh38
rs1022478634 Chr.19: 45785800-45785800 on Build GRCh38
rs754159564 Chr.19: 45785869-45785869 on Build GRCh38
rs61744166 Chr.19: 45785801-45785801 on Build GRCh38
rs919221090 Chr.19: 45785905-45785905 on Build GRCh38
rs1014483622 Chr.19: 45785548-45785548 on Build GRCh38
rs748456041 Chr.19: 45785603-45785603 on Build GRCh38
rs976444821 Chr.19: 45785531-45785531 on Build GRCh38
rs771959831 Chr.19: 45785511-45785511 on Build GRCh38
rs775557869 Chr.19: 45785637-45785637 on Build GRCh38
rs533385634 Chr.19: 45785491-45785491 on Build GRCh38
rs146577305 Chr.19: 45785751-45785751 on Build GRCh38
rs998643210 Chr.19: 45785568-45785568 on Build GRCh38
rs1055606635 Chr.19: 45785525-45785525 on Build GRCh38
rs770454273 Chr.19: 45785820-45785820 on Build GRCh38
rs776045973 Chr.19: 45785845-45785845 on Build GRCh38
rs367705881 Chr.19: 45785918-45785918 on Build GRCh38
rs1016265087 Chr.19: 45785933-45785933 on Build GRCh38
rs560498280 Chr.19: 45785500-45785500 on Build GRCh38
rs771588244 Chr.19: 45785543-45785543 on Build GRCh38
rs761589099 Chr.19: 45785660-45785660 on Build GRCh38
rs766369677 Chr.19: 45785822-45785830 on Build GRCh38
rs374443086 Chr.19: 45785595-45785595 on Build GRCh38
rs771066909 Chr.19: 45785727-45785727 on Build GRCh38
rs370442722 Chr.19: 45785922-45785922 on Build GRCh38
rs758472529 Chr.19: 45785896-45785896 on Build GRCh38
rs747889131 Chr.19: 45785750-45785750 on Build GRCh38
rs777745709 Chr.19: 45785906-45785906 on Build GRCh38
rs767846091 Chr.19: 45785775-45785775 on Build GRCh38
rs751994584 Chr.19: 45785594-45785594 on Build GRCh38
rs746989063 Chr.19: 45785737-45785737 on Build GRCh38
rs780147513 Chr.19: 45785897-45785897 on Build GRCh38
rs981397103 Chr.19: 45785632-45785632 on Build GRCh38
rs747793318 Chr.19: 45785518-45785519 on Build GRCh38
rs754953416 Chr.19: 45785795-45785795 on Build GRCh38
rs754106400 Chr.19: 45785782-45785782 on Build GRCh38
rs377367082 Chr.19: 45785666-45785666 on Build GRCh38
rs749909031 Chr.19: 45785574-45785574 on Build GRCh38
rs996528856 Chr.19: 45785584-45785584 on Build GRCh38
rs760461902 Chr.19: 45785546-45785546 on Build GRCh38
rs781233466 Chr.19: 45785671-45785671 on Build GRCh38
rs758974926 Chr.19: 45785586-45785586 on Build GRCh38
rs757505317 Chr.19: 45785873-45785873 on Build GRCh38
rs749499836 Chr.19: 45785726-45785726 on Build GRCh38
rs765965554 Chr.19: 45785771-45785771 on Build GRCh38
rs769958519 Chr.19: 45785608-45785608 on Build GRCh38
rs894408840 Chr.19: 45785535-45785535 on Build GRCh38
rs762606932 Chr.19: 45785672-45785672 on Build GRCh38
rs767041578 Chr.19: 45785835-45785835 on Build GRCh38
rs762666229 Chr.19: 45785770-45785770 on Build GRCh38
rs1028556794 Chr.19: 45785591-45785591 on Build GRCh38
rs964086302 Chr.19: 45785526-45785526 on Build GRCh38
rs768542645 Chr.19: 45785642-45785642 on Build GRCh38
rs752885905 Chr.19: 45785916-45785917 on Build GRCh38
rs888382840 Chr.19: 45785510-45785510 on Build GRCh38
rs776239814 Chr.19: 45785647-45785647 on Build GRCh38
rs373052030 Chr.19: 45785600-45785600 on Build GRCh38
rs779080979 Chr.19: 45785788-45785788 on Build GRCh38
rs1038327845 Chr.19: 45785497-45785497 on Build GRCh38
rs1049170544 Chr.19: 45785914-45785914 on Build GRCh38
rs765042909 Chr.19: 45785890-45785890 on Build GRCh38

View Full Product Details