Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45790825-45791319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMWD
Entrez Gene ID
1762
Gene Name
dystrophia myotonica, WD repeat containing
Gene Aliases
D19S593E, DMR-N9, DMRN9, gene59
UniGene
Hs.515474
Gene Chromosome Location
Chr.19: 45783006-45792802 on Build GRCh38
Transcripts
NM_004943.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3990123 c.553G>T p.G185C Chr.19: 45790976-45790976 on Build GRCh38
6645735 c.571G>C p.V191L Chr.19: 45790958-45790958 on Build GRCh38
5786646 c.513C>A p.N171K Chr.19: 45791016-45791016 on Build GRCh38
6382299 c.616A>G p.N206D Chr.19: 45790913-45790913 on Build GRCh38
4412352 c.460C>T p.P154S Chr.19: 45791069-45791069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950835617 Chr.19: 45791207-45791207 on Build GRCh38
rs904067975 Chr.19: 45791158-45791158 on Build GRCh38
rs1016356209 Chr.19: 45791233-45791233 on Build GRCh38
rs760385122 Chr.19: 45791116-45791116 on Build GRCh38
rs748616663 Chr.19: 45791247-45791247 on Build GRCh38
rs763722475 Chr.19: 45791117-45791117 on Build GRCh38
rs759234003 Chr.19: 45790987-45790987 on Build GRCh38
rs764471065 Chr.19: 45791124-45791124 on Build GRCh38
rs142180834 Chr.19: 45790980-45790980 on Build GRCh38
rs952018842 Chr.19: 45791225-45791225 on Build GRCh38
rs148167997 Chr.19: 45790977-45790977 on Build GRCh38
rs199670391 Chr.19: 45791033-45791033 on Build GRCh38
rs776211411 Chr.19: 45791122-45791122 on Build GRCh38
rs754396142 Chr.19: 45790879-45790879 on Build GRCh38
rs746130130 Chr.19: 45790943-45790943 on Build GRCh38
rs376268333 Chr.19: 45790882-45790882 on Build GRCh38
rs190299559 Chr.19: 45791042-45791042 on Build GRCh38
rs955535336 Chr.19: 45791182-45791182 on Build GRCh38
rs1008299781 Chr.19: 45791190-45791190 on Build GRCh38
rs961350022 Chr.19: 45790852-45790852 on Build GRCh38
rs781198854 Chr.19: 45791043-45791043 on Build GRCh38
rs114952745 Chr.19: 45791294-45791294 on Build GRCh38
rs575342659 Chr.19: 45791278-45791278 on Build GRCh38
rs369284999 Chr.19: 45790864-45790864 on Build GRCh38
rs1006754171 Chr.19: 45791224-45791224 on Build GRCh38
rs745427704 Chr.19: 45791095-45791095 on Build GRCh38
rs562449014 Chr.19: 45790846-45790846 on Build GRCh38
rs775197959 Chr.19: 45790991-45790991 on Build GRCh38
rs913670239 Chr.19: 45791025-45791025 on Build GRCh38
rs1054494207 Chr.19: 45791273-45791273 on Build GRCh38
rs146902268 Chr.19: 45791260-45791260 on Build GRCh38
rs533950947 Chr.19: 45791001-45791001 on Build GRCh38
rs757414499 Chr.19: 45791129-45791131 on Build GRCh38
rs185009353 Chr.19: 45791037-45791037 on Build GRCh38
rs766129834 Chr.19: 45790857-45790857 on Build GRCh38
rs762802818 Chr.19: 45790856-45790856 on Build GRCh38
rs773119074 Chr.19: 45790855-45790855 on Build GRCh38
rs745710268 Chr.19: 45791090-45791090 on Build GRCh38
rs756704002 Chr.19: 45790875-45790875 on Build GRCh38
rs924333618 Chr.19: 45791255-45791255 on Build GRCh38
rs762840990 Chr.19: 45791037-45791038 on Build GRCh38
rs945057729 Chr.19: 45791271-45791272 on Build GRCh38
rs778935465 Chr.19: 45791129-45791129 on Build GRCh38
rs371927496 Chr.19: 45791085-45791085 on Build GRCh38
rs767019892 Chr.19: 45790988-45790988 on Build GRCh38
rs1026692715 Chr.19: 45791203-45791203 on Build GRCh38
rs925020327 Chr.19: 45791298-45791298 on Build GRCh38
rs758480945 Chr.19: 45790956-45790956 on Build GRCh38
rs757739782 Chr.19: 45790885-45790885 on Build GRCh38
rs760331922 Chr.19: 45790997-45790997 on Build GRCh38
rs961380631 Chr.19: 45791201-45791201 on Build GRCh38
rs769283540 Chr.19: 45791137-45791137 on Build GRCh38
rs974454812 Chr.19: 45791208-45791208 on Build GRCh38
rs749256726 Chr.19: 45791078-45791078 on Build GRCh38
rs1055332105 Chr.19: 45791189-45791189 on Build GRCh38
rs750691327 Chr.19: 45791031-45791031 on Build GRCh38
rs878963994 Chr.19: 45790942-45790942 on Build GRCh38
rs1054732148 Chr.19: 45791119-45791119 on Build GRCh38
rs975369316 Chr.19: 45790960-45790960 on Build GRCh38
rs770709531 Chr.19: 45791084-45791084 on Build GRCh38
rs902880872 Chr.19: 45791093-45791093 on Build GRCh38
rs1865116 Chr.19: 45791252-45791252 on Build GRCh38
rs771899730 Chr.19: 45791107-45791107 on Build GRCh38
rs774856312 Chr.19: 45791168-45791168 on Build GRCh38
rs202006529 Chr.19: 45790862-45790862 on Build GRCh38
rs72626215 Chr.19: 45790878-45790878 on Build GRCh38
rs749199656 Chr.19: 45790974-45790974 on Build GRCh38
rs770013843 Chr.19: 45791277-45791277 on Build GRCh38
rs370317663 Chr.19: 45790940-45790940 on Build GRCh38
rs765636060 Chr.19: 45791024-45791024 on Build GRCh38
rs915783067 Chr.19: 45791244-45791244 on Build GRCh38
rs757795331 Chr.19: 45791016-45791016 on Build GRCh38
rs896711702 Chr.19: 45791195-45791195 on Build GRCh38
rs779127573 Chr.19: 45790895-45790895 on Build GRCh38
rs780192198 Chr.19: 45790959-45790959 on Build GRCh38
rs920533494 Chr.19: 45791221-45791221 on Build GRCh38
rs141123591 Chr.19: 45791248-45791248 on Build GRCh38
rs377687395 Chr.19: 45790965-45790965 on Build GRCh38
rs763669029 Chr.19: 45790998-45790998 on Build GRCh38
rs374221560 Chr.19: 45790873-45790873 on Build GRCh38
rs747201035 Chr.19: 45791038-45791038 on Build GRCh38
rs547407118 Chr.19: 45791193-45791193 on Build GRCh38
rs991420928 Chr.19: 45791236-45791236 on Build GRCh38
rs879002069 Chr.19: 45791092-45791092 on Build GRCh38
rs1015617745 Chr.19: 45791197-45791197 on Build GRCh38
rs949852902 Chr.19: 45791246-45791246 on Build GRCh38
rs1043474435 Chr.19: 45791143-45791143 on Build GRCh38
rs182121100 Chr.19: 45791073-45791073 on Build GRCh38
rs909684056 Chr.19: 45790900-45790900 on Build GRCh38

View Full Product Details