Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49055546-49056071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGB7
Entrez Gene ID
94027
Gene Name
chorionic gonadotropin beta subunit 7
Gene Aliases
CG-beta-a, CGB6
UniGene
Hs.681647
Gene Chromosome Location
Chr.19: 49054274-49058346 on Build GRCh38
Transcripts
NM_033142.1 XM_017027502.1

SNP Detail

SNP ID Chromosome Location
rs966703040 Chr.19: 49055831-49055831 on Build GRCh38
rs917892177 Chr.19: 49055663-49055663 on Build GRCh38
rs373630051 Chr.19: 49055786-49055786 on Build GRCh38
rs1045432320 Chr.19: 49055690-49055690 on Build GRCh38
rs898761905 Chr.19: 49055796-49055796 on Build GRCh38
rs546727970 Chr.19: 49055705-49055705 on Build GRCh38
rs771749475 Chr.19: 49055849-49055849 on Build GRCh38
rs528046811 Chr.19: 49055677-49055677 on Build GRCh38
rs753143593 Chr.19: 49056036-49056036 on Build GRCh38
rs140652088 Chr.19: 49055738-49055738 on Build GRCh38
rs565022041 Chr.19: 49055904-49055904 on Build GRCh38
rs386810090 Chr.19: 49055794-49055795 on Build GRCh38
rs986776906 Chr.19: 49055643-49055643 on Build GRCh38
rs369281749 Chr.19: 49055584-49055584 on Build GRCh38
rs936520213 Chr.19: 49055777-49055777 on Build GRCh38
rs575008513 Chr.19: 49055623-49055623 on Build GRCh38
rs562537827 Chr.19: 49055961-49055961 on Build GRCh38
rs563218444 Chr.19: 49055607-49055607 on Build GRCh38
rs948445225 Chr.19: 49055688-49055688 on Build GRCh38
rs60722428 Chr.19: 49055754-49055754 on Build GRCh38
rs138960898 Chr.19: 49055871-49055871 on Build GRCh38
rs995709541 Chr.19: 49055713-49055713 on Build GRCh38
rs915679232 Chr.19: 49055685-49055685 on Build GRCh38
rs568681143 Chr.19: 49056046-49056046 on Build GRCh38
rs907465981 Chr.19: 49055575-49055575 on Build GRCh38
rs538459965 Chr.19: 49055800-49055800 on Build GRCh38
rs1022836576 Chr.19: 49055855-49055855 on Build GRCh38
rs566529293 Chr.19: 49055787-49055787 on Build GRCh38
rs35772417 Chr.19: 49055732-49055732 on Build GRCh38
rs373795631 Chr.19: 49055972-49055972 on Build GRCh38
rs939724405 Chr.19: 49055887-49055887 on Build GRCh38
rs924475924 Chr.19: 49055666-49055666 on Build GRCh38
rs965938998 Chr.19: 49055650-49055650 on Build GRCh38
rs924531802 Chr.19: 49055753-49055753 on Build GRCh38
rs181988342 Chr.19: 49055967-49055967 on Build GRCh38
rs532094994 Chr.19: 49055910-49055910 on Build GRCh38
rs34929638 Chr.19: 49055771-49055771 on Build GRCh38
rs867994160 Chr.19: 49055909-49055909 on Build GRCh38
rs928388146 Chr.19: 49055879-49055879 on Build GRCh38
rs560836227 Chr.19: 49055589-49055589 on Build GRCh38
rs1032728119 Chr.19: 49055839-49055839 on Build GRCh38
rs754265541 Chr.19: 49055791-49055791 on Build GRCh38
rs1008303229 Chr.19: 49055817-49055817 on Build GRCh38
rs1028389373 Chr.19: 49055618-49055618 on Build GRCh38
rs575124536 Chr.19: 49055809-49055809 on Build GRCh38
rs1054634925 Chr.19: 49055680-49055680 on Build GRCh38
rs969871775 Chr.19: 49055868-49055868 on Build GRCh38
rs1019647099 Chr.19: 49055830-49055830 on Build GRCh38
rs556463705 Chr.19: 49055801-49055801 on Build GRCh38
rs567291783 Chr.19: 49055974-49055974 on Build GRCh38
rs931624565 Chr.19: 49055898-49055898 on Build GRCh38
rs966676772 Chr.19: 49055746-49055746 on Build GRCh38
rs184259189 Chr.19: 49056033-49056033 on Build GRCh38
rs62127892 Chr.19: 49056031-49056031 on Build GRCh38
rs1023952626 Chr.19: 49055567-49055567 on Build GRCh38
rs912531674 Chr.19: 49055647-49055647 on Build GRCh38
rs566392420 Chr.19: 49055979-49055979 on Build GRCh38
rs56131938 Chr.19: 49055656-49055656 on Build GRCh38
rs552267205 Chr.19: 49056022-49056022 on Build GRCh38
rs889950571 Chr.19: 49055808-49055808 on Build GRCh38
rs974591937 Chr.19: 49055592-49055592 on Build GRCh38
rs553588667 Chr.19: 49056049-49056049 on Build GRCh38
rs902616835 Chr.19: 49055947-49055947 on Build GRCh38
rs1050426039 Chr.19: 49055900-49055900 on Build GRCh38
rs1019614584 Chr.19: 49055740-49055740 on Build GRCh38
rs1004507022 Chr.19: 49055698-49055698 on Build GRCh38
rs569168262 Chr.19: 49055762-49055762 on Build GRCh38
rs996129262 Chr.19: 49055798-49055798 on Build GRCh38
rs1011526569 Chr.19: 49056009-49056009 on Build GRCh38
rs544443610 Chr.19: 49055587-49055587 on Build GRCh38
rs12461505 Chr.19: 49055576-49055576 on Build GRCh38
rs977334370 Chr.19: 49055752-49055752 on Build GRCh38
rs556574621 Chr.19: 49056038-49056038 on Build GRCh38
rs944189314 Chr.19: 49055933-49055933 on Build GRCh38
rs537943009 Chr.19: 49056035-49056035 on Build GRCh38
rs1037274280 Chr.19: 49055699-49055699 on Build GRCh38
rs1035490098 Chr.19: 49056053-49056053 on Build GRCh38
rs550794625 Chr.19: 49055953-49055953 on Build GRCh38
rs190693103 Chr.19: 49055793-49055793 on Build GRCh38
rs386810089 Chr.19: 49055769-49055771 on Build GRCh38
rs919547524 Chr.19: 49055895-49055895 on Build GRCh38
rs533953438 Chr.19: 49055996-49055996 on Build GRCh38
rs971065762 Chr.19: 49055728-49055728 on Build GRCh38
rs3810178 Chr.19: 49055843-49055843 on Build GRCh38
rs759911870 Chr.19: 49055986-49055986 on Build GRCh38
rs972435146 Chr.19: 49055890-49055890 on Build GRCh38
rs147980158 Chr.19: 49055795-49055795 on Build GRCh38
rs143498623 Chr.19: 49055794-49055794 on Build GRCh38
rs376978820 Chr.19: 49055821-49055821 on Build GRCh38
rs962906821 Chr.19: 49055588-49055588 on Build GRCh38
rs935833720 Chr.19: 49055676-49055676 on Build GRCh38
rs1028882464 Chr.19: 49055726-49055726 on Build GRCh38
rs35772372 Chr.19: 49055769-49055769 on Build GRCh38
rs1041170566 Chr.19: 49055944-49055944 on Build GRCh38
rs36125845 Chr.19: 49055627-49055627 on Build GRCh38
rs143462803 Chr.19: 49055832-49055832 on Build GRCh38

View Full Product Details