Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49463956-49464450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH16A1
Entrez Gene ID
126133
Gene Name
aldehyde dehydrogenase 16 family member A1
Gene Aliases
-
UniGene
Hs.719998
Gene Chromosome Location
Chr.19: 49453216-49471050 on Build GRCh38
Transcripts
NM_001145396.1 NM_153329.3 XM_011526441.1 XM_011526442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
272327 c.1365C>T p.H455H Chr.19: 49464450-49464450 on Build GRCh38
3797378 c.1323G>A p.L441L Chr.19: 49464255-49464255 on Build GRCh38
1395248 c.1218C>A p.A406A Chr.19: 49464150-49464150 on Build GRCh38
5777053 c.1195-8C>T p.? Chr.19: 49464119-49464119 on Build GRCh38
6598140 c.1228C>T p.R410C Chr.19: 49464160-49464160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769307435 Chr.19: 49464203-49464203 on Build GRCh38
rs761233154 Chr.19: 49464155-49464155 on Build GRCh38
rs375627359 Chr.19: 49464377-49464377 on Build GRCh38
rs750601987 Chr.19: 49464296-49464296 on Build GRCh38
rs774306767 Chr.19: 49464288-49464288 on Build GRCh38
rs377194423 Chr.19: 49463981-49463981 on Build GRCh38
rs546433908 Chr.19: 49464011-49464011 on Build GRCh38
rs138475609 Chr.19: 49464192-49464192 on Build GRCh38
rs995030247 Chr.19: 49464003-49464003 on Build GRCh38
rs765097109 Chr.19: 49464095-49464095 on Build GRCh38
rs917255833 Chr.19: 49464338-49464338 on Build GRCh38
rs866912510 Chr.19: 49464170-49464170 on Build GRCh38
rs536201235 Chr.19: 49463980-49463980 on Build GRCh38
rs778974292 Chr.19: 49464263-49464263 on Build GRCh38
rs756773398 Chr.19: 49464094-49464094 on Build GRCh38
rs745343121 Chr.19: 49464390-49464390 on Build GRCh38
rs376398701 Chr.19: 49463997-49463997 on Build GRCh38
rs369745354 Chr.19: 49464085-49464085 on Build GRCh38
rs370010620 Chr.19: 49464112-49464112 on Build GRCh38
rs759331360 Chr.19: 49464234-49464234 on Build GRCh38
rs765074520 Chr.19: 49464292-49464292 on Build GRCh38
rs371313328 Chr.19: 49464365-49464365 on Build GRCh38
rs143098791 Chr.19: 49464197-49464197 on Build GRCh38
rs774627148 Chr.19: 49464236-49464237 on Build GRCh38
rs1000386645 Chr.19: 49464345-49464345 on Build GRCh38
rs13346311 Chr.19: 49464240-49464240 on Build GRCh38
rs781545715 Chr.19: 49464311-49464311 on Build GRCh38
rs937917658 Chr.19: 49464326-49464326 on Build GRCh38
rs576632177 Chr.19: 49464145-49464145 on Build GRCh38
rs765506860 Chr.19: 49464294-49464294 on Build GRCh38
rs757316825 Chr.19: 49464179-49464179 on Build GRCh38
rs367630533 Chr.19: 49464280-49464280 on Build GRCh38
rs58745947 Chr.19: 49464171-49464171 on Build GRCh38
rs750214804 Chr.19: 49464341-49464341 on Build GRCh38
rs371652548 Chr.19: 49463978-49463978 on Build GRCh38
rs76274328 Chr.19: 49463995-49463995 on Build GRCh38
rs745923965 Chr.19: 49464189-49464189 on Build GRCh38
rs956742140 Chr.19: 49464080-49464080 on Build GRCh38
rs554471064 Chr.19: 49464327-49464327 on Build GRCh38
rs566399437 Chr.19: 49464342-49464342 on Build GRCh38
rs915347915 Chr.19: 49464102-49464102 on Build GRCh38
rs150186880 Chr.19: 49464205-49464205 on Build GRCh38
rs750416447 Chr.19: 49464070-49464070 on Build GRCh38
rs376237943 Chr.19: 49464245-49464245 on Build GRCh38
rs371448117 Chr.19: 49464281-49464281 on Build GRCh38
rs113529719 Chr.19: 49464015-49464015 on Build GRCh38
rs778054606 Chr.19: 49464117-49464117 on Build GRCh38
rs140640605 Chr.19: 49464144-49464144 on Build GRCh38
rs188710863 Chr.19: 49464160-49464160 on Build GRCh38
rs545337650 Chr.19: 49464334-49464334 on Build GRCh38
rs200435991 Chr.19: 49464164-49464164 on Build GRCh38
rs2293008 Chr.19: 49464034-49464034 on Build GRCh38
rs540018165 Chr.19: 49464344-49464344 on Build GRCh38
rs763313609 Chr.19: 49464417-49464417 on Build GRCh38
rs770986242 Chr.19: 49464123-49464123 on Build GRCh38
rs370604251 Chr.19: 49464261-49464261 on Build GRCh38
rs776285715 Chr.19: 49464152-49464152 on Build GRCh38
rs750307829 Chr.19: 49464100-49464100 on Build GRCh38
rs754754768 Chr.19: 49464163-49464163 on Build GRCh38
rs369977868 Chr.19: 49463984-49463984 on Build GRCh38
rs957495847 Chr.19: 49464228-49464228 on Build GRCh38
rs535858907 Chr.19: 49464318-49464318 on Build GRCh38
rs763939268 Chr.19: 49464405-49464405 on Build GRCh38
rs760371149 Chr.19: 49464293-49464293 on Build GRCh38
rs900116709 Chr.19: 49464353-49464353 on Build GRCh38
rs779005097 Chr.19: 49464183-49464183 on Build GRCh38
rs749386156 Chr.19: 49464118-49464118 on Build GRCh38
rs768925017 Chr.19: 49464275-49464275 on Build GRCh38
rs758577738 Chr.19: 49464297-49464297 on Build GRCh38
rs373759432 Chr.19: 49464376-49464376 on Build GRCh38
rs201780425 Chr.19: 49464111-49464111 on Build GRCh38
rs766363661 Chr.19: 49464214-49464214 on Build GRCh38
rs963718024 Chr.19: 49464059-49464059 on Build GRCh38
rs57656227 Chr.19: 49464017-49464017 on Build GRCh38
rs755547269 Chr.19: 49464308-49464308 on Build GRCh38
rs773590065 Chr.19: 49464412-49464412 on Build GRCh38
rs1055704325 Chr.19: 49464167-49464167 on Build GRCh38
rs558103745 Chr.19: 49464346-49464346 on Build GRCh38
rs1007244316 Chr.19: 49464182-49464182 on Build GRCh38
rs964268965 Chr.19: 49464215-49464215 on Build GRCh38
rs374982664 Chr.19: 49464399-49464399 on Build GRCh38
rs759835501 Chr.19: 49463993-49463993 on Build GRCh38
rs373500828 Chr.19: 49464266-49464266 on Build GRCh38
rs772769557 Chr.19: 49464191-49464191 on Build GRCh38
rs893083778 Chr.19: 49464369-49464369 on Build GRCh38
rs1000334154 Chr.19: 49464204-49464204 on Build GRCh38
rs150960356 Chr.19: 49464139-49464139 on Build GRCh38
rs1034409297 Chr.19: 49464007-49464007 on Build GRCh38
rs537993303 Chr.19: 49464202-49464202 on Build GRCh38
rs757550978 Chr.19: 49464260-49464260 on Build GRCh38
rs754029074 Chr.19: 49464176-49464176 on Build GRCh38
rs751727995 Chr.19: 49464429-49464429 on Build GRCh38
rs746341939 Chr.19: 49464132-49464132 on Build GRCh38
rs747386854 Chr.19: 49464193-49464193 on Build GRCh38
rs1041815131 Chr.19: 49464343-49464343 on Build GRCh38
rs780708156 Chr.19: 49464273-49464273 on Build GRCh38
rs576894102 Chr.19: 49464022-49464022 on Build GRCh38
rs61732836 Chr.19: 49464147-49464147 on Build GRCh38
rs772118363 Chr.19: 49464396-49464396 on Build GRCh38
rs753353889 Chr.19: 49464091-49464091 on Build GRCh38
rs1045288160 Chr.19: 49464131-49464131 on Build GRCh38
rs199807886 Chr.19: 49464227-49464227 on Build GRCh38
rs766273860 Chr.19: 49464161-49464161 on Build GRCh38
rs989495546 Chr.19: 49464238-49464238 on Build GRCh38
rs868635624 Chr.19: 49464172-49464172 on Build GRCh38
rs748810234 Chr.19: 49464284-49464284 on Build GRCh38
rs540098012 Chr.19: 49464285-49464285 on Build GRCh38
rs977361627 Chr.19: 49464067-49464067 on Build GRCh38
rs760358272 Chr.19: 49464082-49464082 on Build GRCh38
rs769020141 Chr.19: 49464205-49464205 on Build GRCh38
rs369105748 Chr.19: 49464403-49464403 on Build GRCh38
rs975049400 Chr.19: 49464064-49464064 on Build GRCh38
rs756370303 Chr.19: 49464115-49464115 on Build GRCh38
rs926565454 Chr.19: 49464321-49464321 on Build GRCh38
rs968070124 Chr.19: 49464026-49464026 on Build GRCh38
rs923969423 Chr.19: 49464078-49464078 on Build GRCh38
rs180723300 Chr.19: 49464274-49464274 on Build GRCh38
rs766516992 Chr.19: 49464298-49464298 on Build GRCh38
rs201033569 Chr.19: 49464154-49464154 on Build GRCh38
rs776178060 Chr.19: 49464198-49464198 on Build GRCh38
rs748605370 Chr.19: 49464366-49464366 on Build GRCh38
rs2293009 Chr.19: 49464423-49464423 on Build GRCh38
rs781655331 Chr.19: 49464278-49464278 on Build GRCh38
rs998719705 Chr.19: 49464359-49464359 on Build GRCh38
rs373326957 Chr.19: 49464092-49464092 on Build GRCh38
rs895877773 Chr.19: 49464013-49464013 on Build GRCh38
rs774728276 Chr.19: 49464124-49464124 on Build GRCh38
rs762446931 Chr.19: 49464209-49464209 on Build GRCh38
rs1033224470 Chr.19: 49464046-49464046 on Build GRCh38
rs888777884 Chr.19: 49464335-49464335 on Build GRCh38
rs370793991 Chr.19: 49464386-49464386 on Build GRCh38

View Full Product Details