Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58347231-58347772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A1BG
Entrez Gene ID
1
Gene Name
alpha-1-B glycoprotein
Gene Aliases
A1B, ABG, GAB, HYST2477
UniGene
Hs.529161
Gene Chromosome Location
Chr.19: 58346806-58353499 on Build GRCh38
Transcripts
NM_130786.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2153142 c.1397G>A p.G466D Chr.19: 58347436-58347436 on Build GRCh38
6589076 c.1351G>A p.A451T Chr.19: 58347482-58347482 on Build GRCh38
6390507 c.1219G>A p.A407T Chr.19: 58347614-58347614 on Build GRCh38
6589078 c.1327G>A p.V443M Chr.19: 58347506-58347506 on Build GRCh38
2974300 c.1456G>A p.D486N Chr.19: 58347377-58347377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769339400 Chr.19: 58347374-58347374 on Build GRCh38
rs780797889 Chr.19: 58347458-58347458 on Build GRCh38
rs67397428 Chr.19: 58347697-58347698 on Build GRCh38
rs936326240 Chr.19: 58347666-58347666 on Build GRCh38
rs889511505 Chr.19: 58347659-58347659 on Build GRCh38
rs770249611 Chr.19: 58347456-58347458 on Build GRCh38
rs767646234 Chr.19: 58347321-58347321 on Build GRCh38
rs879884404 Chr.19: 58347701-58347701 on Build GRCh38
rs746134225 Chr.19: 58347689-58347712 on Build GRCh38
rs781255144 Chr.19: 58347642-58347642 on Build GRCh38
rs773174332 Chr.19: 58347686-58347686 on Build GRCh38
rs777501141 Chr.19: 58347335-58347335 on Build GRCh38
rs754557762 Chr.19: 58347533-58347533 on Build GRCh38
rs1014895724 Chr.19: 58347545-58347545 on Build GRCh38
rs373945949 Chr.19: 58347705-58347705 on Build GRCh38
rs990951519 Chr.19: 58347289-58347289 on Build GRCh38
rs755255061 Chr.19: 58347494-58347494 on Build GRCh38
rs776948818 Chr.19: 58347557-58347557 on Build GRCh38
rs564038740 Chr.19: 58347599-58347599 on Build GRCh38
rs745575122 Chr.19: 58347464-58347464 on Build GRCh38
rs996999776 Chr.19: 58347436-58347436 on Build GRCh38
rs890854697 Chr.19: 58347401-58347401 on Build GRCh38
rs771083874 Chr.19: 58347552-58347552 on Build GRCh38
rs980009543 Chr.19: 58347298-58347298 on Build GRCh38
rs943758483 Chr.19: 58347622-58347622 on Build GRCh38
rs772616148 Chr.19: 58347495-58347495 on Build GRCh38
rs1265959 Chr.19: 58347700-58347700 on Build GRCh38
rs150726160 Chr.19: 58347477-58347477 on Build GRCh38
rs767883784 Chr.19: 58347421-58347421 on Build GRCh38
rs772563296 Chr.19: 58347546-58347546 on Build GRCh38
rs547975899 Chr.19: 58347728-58347733 on Build GRCh38
rs757030081 Chr.19: 58347516-58347516 on Build GRCh38
rs762128963 Chr.19: 58347318-58347318 on Build GRCh38
rs758989494 Chr.19: 58347612-58347612 on Build GRCh38
rs752678090 Chr.19: 58347429-58347429 on Build GRCh38
rs771372524 Chr.19: 58347635-58347635 on Build GRCh38
rs747071974 Chr.19: 58347677-58347678 on Build GRCh38
rs759508483 Chr.19: 58347575-58347575 on Build GRCh38
rs770588777 Chr.19: 58347347-58347347 on Build GRCh38
rs778490538 Chr.19: 58347677-58347700 on Build GRCh38
rs758940067 Chr.19: 58347677-58347712 on Build GRCh38
rs773610628 Chr.19: 58347423-58347423 on Build GRCh38
rs769694425 Chr.19: 58347655-58347655 on Build GRCh38
rs776591698 Chr.19: 58347507-58347507 on Build GRCh38
rs199949567 Chr.19: 58347518-58347518 on Build GRCh38
rs564533444 Chr.19: 58347467-58347467 on Build GRCh38
rs995585388 Chr.19: 58347488-58347488 on Build GRCh38
rs970090733 Chr.19: 58347703-58347703 on Build GRCh38
rs748725174 Chr.19: 58347538-58347538 on Build GRCh38
rs549593190 Chr.19: 58347667-58347667 on Build GRCh38
rs768783457 Chr.19: 58347276-58347276 on Build GRCh38
rs113490434 Chr.19: 58347689-58347689 on Build GRCh38
rs778633926 Chr.19: 58347608-58347608 on Build GRCh38
rs923640653 Chr.19: 58347665-58347665 on Build GRCh38
rs765917918 Chr.19: 58347597-58347597 on Build GRCh38
rs573823581 Chr.19: 58347290-58347290 on Build GRCh38
rs371173507 Chr.19: 58347537-58347537 on Build GRCh38
rs186445851 Chr.19: 58347390-58347390 on Build GRCh38
rs529546189 Chr.19: 58347601-58347601 on Build GRCh38
rs62116376 Chr.19: 58347712-58347712 on Build GRCh38
rs902773927 Chr.19: 58347462-58347462 on Build GRCh38
rs751545034 Chr.19: 58347514-58347514 on Build GRCh38
rs139020123 Chr.19: 58347510-58347510 on Build GRCh38
rs960134664 Chr.19: 58347710-58347710 on Build GRCh38
rs763351169 Chr.19: 58347302-58347302 on Build GRCh38
rs932469116 Chr.19: 58347380-58347380 on Build GRCh38
rs200130951 Chr.19: 58347437-58347437 on Build GRCh38
rs1001775652 Chr.19: 58347706-58347706 on Build GRCh38
rs767240825 Chr.19: 58347592-58347592 on Build GRCh38
rs765936482 Chr.19: 58347414-58347415 on Build GRCh38
rs773555659 Chr.19: 58347497-58347497 on Build GRCh38
rs751383546 Chr.19: 58347440-58347440 on Build GRCh38
rs768289377 Chr.19: 58347689-58347700 on Build GRCh38
rs144085420 Chr.19: 58347689-58347690 on Build GRCh38
rs1027037622 Chr.19: 58347693-58347693 on Build GRCh38
rs763435454 Chr.19: 58347403-58347403 on Build GRCh38
rs919809949 Chr.19: 58347371-58347371 on Build GRCh38
rs796832423 Chr.19: 58347252-58347252 on Build GRCh38
rs761742979 Chr.19: 58347587-58347587 on Build GRCh38
rs753191969 Chr.19: 58347609-58347609 on Build GRCh38
rs995451348 Chr.19: 58347681-58347681 on Build GRCh38
rs754928401 Chr.19: 58347325-58347325 on Build GRCh38
rs865796493 Chr.19: 58347261-58347261 on Build GRCh38
rs755735308 Chr.19: 58347522-58347522 on Build GRCh38
rs760816395 Chr.19: 58347424-58347424 on Build GRCh38
rs1265957 Chr.19: 58347310-58347310 on Build GRCh38
rs750009878 Chr.19: 58347520-58347520 on Build GRCh38
rs748780306 Chr.19: 58347527-58347527 on Build GRCh38
rs774591364 Chr.19: 58347416-58347416 on Build GRCh38
rs774738068 Chr.19: 58347487-58347487 on Build GRCh38
rs772167924 Chr.19: 58347392-58347392 on Build GRCh38
rs747367917 Chr.19: 58347629-58347629 on Build GRCh38
rs760335154 Chr.19: 58347594-58347594 on Build GRCh38
rs62116375 Chr.19: 58347698-58347698 on Build GRCh38
rs908471419 Chr.19: 58347324-58347324 on Build GRCh38
rs572621623 Chr.19: 58347360-58347360 on Build GRCh38
rs750239743 Chr.19: 58347593-58347593 on Build GRCh38
rs749744578 Chr.19: 58347479-58347479 on Build GRCh38
rs970741078 Chr.19: 58347291-58347291 on Build GRCh38
rs765654309 Chr.19: 58347621-58347627 on Build GRCh38
rs1012810499 Chr.19: 58347493-58347493 on Build GRCh38
rs773719039 Chr.19: 58347548-58347548 on Build GRCh38
rs528615958 Chr.19: 58347679-58347679 on Build GRCh38
rs912327348 Chr.19: 58347614-58347614 on Build GRCh38
rs761047781 Chr.19: 58347506-58347506 on Build GRCh38
rs1027006415 Chr.19: 58347489-58347489 on Build GRCh38
rs371305168 Chr.19: 58347344-58347344 on Build GRCh38
rs1001109916 Chr.19: 58347463-58347463 on Build GRCh38
rs201773235 Chr.19: 58347362-58347362 on Build GRCh38
rs748492537 Chr.19: 58347490-58347490 on Build GRCh38
rs778239053 Chr.19: 58347616-58347616 on Build GRCh38
rs375150138 Chr.19: 58347576-58347576 on Build GRCh38
rs747174184 Chr.19: 58347549-58347549 on Build GRCh38
rs763895886 Chr.19: 58347439-58347439 on Build GRCh38
rs767365634 Chr.19: 58347519-58347519 on Build GRCh38
rs768998336 Chr.19: 58347412-58347412 on Build GRCh38
rs939886374 Chr.19: 58347343-58347343 on Build GRCh38
rs1042821006 Chr.19: 58347432-58347432 on Build GRCh38
rs373959127 Chr.19: 58347414-58347414 on Build GRCh38
rs751365497 Chr.19: 58347422-58347422 on Build GRCh38
rs766429748 Chr.19: 58347426-58347426 on Build GRCh38
rs921667584 Chr.19: 58347632-58347632 on Build GRCh38
rs757079396 Chr.19: 58347455-58347455 on Build GRCh38
rs12978513 Chr.19: 58347569-58347569 on Build GRCh38
rs751801695 Chr.19: 58347317-58347317 on Build GRCh38
rs749608112 Chr.19: 58347598-58347598 on Build GRCh38
rs1049899055 Chr.19: 58347633-58347633 on Build GRCh38
rs376096264 Chr.19: 58347717-58347717 on Build GRCh38
rs778963034 Chr.19: 58347326-58347326 on Build GRCh38
rs370008583 Chr.19: 58347332-58347332 on Build GRCh38
rs868246140 Chr.19: 58347574-58347574 on Build GRCh38
rs574597204 Chr.19: 58347448-58347448 on Build GRCh38
rs775559380 Chr.19: 58347657-58347657 on Build GRCh38
rs762902651 Chr.19: 58347509-58347509 on Build GRCh38
rs769064384 Chr.19: 58347484-58347484 on Build GRCh38
rs753157383 Chr.19: 58347667-58347678 on Build GRCh38
rs141343506 Chr.19: 58347387-58347387 on Build GRCh38
rs559517090 Chr.19: 58347675-58347675 on Build GRCh38
rs746164945 Chr.19: 58347652-58347652 on Build GRCh38

View Full Product Details