Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9297214-9297719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP2
Entrez Gene ID
8853
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1
UniGene
Hs.555902
Gene Chromosome Location
Chr.2: 9206765-9405683 on Build GRCh38
Transcripts
NM_001135191.1 NM_003887.2 XM_006711898.1 XM_006711901.3 XM_006711902.1 XM_011510403.2 XM_011510404.2 XM_011510405.2 XM_011510406.2 XM_011510407.2 XM_011510408.2 XM_011510409.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6261138 c.291G>T p.A97A Chr.2: 9297391-9297391 on Build GRCh38
1409933 c.291G>A p.A97A Chr.2: 9297391-9297391 on Build GRCh38
1752723 c.306A>C p.S102S Chr.2: 9297406-9297406 on Build GRCh38
4663590 c.222G>A p.Q74Q Chr.2: 9297322-9297322 on Build GRCh38
4096242 c.249C>T p.G83G Chr.2: 9297349-9297349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3811600 Chr.2: 9297284-9297284 on Build GRCh38
rs369806962 Chr.2: 9297349-9297349 on Build GRCh38
rs938127522 Chr.2: 9297617-9297617 on Build GRCh38
rs1016726141 Chr.2: 9297460-9297460 on Build GRCh38
rs889594045 Chr.2: 9297429-9297429 on Build GRCh38
rs377202646 Chr.2: 9297391-9297391 on Build GRCh38
rs375793564 Chr.2: 9297473-9297473 on Build GRCh38
rs772646789 Chr.2: 9297361-9297361 on Build GRCh38
rs762007216 Chr.2: 9297278-9297278 on Build GRCh38
rs764340667 Chr.2: 9297269-9297269 on Build GRCh38
rs767402695 Chr.2: 9297470-9297470 on Build GRCh38
rs560040830 Chr.2: 9297304-9297304 on Build GRCh38
rs369207362 Chr.2: 9297285-9297285 on Build GRCh38
rs982331601 Chr.2: 9297535-9297535 on Build GRCh38
rs770387424 Chr.2: 9297568-9297568 on Build GRCh38
rs775988603 Chr.2: 9297574-9297574 on Build GRCh38
rs552854910 Chr.2: 9297676-9297676 on Build GRCh38
rs548555308 Chr.2: 9297357-9297357 on Build GRCh38
rs568509831 Chr.2: 9297390-9297390 on Build GRCh38
rs769371542 Chr.2: 9297454-9297455 on Build GRCh38
rs368026338 Chr.2: 9297252-9297252 on Build GRCh38
rs777425864 Chr.2: 9297347-9297347 on Build GRCh38
rs989280901 Chr.2: 9297621-9297621 on Build GRCh38
rs914972361 Chr.2: 9297666-9297666 on Build GRCh38
rs759476220 Chr.2: 9297457-9297457 on Build GRCh38
rs144657542 Chr.2: 9297358-9297358 on Build GRCh38
rs775978016 Chr.2: 9297258-9297258 on Build GRCh38
rs369109260 Chr.2: 9297474-9297474 on Build GRCh38
rs570333705 Chr.2: 9297506-9297506 on Build GRCh38
rs751553633 Chr.2: 9297281-9297281 on Build GRCh38
rs761940253 Chr.2: 9297403-9297403 on Build GRCh38
rs926796371 Chr.2: 9297605-9297605 on Build GRCh38
rs373522266 Chr.2: 9297387-9297387 on Build GRCh38
rs954573022 Chr.2: 9297592-9297592 on Build GRCh38
rs900151570 Chr.2: 9297516-9297516 on Build GRCh38
rs752511116 Chr.2: 9297472-9297472 on Build GRCh38
rs142115918 Chr.2: 9297443-9297443 on Build GRCh38
rs773221107 Chr.2: 9297453-9297453 on Build GRCh38
rs957906804 Chr.2: 9297339-9297339 on Build GRCh38
rs539199999 Chr.2: 9297515-9297515 on Build GRCh38
rs746353773 Chr.2: 9297492-9297492 on Build GRCh38
rs776583391 Chr.2: 9297277-9297277 on Build GRCh38
rs755767129 Chr.2: 9297327-9297327 on Build GRCh38
rs1037353256 Chr.2: 9297510-9297510 on Build GRCh38
rs199504138 Chr.2: 9297350-9297350 on Build GRCh38
rs572852070 Chr.2: 9297670-9297670 on Build GRCh38
rs17855821 Chr.2: 9297356-9297356 on Build GRCh38
rs756843927 Chr.2: 9297497-9297497 on Build GRCh38
rs1042436337 Chr.2: 9297686-9297686 on Build GRCh38
rs550762710 Chr.2: 9297499-9297499 on Build GRCh38
rs761004524 Chr.2: 9297261-9297261 on Build GRCh38
rs544895078 Chr.2: 9297424-9297424 on Build GRCh38
rs879054983 Chr.2: 9297509-9297509 on Build GRCh38
rs372237689 Chr.2: 9297280-9297280 on Build GRCh38

View Full Product Details