Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9407359-9407866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB1BP1
Entrez Gene ID
9270
Gene Name
integrin subunit beta 1 binding protein 1
Gene Aliases
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B
UniGene
Hs.467662
Gene Chromosome Location
Chr.2: 9405681-9423569 on Build GRCh38
Transcripts
NM_001319066.1 NM_001319067.1 NM_001319068.1 NM_001319069.1 NM_001319070.1 NM_004763.4 NM_022334.4 NR_134951.1 NR_134952.1 NR_134953.1 NR_134954.1 NR_134955.1 NR_134956.1 NR_134957.1 XM_005246183.4 XM_005246184.4 XM_005246185.4 XM_005246189.4 XM_006711903.3 XM_011510416.2 XM_017005267.1 XM_017005268.1 XM_017005269.1 XM_017005270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
359397 c.514T>A p.Y172N Chr.2: 9407466-9407466 on Build GRCh38
6009976 c.428G>T p.C143F Chr.2: 9407552-9407552 on Build GRCh38
5481772 c.498A>G p.E166E Chr.2: 9407482-9407482 on Build GRCh38
6688173 c.515A>G p.Y172C Chr.2: 9407465-9407465 on Build GRCh38
4400216 c.518A>T p.Q173L Chr.2: 9407462-9407462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902816037 Chr.2: 9407763-9407763 on Build GRCh38
rs556404898 Chr.2: 9407637-9407637 on Build GRCh38
rs865830253 Chr.2: 9407500-9407501 on Build GRCh38
rs200349700 Chr.2: 9407548-9407548 on Build GRCh38
rs562771759 Chr.2: 9407462-9407462 on Build GRCh38
rs902868989 Chr.2: 9407526-9407526 on Build GRCh38
rs778547540 Chr.2: 9407622-9407623 on Build GRCh38
rs759547410 Chr.2: 9407409-9407409 on Build GRCh38
rs1018424885 Chr.2: 9407560-9407560 on Build GRCh38
rs1016992206 Chr.2: 9407584-9407584 on Build GRCh38
rs199860614 Chr.2: 9407439-9407439 on Build GRCh38
rs759545283 Chr.2: 9407731-9407731 on Build GRCh38
rs372189624 Chr.2: 9407519-9407519 on Build GRCh38
rs762673678 Chr.2: 9407504-9407504 on Build GRCh38
rs888856934 Chr.2: 9407786-9407786 on Build GRCh38
rs932996675 Chr.2: 9407776-9407776 on Build GRCh38
rs991308589 Chr.2: 9407712-9407712 on Build GRCh38
rs924497566 Chr.2: 9407822-9407822 on Build GRCh38
rs371225003 Chr.2: 9407438-9407438 on Build GRCh38
rs374426346 Chr.2: 9407619-9407619 on Build GRCh38
rs188763404 Chr.2: 9407608-9407608 on Build GRCh38
rs765247474 Chr.2: 9407417-9407417 on Build GRCh38
rs767633981 Chr.2: 9407411-9407411 on Build GRCh38
rs529557159 Chr.2: 9407414-9407414 on Build GRCh38
rs907338178 Chr.2: 9407546-9407546 on Build GRCh38
rs367974883 Chr.2: 9407470-9407470 on Build GRCh38
rs758212972 Chr.2: 9407427-9407427 on Build GRCh38
rs746089139 Chr.2: 9407634-9407634 on Build GRCh38
rs531717039 Chr.2: 9407541-9407541 on Build GRCh38
rs377453919 Chr.2: 9407622-9407622 on Build GRCh38
rs746058136 Chr.2: 9407464-9407464 on Build GRCh38
rs867815322 Chr.2: 9407526-9407527 on Build GRCh38
rs975291819 Chr.2: 9407612-9407612 on Build GRCh38
rs201259238 Chr.2: 9407542-9407542 on Build GRCh38
rs751167293 Chr.2: 9407431-9407431 on Build GRCh38
rs771268281 Chr.2: 9407461-9407461 on Build GRCh38
rs373071157 Chr.2: 9407621-9407621 on Build GRCh38
rs776737654 Chr.2: 9407412-9407412 on Build GRCh38
rs947265247 Chr.2: 9407746-9407746 on Build GRCh38
rs547624427 Chr.2: 9407605-9407605 on Build GRCh38
rs778246050 Chr.2: 9407449-9407449 on Build GRCh38
rs781674266 Chr.2: 9407390-9407390 on Build GRCh38
rs928733905 Chr.2: 9407696-9407696 on Build GRCh38
rs750226942 Chr.2: 9407418-9407418 on Build GRCh38
rs962903671 Chr.2: 9407607-9407607 on Build GRCh38
rs756722504 Chr.2: 9407611-9407612 on Build GRCh38
rs751242658 Chr.2: 9407755-9407759 on Build GRCh38
rs780422331 Chr.2: 9407577-9407577 on Build GRCh38
rs747147529 Chr.2: 9407578-9407578 on Build GRCh38
rs765542285 Chr.2: 9407757-9407757 on Build GRCh38
rs951420490 Chr.2: 9407678-9407678 on Build GRCh38
rs773028167 Chr.2: 9407609-9407609 on Build GRCh38
rs201847721 Chr.2: 9407407-9407407 on Build GRCh38
rs771124362 Chr.2: 9407406-9407406 on Build GRCh38
rs35336030 Chr.2: 9407815-9407815 on Build GRCh38
rs745355182 Chr.2: 9407641-9407643 on Build GRCh38
rs373174809 Chr.2: 9407547-9407547 on Build GRCh38
rs374769191 Chr.2: 9407469-9407469 on Build GRCh38
rs933803735 Chr.2: 9407765-9407765 on Build GRCh38
rs770177654 Chr.2: 9407396-9407396 on Build GRCh38
rs773518118 Chr.2: 9407398-9407398 on Build GRCh38
rs370443483 Chr.2: 9407630-9407630 on Build GRCh38
rs760094003 Chr.2: 9407653-9407654 on Build GRCh38
rs756652542 Chr.2: 9407446-9407446 on Build GRCh38
rs758599279 Chr.2: 9407681-9407681 on Build GRCh38
rs527741415 Chr.2: 9407561-9407561 on Build GRCh38
rs772465614 Chr.2: 9407576-9407576 on Build GRCh38
rs764366013 Chr.2: 9407783-9407783 on Build GRCh38
rs17850889 Chr.2: 9407531-9407531 on Build GRCh38
rs375404934 Chr.2: 9407405-9407405 on Build GRCh38
rs746211130 Chr.2: 9407562-9407562 on Build GRCh38
rs937432129 Chr.2: 9407697-9407697 on Build GRCh38
rs749672359 Chr.2: 9407459-9407459 on Build GRCh38
rs569589067 Chr.2: 9407675-9407675 on Build GRCh38
rs760808268 Chr.2: 9407477-9407477 on Build GRCh38
rs769922581 Chr.2: 9407487-9407487 on Build GRCh38
rs914422202 Chr.2: 9407743-9407743 on Build GRCh38
rs982578961 Chr.2: 9407558-9407558 on Build GRCh38
rs760183332 Chr.2: 9407638-9407638 on Build GRCh38
rs749183642 Chr.2: 9407527-9407528 on Build GRCh38
rs542886635 Chr.2: 9407429-9407429 on Build GRCh38
rs538623184 Chr.2: 9407744-9407744 on Build GRCh38
rs749631664 Chr.2: 9407399-9407399 on Build GRCh38
rs748587660 Chr.2: 9407394-9407394 on Build GRCh38
rs751202795 Chr.2: 9407530-9407530 on Build GRCh38
rs773191905 Chr.2: 9407500-9407500 on Build GRCh38
rs558218497 Chr.2: 9407761-9407761 on Build GRCh38
rs1055677125 Chr.2: 9407458-9407458 on Build GRCh38
rs113734942 Chr.2: 9407606-9407606 on Build GRCh38

View Full Product Details