Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10408371-10408883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPCAL1
Entrez Gene ID
3241
Gene Name
hippocalcin like 1
Gene Aliases
BDR1, HLP2, VILIP-3
UniGene
Hs.580427
Gene Chromosome Location
Chr.2: 10302124-10427617 on Build GRCh38
Transcripts
NM_001258357.1 NM_001258358.1 NM_001258359.1 NM_002149.3 NM_134421.2 XM_005246160.1 XM_005246161.1 XM_005246162.1 XM_005246163.1 XM_011510346.2 XM_011510347.1 XM_011510348.1 XM_011510349.1 XM_017003950.1 XM_017003951.1 XM_017003952.1 XM_017003953.1

SNP Detail

SNP ID Chromosome Location
rs971429461 Chr.2: 10408491-10408491 on Build GRCh38
rs1039447547 Chr.2: 10408404-10408404 on Build GRCh38
rs1039882614 Chr.2: 10408586-10408586 on Build GRCh38
rs902323663 Chr.2: 10408715-10408715 on Build GRCh38
rs1042640075 Chr.2: 10408702-10408702 on Build GRCh38
rs1024644513 Chr.2: 10408855-10408855 on Build GRCh38
rs759595255 Chr.2: 10408753-10408753 on Build GRCh38
rs956292337 Chr.2: 10408527-10408527 on Build GRCh38
rs565098253 Chr.2: 10408842-10408842 on Build GRCh38
rs966894572 Chr.2: 10408448-10408448 on Build GRCh38
rs187143845 Chr.2: 10408498-10408498 on Build GRCh38
rs569213704 Chr.2: 10408470-10408470 on Build GRCh38
rs1023028250 Chr.2: 10408488-10408488 on Build GRCh38
rs146071973 Chr.2: 10408620-10408620 on Build GRCh38
rs7603265 Chr.2: 10408860-10408860 on Build GRCh38
rs933700394 Chr.2: 10408609-10408609 on Build GRCh38
rs762679720 Chr.2: 10408579-10408579 on Build GRCh38
rs568829789 Chr.2: 10408675-10408675 on Build GRCh38
rs552529244 Chr.2: 10408542-10408542 on Build GRCh38
rs1011921631 Chr.2: 10408839-10408839 on Build GRCh38
rs1032106575 Chr.2: 10408798-10408798 on Build GRCh38
rs931417470 Chr.2: 10408528-10408528 on Build GRCh38
rs115487491 Chr.2: 10408621-10408621 on Build GRCh38
rs1032033348 Chr.2: 10408462-10408462 on Build GRCh38
rs998002450 Chr.2: 10408425-10408425 on Build GRCh38
rs573021091 Chr.2: 10408762-10408762 on Build GRCh38
rs1013445439 Chr.2: 10408487-10408487 on Build GRCh38
rs918179440 Chr.2: 10408482-10408482 on Build GRCh38
rs959045276 Chr.2: 10408479-10408487 on Build GRCh38
rs876614 Chr.2: 10408451-10408451 on Build GRCh38
rs1004173751 Chr.2: 10408629-10408629 on Build GRCh38
rs767690964 Chr.2: 10408838-10408838 on Build GRCh38
rs924747157 Chr.2: 10408500-10408500 on Build GRCh38
rs914924847 Chr.2: 10408546-10408546 on Build GRCh38
rs550932107 Chr.2: 10408399-10408399 on Build GRCh38
rs1029479558 Chr.2: 10408430-10408430 on Build GRCh38
rs1020781956 Chr.2: 10408415-10408415 on Build GRCh38
rs540399362 Chr.2: 10408774-10408774 on Build GRCh38
rs971420635 Chr.2: 10408856-10408856 on Build GRCh38
rs1017354637 Chr.2: 10408477-10408477 on Build GRCh38
rs876616 Chr.2: 10408401-10408401 on Build GRCh38
rs80339500 Chr.2: 10408481-10408481 on Build GRCh38
rs901805637 Chr.2: 10408700-10408700 on Build GRCh38
rs876615 Chr.2: 10408407-10408407 on Build GRCh38
rs554836079 Chr.2: 10408596-10408596 on Build GRCh38
rs3771130 Chr.2: 10408700-10408700 on Build GRCh38
rs946390570 Chr.2: 10408557-10408557 on Build GRCh38
rs971462015 Chr.2: 10408720-10408720 on Build GRCh38
rs115769243 Chr.2: 10408745-10408745 on Build GRCh38
rs1038284683 Chr.2: 10408664-10408664 on Build GRCh38
rs1031840278 Chr.2: 10408769-10408769 on Build GRCh38
rs956199169 Chr.2: 10408837-10408837 on Build GRCh38
rs149109759 Chr.2: 10408492-10408492 on Build GRCh38
rs761524972 Chr.2: 10408434-10408434 on Build GRCh38
rs574541779 Chr.2: 10408684-10408684 on Build GRCh38
rs571102161 Chr.2: 10408583-10408583 on Build GRCh38

View Full Product Details