Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24821807-24822304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY3
Entrez Gene ID
109
Gene Name
adenylate cyclase 3
Gene Aliases
AC-III, AC3
UniGene
Hs.467898
Gene Chromosome Location
Chr.2: 24819169-24920017 on Build GRCh38
Transcripts
NM_001320613.1 NM_004036.4 XM_006711925.1 XM_011532489.1 XM_011532490.1 XM_011532491.1 XM_011532492.2 XM_011532493.1 XM_011532494.1 XM_011532496.1 XM_017003186.1 XM_017003187.1 XM_017003188.1 XM_017003189.1 XM_017003190.1 XM_017003191.1 XM_017003192.1 XM_017003193.1 NM_001199803.2 NM_001322101.1 NM_024322.3 NR_136182.1 NR_136183.1 NR_136184.1

Gene Symbol
CENPO
Entrez Gene ID
79172
Gene Name
centromere protein O
Gene Aliases
CENP-O, ICEN-36, MCM21R
UniGene
Hs.731569
Gene Chromosome Location
Chr.2: 24793115-24822376 on Build GRCh38
Transcripts
NM_001320613.1 NM_004036.4 XM_006711925.1 XM_011532489.1 XM_011532490.1 XM_011532491.1 XM_011532492.2 XM_011532493.1 XM_011532494.1 XM_011532496.1 XM_017003186.1 XM_017003187.1 XM_017003188.1 XM_017003189.1 XM_017003190.1 XM_017003191.1 XM_017003192.1 XM_017003193.1 NM_001199803.2 NM_001322101.1 NM_024322.3 NR_136182.1 NR_136183.1 NR_136184.1

SNP Detail

SNP ID Chromosome Location
rs927352056 Chr.2: 24821907-24821907 on Build GRCh38
rs1057041357 Chr.2: 24821915-24821915 on Build GRCh38
rs537951258 Chr.2: 24822210-24822210 on Build GRCh38
rs879791048 Chr.2: 24821885-24821886 on Build GRCh38
rs764404545 Chr.2: 24822079-24822079 on Build GRCh38
rs924315125 Chr.2: 24822215-24822215 on Build GRCh38
rs989990008 Chr.2: 24822217-24822217 on Build GRCh38
rs745669836 Chr.2: 24822271-24822271 on Build GRCh38
rs753178115 Chr.2: 24822044-24822044 on Build GRCh38
rs141875727 Chr.2: 24821910-24821910 on Build GRCh38
rs992821754 Chr.2: 24822174-24822174 on Build GRCh38
rs71325758 Chr.2: 24821885-24821886 on Build GRCh38
rs1050203638 Chr.2: 24822038-24822038 on Build GRCh38
rs763469786 Chr.2: 24822064-24822064 on Build GRCh38
rs1050582479 Chr.2: 24822239-24822239 on Build GRCh38
rs184372866 Chr.2: 24822015-24822015 on Build GRCh38
rs930824419 Chr.2: 24822221-24822221 on Build GRCh38
rs1037467344 Chr.2: 24822009-24822009 on Build GRCh38
rs991216452 Chr.2: 24821837-24821837 on Build GRCh38
rs932477603 Chr.2: 24822238-24822238 on Build GRCh38
rs753418735 Chr.2: 24822211-24822211 on Build GRCh38
rs567716165 Chr.2: 24821987-24821987 on Build GRCh38
rs917260821 Chr.2: 24822196-24822196 on Build GRCh38
rs1036629524 Chr.2: 24822216-24822216 on Build GRCh38
rs967917777 Chr.2: 24822094-24822094 on Build GRCh38
rs927405744 Chr.2: 24822158-24822158 on Build GRCh38
rs915085256 Chr.2: 24821847-24821847 on Build GRCh38
rs147123606 Chr.2: 24822173-24822173 on Build GRCh38
rs566613587 Chr.2: 24822056-24822056 on Build GRCh38
rs920832080 Chr.2: 24822220-24822220 on Build GRCh38
rs1058064 Chr.2: 24821886-24821886 on Build GRCh38
rs10174476 Chr.2: 24821997-24821997 on Build GRCh38
rs921193195 Chr.2: 24822232-24822232 on Build GRCh38
rs374613429 Chr.2: 24821919-24821919 on Build GRCh38
rs980559262 Chr.2: 24822153-24822153 on Build GRCh38
rs3198667 Chr.2: 24821894-24821894 on Build GRCh38
rs971060659 Chr.2: 24821867-24821867 on Build GRCh38
rs1051392419 Chr.2: 24822257-24822258 on Build GRCh38
rs576205416 Chr.2: 24822006-24822006 on Build GRCh38
rs916996709 Chr.2: 24821852-24821852 on Build GRCh38
rs555687602 Chr.2: 24822193-24822193 on Build GRCh38
rs968244539 Chr.2: 24821851-24821851 on Build GRCh38
rs1051158743 Chr.2: 24821989-24821989 on Build GRCh38
rs765294214 Chr.2: 24821996-24821996 on Build GRCh38
rs76078428 Chr.2: 24821871-24821871 on Build GRCh38
rs537821968 Chr.2: 24822257-24822257 on Build GRCh38
rs778868363 Chr.2: 24822222-24822222 on Build GRCh38
rs912632850 Chr.2: 24822258-24822258 on Build GRCh38
rs1018605986 Chr.2: 24822072-24822072 on Build GRCh38
rs898739875 Chr.2: 24822012-24822012 on Build GRCh38
rs3198665 Chr.2: 24821885-24821885 on Build GRCh38
rs921055879 Chr.2: 24821908-24821908 on Build GRCh38
rs754473225 Chr.2: 24822263-24822263 on Build GRCh38
rs956538833 Chr.2: 24822076-24822076 on Build GRCh38
rs958779338 Chr.2: 24822166-24822166 on Build GRCh38
rs755822087 Chr.2: 24822097-24822097 on Build GRCh38
rs17799872 Chr.2: 24822088-24822088 on Build GRCh38

View Full Product Details