Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24822422-24822922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY3
Entrez Gene ID
109
Gene Name
adenylate cyclase 3
Gene Aliases
AC-III, AC3
UniGene
Hs.467898
Gene Chromosome Location
Chr.2: 24819169-24920017 on Build GRCh38
Transcripts
NM_001320613.1 NM_004036.4 XM_006711925.1 XM_011532489.1 XM_011532490.1 XM_011532491.1 XM_011532492.2 XM_011532493.1 XM_011532494.1 XM_011532496.1 XM_017003186.1 XM_017003187.1 XM_017003188.1 XM_017003189.1 XM_017003190.1 XM_017003191.1 XM_017003192.1 XM_017003193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354535 c.3000C>A p.N1000K Chr.2: 24822514-24822514 on Build GRCh38
2911542 c.2940G>A p.T980T Chr.2: 24822574-24822574 on Build GRCh38
720777 c.2951C>T p.A984V Chr.2: 24822563-24822563 on Build GRCh38
1326282 c.2927C>T p.T976I Chr.2: 24822587-24822587 on Build GRCh38
1019188 c.2884-2A>C p.? Chr.2: 24822632-24822632 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750597549 Chr.2: 24822493-24822493 on Build GRCh38
rs773402151 Chr.2: 24822599-24822599 on Build GRCh38
rs937650884 Chr.2: 24822882-24822882 on Build GRCh38
rs142423059 Chr.2: 24822525-24822525 on Build GRCh38
rs373993570 Chr.2: 24822473-24822473 on Build GRCh38
rs139344556 Chr.2: 24822547-24822547 on Build GRCh38
rs767039358 Chr.2: 24822501-24822501 on Build GRCh38
rs779884635 Chr.2: 24822655-24822655 on Build GRCh38
rs949417344 Chr.2: 24822870-24822870 on Build GRCh38
rs542854249 Chr.2: 24822458-24822458 on Build GRCh38
rs745831583 Chr.2: 24822649-24822649 on Build GRCh38
rs758333474 Chr.2: 24822651-24822651 on Build GRCh38
rs753074005 Chr.2: 24822627-24822627 on Build GRCh38
rs768279980 Chr.2: 24822485-24822486 on Build GRCh38
rs543653069 Chr.2: 24822846-24822846 on Build GRCh38
rs972581711 Chr.2: 24822783-24822783 on Build GRCh38
rs931280783 Chr.2: 24822508-24822508 on Build GRCh38
rs747658762 Chr.2: 24822871-24822871 on Build GRCh38
rs75780875 Chr.2: 24822603-24822603 on Build GRCh38
rs767825755 Chr.2: 24822878-24822878 on Build GRCh38
rs753316883 Chr.2: 24822635-24822635 on Build GRCh38
rs199532565 Chr.2: 24822449-24822449 on Build GRCh38
rs986398083 Chr.2: 24822749-24822749 on Build GRCh38
rs765412411 Chr.2: 24822488-24822488 on Build GRCh38
rs777264230 Chr.2: 24822532-24822532 on Build GRCh38
rs759757473 Chr.2: 24822609-24822609 on Build GRCh38
rs919992695 Chr.2: 24822823-24822823 on Build GRCh38
rs756793005 Chr.2: 24822637-24822637 on Build GRCh38
rs549230016 Chr.2: 24822751-24822751 on Build GRCh38
rs191841661 Chr.2: 24822768-24822768 on Build GRCh38
rs766949371 Chr.2: 24822606-24822606 on Build GRCh38
rs767933376 Chr.2: 24822614-24822614 on Build GRCh38
rs371465590 Chr.2: 24822608-24822608 on Build GRCh38
rs776650898 Chr.2: 24822574-24822574 on Build GRCh38
rs990811311 Chr.2: 24822848-24822848 on Build GRCh38
rs146622426 Chr.2: 24822565-24822565 on Build GRCh38
rs1031359436 Chr.2: 24822577-24822577 on Build GRCh38
rs79421293 Chr.2: 24822544-24822544 on Build GRCh38
rs770197713 Chr.2: 24822598-24822598 on Build GRCh38
rs755754493 Chr.2: 24822633-24822633 on Build GRCh38
rs961475577 Chr.2: 24822766-24822766 on Build GRCh38
rs189352896 Chr.2: 24822462-24822462 on Build GRCh38
rs776942776 Chr.2: 24822481-24822481 on Build GRCh38
rs147179390 Chr.2: 24822514-24822514 on Build GRCh38
rs913689877 Chr.2: 24822839-24822839 on Build GRCh38
rs962438707 Chr.2: 24822687-24822687 on Build GRCh38
rs953983877 Chr.2: 24822706-24822707 on Build GRCh38
rs763538939 Chr.2: 24822497-24822497 on Build GRCh38
rs774782285 Chr.2: 24822756-24822756 on Build GRCh38
rs748857384 Chr.2: 24822535-24822535 on Build GRCh38
rs746848833 Chr.2: 24822658-24822658 on Build GRCh38
rs763435563 Chr.2: 24822673-24822673 on Build GRCh38
rs751836891 Chr.2: 24822505-24822505 on Build GRCh38
rs972126728 Chr.2: 24822715-24822715 on Build GRCh38
rs776095938 Chr.2: 24822650-24822652 on Build GRCh38
rs931414926 Chr.2: 24822824-24822824 on Build GRCh38
rs752403966 Chr.2: 24822529-24822529 on Build GRCh38
rs986348988 Chr.2: 24822527-24822527 on Build GRCh38
rs776999333 Chr.2: 24822471-24822471 on Build GRCh38
rs144282882 Chr.2: 24822886-24822886 on Build GRCh38
rs377745631 Chr.2: 24822575-24822575 on Build GRCh38
rs892876643 Chr.2: 24822895-24822895 on Build GRCh38
rs566079421 Chr.2: 24822744-24822744 on Build GRCh38
rs916608835 Chr.2: 24822859-24822860 on Build GRCh38
rs920023886 Chr.2: 24822504-24822504 on Build GRCh38
rs987081359 Chr.2: 24822719-24822719 on Build GRCh38
rs924235619 Chr.2: 24822874-24822874 on Build GRCh38
rs759422318 Chr.2: 24822470-24822470 on Build GRCh38
rs761750343 Chr.2: 24822482-24822482 on Build GRCh38
rs773596441 Chr.2: 24822669-24822669 on Build GRCh38
rs144045466 Chr.2: 24822714-24822714 on Build GRCh38
rs944882532 Chr.2: 24822796-24822796 on Build GRCh38
rs755558582 Chr.2: 24822531-24822531 on Build GRCh38
rs76847777 Chr.2: 24822648-24822648 on Build GRCh38
rs116997258 Chr.2: 24822892-24822892 on Build GRCh38
rs527464781 Chr.2: 24822745-24822745 on Build GRCh38
rs547864204 Chr.2: 24822685-24822685 on Build GRCh38
rs770835810 Chr.2: 24822541-24822541 on Build GRCh38
rs769788756 Chr.2: 24822546-24822546 on Build GRCh38
rs976638672 Chr.2: 24822827-24822827 on Build GRCh38
rs538106284 Chr.2: 24822869-24822869 on Build GRCh38
rs374537031 Chr.2: 24822663-24822663 on Build GRCh38
rs1013824059 Chr.2: 24822662-24822662 on Build GRCh38
rs1040135012 Chr.2: 24822536-24822536 on Build GRCh38
rs367794544 Chr.2: 24822671-24822671 on Build GRCh38

View Full Product Details