Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24827468-24827974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY3
Entrez Gene ID
109
Gene Name
adenylate cyclase 3
Gene Aliases
AC-III, AC3
UniGene
Hs.467898
Gene Chromosome Location
Chr.2: 24819169-24920017 on Build GRCh38
Transcripts
NM_001320613.1 NM_004036.4 XM_006711925.1 XM_011532489.1 XM_011532490.1 XM_011532491.1 XM_011532492.2 XM_011532493.1 XM_011532494.1 XM_011532496.1 XM_017003186.1 XM_017003187.1 XM_017003188.1 XM_017003189.1 XM_017003190.1 XM_017003191.1 XM_017003192.1 XM_017003193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4618216 c.2406C>T p.Y802Y Chr.2: 24827928-24827928 on Build GRCh38
4772940 c.2442G>T p.M814I Chr.2: 24827599-24827599 on Build GRCh38
1019192 c.2454G>A p.E818E Chr.2: 24827587-24827587 on Build GRCh38
4979480 c.2479C>T p.L827F Chr.2: 24827562-24827562 on Build GRCh38
6594911 c.2391C>T p.P797P Chr.2: 24827943-24827943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747701709 Chr.2: 24827574-24827574 on Build GRCh38
rs921339565 Chr.2: 24827491-24827491 on Build GRCh38
rs1045465172 Chr.2: 24827714-24827714 on Build GRCh38
rs759333413 Chr.2: 24827857-24827857 on Build GRCh38
rs573075609 Chr.2: 24827577-24827577 on Build GRCh38
rs146269706 Chr.2: 24827852-24827852 on Build GRCh38
rs116538472 Chr.2: 24827538-24827538 on Build GRCh38
rs867833762 Chr.2: 24827749-24827749 on Build GRCh38
rs775511537 Chr.2: 24827595-24827595 on Build GRCh38
rs897804782 Chr.2: 24827495-24827495 on Build GRCh38
rs750787613 Chr.2: 24827504-24827504 on Build GRCh38
rs577695729 Chr.2: 24827699-24827699 on Build GRCh38
rs1045815907 Chr.2: 24827828-24827828 on Build GRCh38
rs775904787 Chr.2: 24827945-24827945 on Build GRCh38
rs201882277 Chr.2: 24827535-24827535 on Build GRCh38
rs538748727 Chr.2: 24827731-24827731 on Build GRCh38
rs767110396 Chr.2: 24827885-24827885 on Build GRCh38
rs565303281 Chr.2: 24827889-24827889 on Build GRCh38
rs937876218 Chr.2: 24827735-24827735 on Build GRCh38
rs759846027 Chr.2: 24827880-24827880 on Build GRCh38
rs112239820 Chr.2: 24827631-24827631 on Build GRCh38
rs765788830 Chr.2: 24827628-24827628 on Build GRCh38
rs749363565 Chr.2: 24827927-24827927 on Build GRCh38
rs774738640 Chr.2: 24827937-24827937 on Build GRCh38
rs751923237 Chr.2: 24827647-24827647 on Build GRCh38
rs1045870709 Chr.2: 24827952-24827952 on Build GRCh38
rs952334048 Chr.2: 24827532-24827532 on Build GRCh38
rs201034328 Chr.2: 24827870-24827870 on Build GRCh38
rs753347971 Chr.2: 24827903-24827903 on Build GRCh38
rs532463063 Chr.2: 24827892-24827892 on Build GRCh38
rs755796043 Chr.2: 24827686-24827687 on Build GRCh38
rs752377174 Chr.2: 24827663-24827663 on Build GRCh38
rs374433056 Chr.2: 24827638-24827638 on Build GRCh38
rs773867955 Chr.2: 24827630-24827630 on Build GRCh38
rs776831636 Chr.2: 24827604-24827604 on Build GRCh38
rs777721834 Chr.2: 24827580-24827580 on Build GRCh38
rs945015831 Chr.2: 24827696-24827696 on Build GRCh38
rs763500351 Chr.2: 24827876-24827876 on Build GRCh38
rs752824508 Chr.2: 24827867-24827870 on Build GRCh38
rs777026103 Chr.2: 24827865-24827865 on Build GRCh38
rs767194206 Chr.2: 24827652-24827652 on Build GRCh38
rs377506465 Chr.2: 24827928-24827928 on Build GRCh38
rs1037666380 Chr.2: 24827557-24827557 on Build GRCh38
rs760728863 Chr.2: 24827569-24827569 on Build GRCh38
rs373035912 Chr.2: 24827918-24827918 on Build GRCh38
rs372148762 Chr.2: 24827853-24827853 on Build GRCh38
rs897855322 Chr.2: 24827831-24827831 on Build GRCh38
rs542868330 Chr.2: 24827822-24827822 on Build GRCh38
rs764980386 Chr.2: 24827872-24827872 on Build GRCh38
rs12105415 Chr.2: 24827771-24827771 on Build GRCh38
rs556210357 Chr.2: 24827643-24827643 on Build GRCh38
rs770775631 Chr.2: 24827591-24827591 on Build GRCh38
rs752159446 Chr.2: 24827656-24827656 on Build GRCh38
rs766756310 Chr.2: 24827497-24827499 on Build GRCh38
rs767804097 Chr.2: 24827862-24827864 on Build GRCh38
rs770197739 Chr.2: 24827920-24827920 on Build GRCh38
rs925205210 Chr.2: 24827732-24827732 on Build GRCh38
rs543479985 Chr.2: 24827863-24827863 on Build GRCh38
rs778757630 Chr.2: 24827906-24827906 on Build GRCh38
rs773315426 Chr.2: 24827922-24827922 on Build GRCh38
rs369803323 Chr.2: 24827942-24827942 on Build GRCh38
rs758279809 Chr.2: 24827912-24827912 on Build GRCh38
rs975767145 Chr.2: 24827545-24827545 on Build GRCh38
rs754633404 Chr.2: 24827651-24827651 on Build GRCh38
rs993760930 Chr.2: 24827509-24827509 on Build GRCh38
rs1028116101 Chr.2: 24827824-24827824 on Build GRCh38
rs768818963 Chr.2: 24827600-24827600 on Build GRCh38
rs772433588 Chr.2: 24827943-24827943 on Build GRCh38
rs758812918 Chr.2: 24827520-24827520 on Build GRCh38
rs1027891771 Chr.2: 24827531-24827531 on Build GRCh38
rs913589265 Chr.2: 24827621-24827621 on Build GRCh38
rs748990847 Chr.2: 24827587-24827587 on Build GRCh38
rs757211680 Chr.2: 24827856-24827856 on Build GRCh38
rs991120544 Chr.2: 24827941-24827941 on Build GRCh38
rs576369380 Chr.2: 24827854-24827854 on Build GRCh38
rs190125602 Chr.2: 24827948-24827948 on Build GRCh38
rs766839931 Chr.2: 24827877-24827877 on Build GRCh38
rs551713709 Chr.2: 24827748-24827748 on Build GRCh38
rs572315516 Chr.2: 24827796-24827796 on Build GRCh38
rs762026508 Chr.2: 24827616-24827616 on Build GRCh38
rs185180064 Chr.2: 24827917-24827917 on Build GRCh38
rs751226954 Chr.2: 24827534-24827534 on Build GRCh38
rs560339786 Chr.2: 24827911-24827911 on Build GRCh38
rs934311006 Chr.2: 24827922-24827924 on Build GRCh38
rs372560995 Chr.2: 24827947-24827947 on Build GRCh38
rs547084770 Chr.2: 24827567-24827567 on Build GRCh38
rs779512665 Chr.2: 24827528-24827528 on Build GRCh38
rs373053710 Chr.2: 24827869-24827869 on Build GRCh38
rs755558661 Chr.2: 24827890-24827890 on Build GRCh38
rs139420185 Chr.2: 24827904-24827904 on Build GRCh38
rs1031991377 Chr.2: 24827736-24827739 on Build GRCh38
rs767432609 Chr.2: 24827703-24827704 on Build GRCh38
rs760947703 Chr.2: 24827598-24827598 on Build GRCh38
rs774744041 Chr.2: 24827878-24827878 on Build GRCh38

View Full Product Details