Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26931322-26931725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL5
Entrez Gene ID
56896
Gene Name
dihydropyrimidinase like 5
Gene Aliases
CRAM, CRMP-5, CRMP5, Ulip6
UniGene
Hs.299315
Gene Chromosome Location
Chr.2: 26848101-26950351 on Build GRCh38
Transcripts
NM_001253723.1 NM_001253724.1 NM_020134.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6649179 c.692G>A p.R231H Chr.2: 26931662-26931662 on Build GRCh38
5894253 c.714+7C>T p.? Chr.2: 26931691-26931691 on Build GRCh38
7365441 c.682G>T p.A228S Chr.2: 26931652-26931652 on Build GRCh38
442722 c.691C>T p.R231C Chr.2: 26931661-26931661 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771627088 Chr.2: 26931698-26931698 on Build GRCh38
rs766460213 Chr.2: 26931632-26931632 on Build GRCh38
rs758987705 Chr.2: 26931591-26931591 on Build GRCh38
rs908059450 Chr.2: 26931493-26931493 on Build GRCh38
rs773922062 Chr.2: 26931598-26931600 on Build GRCh38
rs79263793 Chr.2: 26931379-26931379 on Build GRCh38
rs914195826 Chr.2: 26931343-26931343 on Build GRCh38
rs956588619 Chr.2: 26931369-26931369 on Build GRCh38
rs1010507699 Chr.2: 26931582-26931582 on Build GRCh38
rs909849829 Chr.2: 26931533-26931533 on Build GRCh38
rs530348832 Chr.2: 26931527-26931527 on Build GRCh38
rs199857288 Chr.2: 26931623-26931623 on Build GRCh38
rs1030893032 Chr.2: 26931358-26931358 on Build GRCh38
rs753542089 Chr.2: 26931622-26931622 on Build GRCh38
rs760592481 Chr.2: 26931610-26931610 on Build GRCh38
rs753318314 Chr.2: 26931678-26931678 on Build GRCh38
rs979727372 Chr.2: 26931602-26931602 on Build GRCh38
rs1021961011 Chr.2: 26931419-26931419 on Build GRCh38
rs965551756 Chr.2: 26931512-26931512 on Build GRCh38
rs891640660 Chr.2: 26931558-26931558 on Build GRCh38
rs751675163 Chr.2: 26931640-26931640 on Build GRCh38
rs899270153 Chr.2: 26931522-26931522 on Build GRCh38
rs1008103362 Chr.2: 26931545-26931545 on Build GRCh38
rs542082136 Chr.2: 26931532-26931532 on Build GRCh38
rs367898236 Chr.2: 26931445-26931445 on Build GRCh38
rs996189652 Chr.2: 26931529-26931529 on Build GRCh38
rs796195823 Chr.2: 26931531-26931531 on Build GRCh38
rs546270376 Chr.2: 26931544-26931544 on Build GRCh38
rs773944952 Chr.2: 26931590-26931590 on Build GRCh38
rs372183731 Chr.2: 26931568-26931568 on Build GRCh38
rs149150079 Chr.2: 26931619-26931619 on Build GRCh38
rs72804841 Chr.2: 26931553-26931553 on Build GRCh38
rs373933041 Chr.2: 26931391-26931391 on Build GRCh38
rs778360884 Chr.2: 26931691-26931691 on Build GRCh38
rs145194693 Chr.2: 26931483-26931483 on Build GRCh38
rs12463779 Chr.2: 26931469-26931469 on Build GRCh38
rs1003881932 Chr.2: 26931355-26931355 on Build GRCh38
rs892419755 Chr.2: 26931696-26931696 on Build GRCh38
rs72851870 Chr.2: 26931699-26931699 on Build GRCh38
rs759193797 Chr.2: 26931691-26931692 on Build GRCh38
rs199608212 Chr.2: 26931600-26931600 on Build GRCh38
rs749576469 Chr.2: 26931682-26931682 on Build GRCh38
rs1037778743 Chr.2: 26931520-26931520 on Build GRCh38
rs371775143 Chr.2: 26931446-26931446 on Build GRCh38
rs72851869 Chr.2: 26931519-26931519 on Build GRCh38
rs376628981 Chr.2: 26931595-26931595 on Build GRCh38
rs745513904 Chr.2: 26931695-26931695 on Build GRCh38
rs756667382 Chr.2: 26931681-26931681 on Build GRCh38
rs755044882 Chr.2: 26931666-26931666 on Build GRCh38
rs906375698 Chr.2: 26931569-26931569 on Build GRCh38
rs760677606 Chr.2: 26931356-26931356 on Build GRCh38
rs573831826 Chr.2: 26931432-26931432 on Build GRCh38
rs781421525 Chr.2: 26931674-26931674 on Build GRCh38
rs528653729 Chr.2: 26931576-26931576 on Build GRCh38
rs770640809 Chr.2: 26931684-26931684 on Build GRCh38
rs968393398 Chr.2: 26931374-26931374 on Build GRCh38

View Full Product Details